Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Endothelin A Receptor Antibodies

          Invitrogen

          Endothelin A Receptor Polyclonal Antibody

          Advanced Verification
          5 Published Figures
          13 References
          View all (9) Endothelin A Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Endothelin A Receptor Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          • Published Figures (5)
          • Advanced Verification (1)
          Endothelin A Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Endothelin A Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Endothelin A Receptor Antibody (PA3-065) in ICC/IF

          Immunofluorescent analysis of Endothelin A Receptor in ETA-transfected cells using a polyclonal antibody (Product # PA3-065). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Endothelin A Receptor Antibody in Immunocytochemistry (ICC/IF)
          Endothelin A Receptor Antibody in Immunohistochemistry (IHC)
          Endothelin A Receptor Antibody in Immunohistochemistry (IHC)
          Endothelin A Receptor Antibody in Western Blot (WB)
          Endothelin A Receptor Antibody in Western Blot (WB)
          Endothelin A Receptor Antibody in Multiplex Immunohistochemistry (mIHC)
          Endothelin A Receptor Antibody in Western Blot (WB)
          Endothelin A Receptor Antibody in Western Blot (WB)
          Endothelin A Receptor Antibody in Immunocytochemistry (ICC/IF)
          Endothelin A Receptor Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Endothelin A Receptor Antibody in Western Blot (WB)
          Endothelin A Receptor Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Endothelin A Receptor Polyclonal Antibody

          Product Details

          PA3-065

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:5,000
          View 7 publications 7 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Multiplex Immunohistochemistry (mIHC)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          1:500-1:2,000
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:5,000
          View 1 publication 1 publication

          Immunoelectrophoresis (IE)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          COOH-terminal amino acids of mouse ETA (408-427) KNQEQNNHNTERSSHKDSMN coupled to KLH
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11153077

          Product Specific Information

          PA3-065 detects Endothelin Receptor A from human, mouse, and rat samples.



          PA3-065 has been successfully used in Western blot, immunocytochemistry, and immunohistochemistry procedures. By western blot, this antibody detects a 46kDa protein corresponding to mouse Endothelin Receptor A.

          Target Information

          This gene encodes the receptor for endothelin-1, a peptide that plays a role in potent and long-lasting vasoconstriction. This receptor associates with guanine-nucleotide-binding (G) proteins, and this coupling activates a phosphatidylinositol-calcium second messenger system. Polymorphisms in this gene have been linked to migraine headache resistance. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Endothelin A; endothelin A receptor; endothelin receptor subtype A; endothelin-1 receptor-like; endothelin-1-specific receptor; ET-A; ET-AR; ET-AR delta 3-4; ET-AR delta 4; ETAR-L; ETAR-U3; ETAR-U8; G protein-coupled receptor; G-protein coupled receptor; G-protein coupled receptor 10; unnamed protein product

          View more View less

          Gene Aliases: AEA001; Endor; ET-A; ET-AR; ETA; ETA-R; ETAR; ETRA; Gpcr10; hET-AR; MFDA; Mhdaaea1; RGD1559432

          View more View less

          UniProt ID: (Human) P25101, (Mouse) Q61614, (Rat) P26684

          View more View less

          Entrez Gene ID: (Human) 1909, (Mouse) 13617, (Rat) 24326

          View more View less

          Function(s)
          phosphatidylinositol phospholipase C activity G-protein coupled receptor activity endothelin receptor activity protein binding
          Process(es)
          mitotic cell cycle angiogenesis patterning of blood vessels response to hypoxia in utero embryonic development histamine secretion positive regulation of protein phosphorylation blood vessel remodeling response to amphetamine regulation of heart rate heart process glomerular filtration cardiac chamber formation left ventricular cardiac muscle tissue morphogenesis atrial cardiac muscle tissue development cardiac neural crest cell migration involved in outflow tract morphogenesis noradrenergic neuron differentiation protein phosphorylation cellular calcium ion homeostasis mitochondrion organization G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration Rho protein signal transduction specification of segmental identity, maxillary segment axon guidance heart development respiratory gaseous exchange blood circulation regulation of blood pressure positive regulation of cell proliferation response to wounding gene expression regulation of glucose transport neural crest cell development neural crest cell differentiation neural crest cell fate commitment artery smooth muscle contraction neuron remodeling heparin metabolic process thyroid gland development neuron projection development response to lipopolysaccharide glomerulus development cellular response to oxidative stress embryonic heart tube development aorta development specification of segmental identity, mandibular segment vasoconstriction norepinephrine metabolic process middle ear morphogenesis positive regulation of odontogenesis negative regulation of apoptotic process cellular response to human chorionic gonadotropin stimulus developmental pigmentation fibroblast proliferation enteric nervous system development sympathetic nervous system development smooth muscle cell proliferation axon extension embryonic skeletal system development regulation of epithelial cell proliferation neuromuscular process positive regulation of release of sequestered calcium ion into cytosol positive regulation of calcium ion transport sodium ion homeostasis canonical Wnt signaling pathway head development face development axonogenesis involved in innervation establishment of endothelial barrier pharyngeal arch artery morphogenesis renal sodium ion absorption cellular response to hydrogen peroxide positive regulation of ERK1 and ERK2 cascade calcium ion transmembrane transport cellular response to mechanical stimulus cellular response to follicle-stimulating hormone stimulus cellular response to luteinizing hormone stimulus semaphorin-plexin signaling pathway protein transmembrane transport glomerular endothelium development glomerular visceral epithelial cell differentiation endothelin receptor signaling pathway positive regulation of neutrophil chemotaxis positive regulation of kidney development renal albumin absorption vascular smooth muscle cell development mesenchymal cell apoptotic process cellular response to toxic substance sympathetic neuron axon guidance cAMP/PKA signal transduction glomerular visceral epithelial cell apoptotic process meiotic cell cycle process involved in oocyte maturation cranial skeletal system development response to acetylcholine regulation of protein localization to cell leading edge smooth muscle contraction signal transduction activation of adenylate cyclase activity cell proliferation vascular smooth muscle contraction renal system process maternal process involved in parturition
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch