Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PPAR gamma Antibodies

          Invitrogen

          PPAR gamma Polyclonal Antibody

          Advanced Verification
          23 Published Figures
          40 References
          View all (34) PPAR gamma antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PPAR gamma Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (8)
          • Published Figures (23)
          • Advanced Verification (2)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 33

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PPAR gamma Antibody (PA3-821A) in ICC/IF

          Immunofluorescence analysis of PPAR Gamma was done on 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with PPAR gamma Rabbit Polyclonal Antibody (Product # PA3-821A) at 1:250 dilution in 0.1% BSA and incubated for 3 hours at r... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Immunocytochemistry (ICC/IF)
          PPAR gamma Antibody in Immunohistochemistry (IHC)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Western Blot (WB)
          PPAR gamma Antibody in Flow Cytometry (Flow)
          PPAR gamma Antibody in Flow Cytometry (Flow)
          PPAR gamma Antibody in Flow Cytometry (Flow)
          PPAR gamma Antibody in Immunoprecipitation, ChIP Assay (IP, ChIP)
          PPAR gamma Antibody in ChIP Assay (ChIP)
          PPAR gamma Antibody in ChIP Assay (ChIP)
          PPAR gamma Antibody
          PPAR gamma Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PPAR gamma Polyclonal Antibody

          Product Details

          PA3-821A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,500
          View 21 publications 21 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:250
          View 4 publications 4 publications

          Flow Cytometry (Flow)

          -
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          ChIP assay (ChIP)

          -
          View 3 publications 3 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Gel Shift (EMSA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues M(284) M G E D K I K F K H I T P L(298) of mouse PPAR gamma 2.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2166056

          Product Specific Information

          PA3-821A detects peroxisome proliferator activated receptor (PPAR) gamma from human and mouse samples. This antibody is a reproduction of PA3-821, using the same immunizing peptide and new host rabbits.

          PA3-821A has been successfully used in IF and Western blot procedures. Previous batches of this antibody have been used in gel shift, immunocytochemistry, and immunofluoresence procedures. By Western blot, this antibody detects a 57 kDa protein representing PPAR gamma 2 from differentiated mouse 3T3-L1 cell lysate. Immunolocalization of previous batches gives predominant nuclear staining.

          The PA3-821A immunizing peptide corresponds to amino acid residues 284-298 from mouse PPAR gamma 2. This sequence is completely conserved in PPAR gamma 1, but exhibits no significant homology with PPAR alpha or NUC1. This sequence is completely conserved in mouse, human, and rat.

          Target Information

          This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: HUMPPARG; ligand-activated nuclear transcription factor; ligand-activated transcription factor; nuclear receptor; nuclear receptor family member; Pparg; nuclear receptor subfamily 1 group C member 3; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated receptor-gamma 5; peroxisome proliferator-activated receptor-gamma splicing; PPAR gamma; PPAR-gamma; PPAR-gamma-1a; PPAR-gamma-1b; Pparg; PPARy; unnamed protein product

          View more View less

          Gene Aliases: CIMT1; FPLD3; GLM1; NR1C3; PPAR-gamma; PPAR-gamma2; PPARG1; PPARG2; PPARG5; PPARgamma; PPARgamma2

          View more View less

          UniProt ID: (Human) P37231, (Rat) O88275, (Mouse) P37238

          View more View less

          Entrez Gene ID: (Human) 5468, (Rat) 25664, (Mouse) 19016

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription cofactor binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding nucleic acid binding DNA binding chromatin binding double-stranded DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding prostaglandin receptor activity protein binding zinc ion binding enzyme binding peptide binding identical protein binding sequence-specific DNA binding retinoid X receptor binding arachidonic acid binding DBD domain binding LBD domain binding WW domain binding alpha-actinin binding R-SMAD binding E-box binding STAT family protein binding DNA-binding transcription factor binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding transcription coactivator binding protein phosphatase binding estrogen receptor binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter negative regulation of cytokine production placenta development response to dietary excess diet induced thermogenesis wound healing involved in inflammatory response negative regulation of acute inflammatory response regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter fatty acid metabolic process response to stress heart development regulation of blood pressure negative regulation of cell proliferation response to xenobiotic stimulus response to light stimulus response to mechanical stimulus hormone-mediated signaling pathway regulation of gene expression negative regulation of cardiac muscle hypertrophy positive regulation of gene expression negative regulation of gene expression negative regulation of macrophage derived foam cell differentiation positive regulation of cholesterol efflux negative regulation of cholesterol storage negative regulation of lipid storage negative regulation of smooth muscle cell migration long-chain fatty acid transport negative regulation of angiogenesis regulation of lipid metabolic process fatty acid oxidation cell differentiation monocyte differentiation negative regulation of cell growth BMP signaling pathway negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of BMP signaling pathway intracellular receptor signaling pathway epithelial cell differentiation response to caffeine organ regeneration positive regulation of prostaglandin biosynthetic process response to nutrient levels response to food positive regulation of cholesterol transport positive regulation of intracellular cholesterol transport cellular response to insulin stimulus cellular response to hormone stimulus negative regulation of collagen biosynthetic process response to vitamin A response to lipid peroxisome proliferator activated receptor signaling pathway response to immobilization stress cellular response to platelet-derived growth factor stimulus glucose homeostasis response to starvation regulation of circadian rhythm mRNA transcription from RNA polymerase II promoter positive regulation of apoptotic process negative regulation of MAPK cascade negative regulation of blood vessel endothelial cell migration response to estrogen cell fate commitment fat cell differentiation regulation of fat cell differentiation positive regulation of fat cell differentiation negative regulation of osteoblast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of fatty acid metabolic process positive regulation of transcription from RNA polymerase II promoter positive regulation of fatty acid oxidation regulation of hormone secretion retinoic acid receptor signaling pathway cell maturation negative regulation of smooth muscle cell proliferation positive regulation of oligodendrocyte differentiation positive regulation of protein secretion negative regulation of inflammatory response white fat cell differentiation brown fat cell differentiation positive regulation of phagocytosis, engulfment negative regulation of interferon-gamma-mediated signaling pathway positive regulation of SMAD protein import into nucleus negative regulation of SMAD protein import into nucleus positive regulation of adiponectin secretion response to linoleic acid cellular response to lithium ion cellular response to retinoic acid cellular response to vitamin E cellular response to prostaglandin stimulus cellular response to prostaglandin E stimulus cellular response to low-density lipoprotein particle stimulus cellular response to hyperoxia cellular response to hypoxia negative regulation of mitochondrial fission negative regulation of peptide hormone secretion cold-induced thermogenesis positive regulation of lipoprotein transport beige fat cell differentiation negative regulation of cellular response to insulin stimulus negative regulation of extracellular matrix assembly negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter regulation of protein localization to plasma membrane negative regulation of cardiac muscle hypertrophy in response to stress negative regulation of cellular response to transforming growth factor beta stimulus positive regulation of adipose tissue development negative regulation of connective tissue replacement involved in inflammatory response wound healing negative regulation of vascular smooth muscle cell proliferation negative regulation of receptor signaling pathway via STAT positive regulation of vascular associated smooth muscle cell apoptotic process negative regulation of vascular endothelial cell proliferation negative regulation of pancreatic stellate cell proliferation positive regulation of apoptotic signaling pathway G-protein coupled receptor signaling pathway response to nutrient innate immune response positive regulation of lipid metabolic process regulation of transport negative regulation of multicellular organismal process lipid homeostasis nuclear receptor-mediated signaling pathway regulation of cellular response to insulin stimulus inflammatory response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch