Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: TEKVAVDPET VFKRPRECDS PSYQKRQRMA LLARKQGAGD SLIAGSAMSK
Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.
CT45A3 (Cancer/Testis Antigen Family 45 Member A3) is a Protein Coding gene. An important paralog of this gene is CT45A1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 45-1; cancer/testis antigen 45-3; cancer/testis antigen 45-4; cancer/testis antigen 45A1; cancer/testis antigen 45A3; cancer/testis antigen 45A4; cancer/testis antigen CT45-1; cancer/testis antigen CT45-3; cancer/testis antigen family 45 member A4; unnamed protein product
Gene Aliases: CT45; CT45-1; CT45-3; CT45-4; CT45.1; CT45.3; CT45.4; CT45.5; CT45.6; CT455; CT45A4
UniProt ID: (Human) Q5HYN5, (Human) P0DMU8, (Human) Q8NHU0, (Human) P0DMU7
Entrez Gene ID: (Human) 541466, (Human) 102723631, (Human) 441519, (Human) 441521, (Human) 541465
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support