Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KGEAKLLKDG KVFRVVRENC WDPEVRIREM DQKGVTVQAL STVPVMFSYW AKPEDTLNLC QLLNNDLAST VVSYPRRFVG LGTLPMQAPE LAVKEMERCV KELGFPGVQI GTHVNEWDLN AQELFPVYAA AERLKCSLFV HPW
Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 72%.
The neuronal excitotoxin quinolinate is an intermediate in the de novo synthesis pathway of NAD from tryptophan, and has been implicated in the pathogenesis of several neurodegenerative disorders. Quinolinate is derived from alpha-amino-beta-carboxy-muconate-epsilon-semialdehyde (ACMS). ACMSD (ACMS decarboxylase; EC 4.1.1.45) can divert ACMS to a benign catabolite and thus prevent the accumulation of quinolinate from ACMS.[supplied by OMIM, Oct 2004]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: picolinate carboxylase; unnamed protein product
UniProt ID: (Human) Q8TDX5
Entrez Gene ID: (Human) 130013
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support