Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KLKEDSIPAP RSVGSIKINF TPRVFPTALR ESQVAEEEEW LHKQAEARRA MNTDIAELCD LKEEEKNPEW LKDKGNKLFA TENYLAAINA
Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 87%.
This gene encodes a tetratricopeptide repeat domain-containing protein. The encoded protein interacts with estrogen receptors and the heat shock proteins, Hsp70 and Hsp90. A chromosomal translocation involving this gene is associated with a susceptibility to developmental dyslexia. Mutations in this gene are associated with deficits in reading and spelling. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dynein assembly factor 4, axonemal; dyslexia susceptibility 1 candidate 1; dyslexia susceptibility 1 candidate gene 1 protein; FLJ37882; MGC70618; unnamed protein product
Gene Aliases: CILD25; DYX1; DYX1C1; DYXC1; EKN1; pf23; RD
UniProt ID: (Human) Q8WXU2
Entrez Gene ID: (Human) 161582
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support