Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: KERDHISERR ERERSTSMRK SSNDRDGKEK LEKNSTSLKE KEHNKEPDSS VSKEVDDKDA PRTEENKIQH NGNCQLNEEN LSTKTEAV
Highest antigen sequence indentity to the following orthologs: Rat - 69%, Mouse - 69%.
SFRS12 belongs to the superfamily of serine/arginine-rich splicing factors. It modulates splice site selection by regulating the activities of other SR proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: serine-arginine-rich splicing regulatory protein 508; serine/arginine-rich-splicing regulatory protein 86; splicing factor, arginine/serine-rich 12; splicing regulatory glutamic acid/lysine-rich protein 1; unnamed protein product
Gene Aliases: SFRS12; SRrp508; SRrp86
UniProt ID: (Human) Q8WXA9
Entrez Gene ID: (Human) 140890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support