Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Clathrin Light Chain Antibodies

          Invitrogen

          Clathrin Light Chain Polyclonal Antibody

          View all (8) Clathrin Light Chain antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Clathrin Light Chain Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (20)
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Clathrin Light Chain Antibody (PA5-100037) in IHC (P)

          Immunohistochemistry analysis of Clathrin Light Chain in human normal tissues adjacent to colorectal cancer. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with Clathrin Light Chain polyclonal antibody (Product # PA5-100037) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Clathrin Light Chain Antibody in Western Blot (WB)
          Clathrin Light Chain Antibody in Western Blot (WB)
          Clathrin Light Chain Antibody in Western Blot (WB)
          Clathrin Light Chain Polyclonal Antibody

          Product Details

          PA5-100037

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CLTA(Accession P09496), corresponding to amino acid residues G58-E108.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2815567

          Product Specific Information

          Antibody detects endogenous levels of total CLTA.

          Target Information

          Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CHC17; Clathrin light chain A; Clathrin light chain B; clathrin light-chain A; clathrin light-chain B; clathrin, light chain (Lca); clathrin, light chain (Lcb); clathrin, light polypeptide (Lca); clathrin, light polypeptide (Lcb); clathryn light chain (LCA2); CLH17; CLTA; CLTB; CLTC; CTLA; CTLB; KIAA0034; Lca; Lcb; unnamed protein product

          View more View less

          Gene Aliases: 2310046E19Rik; AV026556; CLTA; CLTB; LCA; LCA1; LCA2; LCA3; LCB

          View more View less

          UniProt ID: (Human) P09496, (Human) P09497, (Rat) P08081, (Rat) P08082, (Mouse) Q6IRU5

          View more View less

          Entrez Gene ID: (Human) 1211, (Human) 1212, (Mouse) 12757, (Rat) 83800, (Rat) 116561, (Mouse) 74325

          View more View less

          Function(s)
          structural molecule activity protein binding clathrin heavy chain binding peptide binding macromolecular complex binding GTPase binding neurotransmitter transporter activity vesicle coat protein
          Process(es)
          intracellular protein transport endocytosis vesicle-mediated transport clathrin coat assembly synaptic vesicle endocytosis cell division clathrin-mediated endocytosis neurotransmitter transport neurotransmitter secretion synaptic vesicle coating biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-hn5gs:80/100.66.79.229:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline