Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NLRP3 Antibodies

          Invitrogen

          Phospho-NLRP3 (Ser295) Polyclonal Antibody

          3 Published Figures
          5 References
          View all (38) NLRP3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-NLRP3 (Ser295) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (3)
          Phospho-NLRP3 (Ser295) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-NLRP3 (Ser295) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-NLRP3 (Ser295) Antibody (PA5-105071) in IHC (P)

          Immunohistochemistry analysis of Phospho-NLRP3 (Ser295) in human colorectal cancer. The sample was formaldehyde fixed and a heat mediated antigen retrieval step in citrate buffer was performed. Samples were incubated with Phospho-NLRP3 (Ser295) polyclonal antibody (Product # PA5-105071) using a dilution of 1:100 (4°C overnight) followed by HRP conjugated anti-Rabbit secondary antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-NLRP3 (Ser295) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-NLRP3 (Ser295) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-NLRP3 (Ser295) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-NLRP3 (Ser295) Antibody in Western Blot (WB)
          Phospho-NLRP3 (Ser295) Antibody in Western Blot (WB)
          Phospho-NLRP3 (Ser295) Antibody in Western Blot (WB)
          Phospho-NLRP3 (Ser295) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-NLRP3 (Ser295) Polyclonal Antibody

          Product Details

          PA5-105071

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human NLRP3(Accession Q96P20), corresponding to amino acid residues around phosphorylated Ser295.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816544

          Product Specific Information

          Antibody detects endogenous levels of NLRP3 only when phosphorylated at Ser295.

          Target Information

          This gene encodes a pyrin-like protein containing a pyrin domain, a nucleotide-binding site domain, and a leucine-rich repeat motif. This protein interacts with the apoptosis-associated speck-like protein PYCARD/ASC, which contains a caspase recruitment domain, and is a member of the NALP3 inflammasome complex. This complex functions as an upstream activator of NF-kappaB signaling, and it plays a role in the regulation of inflammation, the immune response, and apoptosis. Mutations in this gene are associated with familial cold autoinflammatory syndrome, and neonatal-onset multisystem inflammatory disease. Multiple alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene. Alternative 5' UTR structures are suggested by available data; however, insufficient evidence is available to determine if all of the represented 5' UTR splice patterns are biologically valid.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: caterpiller protein 1.1; cold auto-inflammatory syndrome 1; PYPAF1; NALP3; C1ORF7; cold autoinflammatory syndrome 1 protein; cold autoinflammatory syndrome 1 protein homolog; cold-induced autoinflammatory syndrome 1 protein; cryopyrin; cryopyrin, NACHT, LRR and PYD domains - containing protein 3; deafness, autosomal dominant 34; FLJ95925; mast cell maturation-associated-inducible protein 1; member of the NALP family of inflammatory mediators; MMIG-1; NACHT domain-, leucine-rich repeat-, and PYD-containing protein 3; NACHT, LRR and PYD containing protein 3; NACHT/LRR/pyrin domain-containing protein 3; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 3; PYD, NACHT, and LRR containing protein; PYRIN-containing APAF1-like protein 1; unnamed protein product

          View more View less

          Gene Aliases: AGTAVPRL; AII; AII/AVP; AVP; C1orf7; CIAS1; CLR1.1; DFNA34; FCAS; FCAS1; FCU; KEFH; Mmig1; MWS; NALP3; PYPAF1

          View more View less

          UniProt ID: (Human) Q96P20, (Mouse) Q8R4B8

          View more View less

          Entrez Gene ID: (Human) 114548, (Mouse) 216799

          View more View less

          Function(s)
          protein binding ATP binding ATPase activity protein binding, bridging signaling adaptor activity identical protein binding peptidoglycan binding ADP binding sequence-specific DNA binding binding, bridging phosphatidylinositol-4-phosphate binding DNA-binding transcription factor binding small molecule sensor activity cysteine-type endopeptidase activator activity molecular condensate scaffold activity phosphatidylinositol phosphate binding
          Process(es)
          pattern recognition receptor signaling pathway acute inflammatory response negative regulation of acute inflammatory response positive regulation of cytokine production involved in immune response positive regulation of type 2 immune response inflammatory response osmosensory signaling pathway protein secretion detection of biotic stimulus response to virus negative regulation of interleukin-1 beta production positive regulation of interleukin-1 beta production positive regulation of interleukin-13 production positive regulation of interleukin-18 production positive regulation of interleukin-4 production positive regulation of interleukin-5 production interleukin-18-mediated signaling pathway NLRP3 inflammasome complex assembly response to ethanol positive regulation of T-helper 2 cell differentiation positive regulation of transcription from RNA polymerase II promoter regulation of inflammatory response negative regulation of inflammatory response positive regulation of inflammatory response defense response to Gram-positive bacterium protein homooligomerization protein maturation defense response to virus pyroptosis interleukin-1-mediated signaling pathway cellular response to lipopolysaccharide cellular response to peptidoglycan cellular response to virus positive regulation of NLRP3 inflammasome complex assembly negative regulation of NIK/NF-kappaB signaling positive regulation of NIK/NF-kappaB signaling positive regulation of T-helper 17 cell differentiation positive regulation of T-helper 2 cell cytokine production apoptotic process defense response signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0