Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FABP1 Antibodies

          Invitrogen

          Phospho-FABP1 (Ser11) Polyclonal Antibody

          View all (62) FABP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-FABP1 (Ser11) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Phospho-FABP1 (Ser11) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-FABP1 (Ser11) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-FABP1 (Ser11) Antibody (PA5-105330) in IHC (P)

          Immunohistochemistry analysis of paraffin-embedded Phospho-FABP1 (Ser11) in rat liver tissue sections. Antigen retrieval was performed using citrate buffer. Samples were blocked with blocking buffer (1.5 hr, 22°C), incubated with Phospho-FABP1 (Ser11) polyclonal antibody (Product # PA5-105330) using a dilution of 1:100 (1.5 hr, 22°C), followed by HRP conjugated goat anti-rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-FABP1 (Ser11) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-FABP1 (Ser11) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-FABP1 (Ser11) Polyclonal Antibody

          Product Details

          PA5-105330

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human FABP1(Accession P07148), corresponding to amino acid residues around phosphorylated Ser11.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816758

          Product Specific Information

          Antibody detects endogenous levels of FABP1 only when phosphorylated at Ser11.

          Target Information

          FABP1 encodes the fatty acid binding protein found in liver. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. FABP1 and FABP6 (the ileal fatty acid binding protein) are also able to bind bile acids. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 14 kDa selenium-binding protein; FABP1; fatty acid binding protein; fatty acid binding protein (FABP); fatty acid binding protein 1, liver; fatty acid-binding protein 1; L-FABP; liver fatty acid binding protein p14; liver-type fatty acid-binding protein; squalene- and sterol-carrier protein; sterol carrier protein; unnamed protein product; Z-protein

          View more View less

          Gene Aliases: FABPL; Fabplg; L-FABP; p14; SCP; SCP.

          View more View less

          UniProt ID: (Human) P07148, (Rat) P02692, (Mouse) P12710

          View more View less

          Entrez Gene ID: (Human) 2168, (Rat) 24360, (Mouse) 14080

          View more View less

          Function(s)
          chromatin binding lipid transporter activity long-chain fatty acid transporter activity fatty acid binding protein binding lipid binding cholesterol binding antioxidant activity sterol binding long-chain fatty acyl-CoA binding intermembrane lipid transfer activity molecular carrier activity phospholipid binding bile acid binding lysophospholipid transporter activity oleic acid binding heterocyclic compound binding binding
          Process(es)
          long-chain fatty acid metabolic process cell activation liver development reduction of food intake in response to dietary excess glucose metabolic process glycerol metabolic process cellular aldehyde metabolic process glycolytic process glutamine metabolic process lipid metabolic process fatty acid metabolic process fatty acid biosynthetic process fatty acid beta-oxidation unsaturated fatty acid biosynthetic process acylglycerol metabolic process triglyceride metabolic process phospholipid metabolic process glutathione metabolic process lipid transport membrane budding inflammatory response response to oxidative stress response to nutrient cholesterol metabolic process bile acid metabolic process cell proliferation adult feeding behavior response to xenobiotic stimulus response to wounding gene expression fatty acid transport long-chain fatty acid transport lipid catabolic process diterpenoid metabolic process arachidonic acid metabolic process fatty acid oxidation triglyceride biosynthetic process digestive system process positive regulation of fatty acid beta-oxidation response to food bile acid secretion cholesterol efflux response to vitamin B3 response to lipid chylomicron assembly very-low-density lipoprotein particle clearance multicellular organism growth hepatic stellate cell activation cannabinoid signaling pathway lipoprotein metabolic process cellular ketone metabolic process retinol metabolic process response to starvation cholesterol homeostasis xenobiotic transport lipoprotein transport long-chain fatty acid import monoacylglycerol metabolic process acylglycerol biosynthetic process digestive tract development smooth muscle tissue development regulation of catalytic activity intestinal absorption positive regulation of hydrolase activity lysophospholipid transport fatty acid primary amide metabolic process cellular response to hydrogen peroxide triglyceride homeostasis response to fatty acid response to cholesterol cellular response to hypoxia endocannabinoid signaling pathway energy homeostasis renal protein absorption connective tissue replacement cellular oxidant detoxification intermembrane lipid transfer lipid droplet formation intracellular signaling cassette response to paracetamol fatty acid derivative metabolic process glucose transmembrane transport response to tetrachloromethane cellular detoxification protein secretion macrophage differentiation response to peptide regulation of lipid metabolic process triglyceride catabolic process heme catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0