Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PKA alpha/beta/gamma Antibodies

          Invitrogen

          Phospho-PKA alpha/beta/gamma (Thr198) Polyclonal Antibody

          1 Reference
          View all (5) PKA alpha/beta/gamma antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-PKA alpha/beta/gamma (Thr198) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          Phospho-PKA alpha/beta/gamma (Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-PKA alpha/beta/gamma (Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PKA alpha/beta/gamma (Thr198) Antibody (PA5-105515) in IHC (P)

          Immunohistochemistry analysis of paraffin-embedded Phospho-PKA alpha/beta (Thr197) in human kidney tissue sections. Antigen retrieval was performed using citrate buffer. Samples were blocked with blocking buffer (1.5 hr, 22°C), incubated with Phospho-PKA alpha/beta (Thr197) polyclonal antibody (Product # PA5-105515) using a dilution of 1:100 (1.5 hr, 22°C), followed by HRP conjugated goat anti-rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PKA alpha/beta/gamma (Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-PKA alpha/beta/gamma (Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-PKA alpha/beta/gamma (Thr198) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-PKA alpha/beta/gamma (Thr198) Antibody in Western Blot (WB)
          Phospho-PKA alpha/beta/gamma (Thr198) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-105515

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human PRKACA(Accession P17612), corresponding to amino acid residues around phosphorylated Thr198.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816943

          Product Specific Information

          Antibody detects endogenous levels of PKA alpha/beta/gamma CAT only when phosphorylated at Thr198.

          Target Information

          Protein kinase A (PKA, cAMP-dependent protein kinase) is a key element of a ubiquitous signaling pathway important in the cell cycle, cellular communication, memory formation and behavior. PKA is composed of two catalytic (PKAc; Protein Kinase A catalytic subunit) and two regulatory subunits (PKAr). Upon binding cAMP, the complex dissociates to PKAr dimer and two activated PKAc ser/thr protein kinase catalytic monomers. The released PKAc can translocate into the nucleus and exert a regulatory role in the activation of multiple nuclear hormone receptors. However, PKAc-mediated activation of tonicity-dependent gene expression is cAMP independent. Humans express three types of PKAc subunit - PKAc alpha is present in most human tissues, PKAc beta and gamma are tissue-specific, the later is found in testes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C-alpha-s; alternatively spliced isoform; cAMP-dependent protein kinase C beta; cAMP-dependent protein kinase catalytic subunit C alpha; Cbeta2 PKA; Cs-PKA; PKA C-alpha; PKA C-beta; PKA C-gamma; protein kinase A; protein kinase A catalytic subunit; protein kinase A catalytic subunit beta; protein kinase, cAMP dependent, catalytic, beta; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, beta catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; protein kinase, cAMP-dependent, catalytic, beta; protein kinase, cAMP-dependent, catalytic, gamma; protein kinase, cAMP-dependent, gamma catalytic subunit; serine(threonine) protein kinase; sperm cAMP-dependent protein kinase catalytic subunit; unnamed protein product

          View more View less

          Gene Aliases: BDPLT19; CAFD1; CAFD2; CbPKA; Cs-PKA; KAPG; PKA C-beta; PKACA; PKACB; PKACg; PKCA1; PKCD; PPNAD4

          View more View less

          UniProt ID: (Human) P17612, (Human) P22694, (Human) P22612, (Mouse) P05132, (Mouse) P68181, (Rat) P68182

          View more View less

          Entrez Gene ID: (Human) 5566, (Human) 5567, (Human) 5568, (Mouse) 18747, (Rat) 25636, (Mouse) 18749, (Rat) 293508

          View more View less

          Function(s)
          magnesium ion binding protein kinase activity protein serine/threonine kinase activity cAMP-dependent protein kinase activity protein serine/threonine/tyrosine kinase activity protein binding ATP binding potassium channel inhibitor activity protein kinase binding protein domain specific binding manganese ion binding protein kinase A regulatory subunit binding channel activator activity protein serine kinase activity histone H1-4S35 kinase activity small GTPase binding ubiquitin protein ligase binding macromolecular complex binding cyclic nucleotide-dependent protein kinase activity
          Process(es)
          renal water homeostasis adenylate cyclase-activating G-protein coupled receptor signaling pathway spermatogenesis male gonad development positive regulation of insulin secretion high-density lipoprotein particle assembly vascular endothelial cell response to laminar fluid shear stress neural tube closure protein phosphorylation adenylate cyclase-modulating G-protein coupled receptor signaling pathway cellular response to nutrient levels TORC1 signaling negative regulation of smoothened signaling pathway negative regulation of meiotic cell cycle regulation of protein processing negative regulation of TORC1 signaling mesoderm formation chromatin remodeling mRNA processing adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway positive regulation of cell proliferation positive regulation of triglyceride catabolic process peptidyl-serine phosphorylation peptidyl-threonine phosphorylation cellular potassium ion homeostasis cellular response to heat insulin secretion involved in cellular response to glucose stimulus interleukin-2-mediated signaling pathway regulation of cellular respiration positive regulation of cholesterol biosynthetic process regulation of osteoblast differentiation positive regulation of gluconeogenesis negative regulation of glycolytic process protein autophosphorylation positive regulation of protein export from nucleus sperm capacitation spontaneous exocytosis of neurotransmitter positive regulation of phagocytosis modulation of synaptic transmission regulation of cell cycle regulation of synaptic transmission, glutamatergic regulation of proteasomal protein catabolic process cellular response to cold regulation of microtubule cytoskeleton organization cellular response to glucose stimulus cellular response to parathyroid hormone stimulus cellular response to glucagon stimulus postsynaptic modulation of chemical synaptic transmission negative regulation of protein localization to chromatin cAMP/PKA signal transduction positive regulation of protein localization to plasma membrane negative regulation of glycolytic process through fructose-6-phosphate protein localization to lipid particle regulation of bicellular tight junction assembly negative regulation of meiotic cell cycle process involved in oocyte maturation regulation of heart rate regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion regulation of macroautophagy cytokine-mediated signaling pathway negative regulation of interleukin-2 production mitochondrial protein catabolic process positive regulation of calcium-mediated signaling regulation of cardiac muscle contraction cellular response to epinephrine stimulus cell communication by electrical coupling involved in cardiac conduction regulation of cardiac conduction signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline