Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • LRP6 Antibodies

          Invitrogen

          Phospho-LRP6 (Thr1479) Polyclonal Antibody

          View all (29) LRP6 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-LRP6 (Thr1479) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-LRP6 (Thr1479) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-LRP6 (Thr1479) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-LRP6 (Thr1479) Antibody (PA5-106063) in WB

          Western blot analysis of Phospho-LRP6 (Thr1479) in MDA-MB-435 cells lysate (absence (-) or presence (+) of non-phospho and phospho peptide). Samples were incubated with Phospho-LRP6 (Thr1479) polyclonal antibody (Product # PA5-106063). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-LRP6 (Thr1479) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-LRP6 (Thr1479) Polyclonal Antibody

          Product Details

          PA5-106063

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human LRP6(Accession O75581), corresponding to amino acid residues around phosphorylated Thr1479.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2817462

          Product Specific Information

          Antibody detects endogenous levels of LRP6 only when phosphorylated at Thr1479.

          Target Information

          LRP6 is essential for the Wnt/beta catenin signaling pathway, probably by acting as a coreceptor together with Frizzled for Wnt. It is a specific, high-affinity receptor for DKK1 and DKK2, but not DKK3. The interaction with DKK1 blocks LRP6-mediated Wnt/beta catenin signaling via LRP6 removal via Kremen proteins-mediated endocytosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: crooked; FLJ90062; FLJ90421; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; low density lipoprotein-related protein 6; lpr6; LRP-6; transmembrane protein; unnamed protein product

          View more View less

          Gene Aliases: ADCAD2; C030016K15Rik; Cd; EVR8; Gw; OPTA4; ska26; ska; skax26; STHAG7

          View more View less

          UniProt ID: (Human) O75581, (Mouse) O88572

          View more View less

          Entrez Gene ID: (Human) 4040, (Mouse) 16974

          View more View less

          Function(s)
          low-density lipoprotein receptor activity receptor binding frizzled binding protein binding coreceptor activity Wnt-protein binding kinase inhibitor activity toxin transporter activity protein serine/threonine kinase inhibitor activity identical protein binding protein homodimerization activity Wnt-activated receptor activity apolipoprotein binding
          Process(es)
          regulation of transcription, DNA-templated positive regulation of cytosolic calcium ion concentration chemical synaptic transmission nervous system development tissue development neural crest formation neural crest cell differentiation Wnt signaling pathway vesicle-mediated transport midbrain development negative regulation of smooth muscle cell apoptotic process regulation of apoptotic process chordate embryonic development response to peptide hormone positive regulation of cell cycle positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter anatomical structure formation involved in morphogenesis transmembrane transport canonical Wnt signaling pathway cellular response to cholesterol dopaminergic neuron differentiation protein localization to plasma membrane cell-cell adhesion midbrain dopaminergic neuron differentiation gastrulation with mouth forming second somitogenesis neural tube closure heart looping positive regulation of mesenchymal cell proliferation pericardium morphogenesis axis elongation embryonic pattern specification dorsal/ventral axis specification anterior/posterior pattern specification positive regulation of neuron projection development cerebellum morphogenesis thalamus development cerebral cortex cell migration forebrain radial glial cell differentiation forebrain generation of neurons neural tube development formation of radial glial scaffolds cerebral cortex development embryonic limb morphogenesis forebrain development midbrain-hindbrain boundary development limb morphogenesis embryonic forelimb morphogenesis embryonic hindlimb morphogenesis external genitalia morphogenesis post-anal tail morphogenesis cell migration involved in gastrulation regulation of cell proliferation odontogenesis of dentin-containing tooth embryonic digit morphogenesis positive regulation of apoptotic process negative regulation of fat cell differentiation positive regulation of ossification positive regulation of bone resorption bone remodeling embryonic camera-type eye morphogenesis generation of neurons skeletal system morphogenesis negative regulation of epithelial cell proliferation response to folic acid palate development convergent extension retina morphogenesis in camera-type eye embryonic retina morphogenesis in camera-type eye regulation of cell development face morphogenesis branching involved in mammary gland duct morphogenesis trachea cartilage morphogenesis mammary placode formation mammary gland duct morphogenesis establishment of blood-brain barrier primitive streak formation receptor-mediated endocytosis of low-density lipoprotein particle involved in cholesterol transport axis elongation involved in somitogenesis Norrin signaling pathway establishment of blood-retinal barrier negative regulation of non-canonical Wnt signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0