Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PRK1/PRK2 Antibodies

          Invitrogen

          PRK1/PRK2 Polyclonal Antibody

          View all (3) PRK1/PRK2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PRK1/PRK2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          PRK1/PRK2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PRK1/PRK2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PRK1/PRK2 Antibody (PA5-106263) in WB

          Western blot analysis of PRK1/PRK2 in HeLa cell lysate (left lane: treated with blocking peptide). Samples were incubated with PRK1/PRK2 polyclonal antibody (Product # PA5-106263). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PRK1/PRK2 Antibody in Western Blot (WB)
          PRK1/PRK2 Antibody in Western Blot (WB)
          PRK1/PRK2 Polyclonal Antibody

          Product Details

          PA5-106263

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human PKN1(Accession Q16512), corresponding to amino acid residues L748-W798.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2817645

          Product Specific Information

          Antibody detects endogenous levels of total PRK1/2.

          Target Information

          Receptor-like kinase involved in the control of pollen germination and pollen tube polar growth.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cardiolipin-activated protein kinase Pak2; p140 kinase; PAK-1; PKN gamma; Protease-activated kinase 1; protease-activated kinase 2; Protein kinase C-like 1; Protein kinase C-like 2; Protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase PKN; Protein kinase PKN-alpha; Protein-kinase C-related kinase 1; Protein-kinase C-related kinase 2; serine-threonine kinase N; Serine-threonine protein kinase N; serine/threonine kinase 3; serine/threonine kinase 7; serine/threonine protein kinase N; Serine/threonine-protein kinase N1; Serine/threonine-protein kinase N2; unnamed protein product

          View more View less

          Gene Aliases: 6030436C20Rik; AI507382; DBK; F730027O18Rik; PAK-1; Pak-2; PAK1; PAK2; PKN; PKN-ALPHA; PKN1; PKN2; PRK1; PRK2; PRKCL1; PRKCL2; PRO2042; Stk3; STK7

          View more View less

          UniProt ID: (Human) Q16512, (Human) Q16513, (Rat) Q63433, (Mouse) P70268, (Mouse) Q8BWW9

          View more View less

          Entrez Gene ID: (Human) 5585, (Human) 5586, (Rat) 29355, (Mouse) 320795, (Mouse) 109333, (Rat) 207122

          View more View less

          Function(s)
          nucleotide binding chromatin binding transcription coactivator activity protein kinase activity protein serine/threonine kinase activity protein kinase C activity protein kinase C binding protein binding ATP binding kinase activity transferase activity small GTPase binding histone kinase activity (H3-T11 specific) histone binding histone deacetylase binding androgen receptor binding protein serine kinase activity RNA binding protein tyrosine kinase activity cadherin binding RNA polymerase binding GTP-Rho binding ligand-dependent nuclear receptor transcription coactivator activity Rac GTPase binding transferase activity, transferring phosphorus-containing groups poly(A) RNA binding non-receptor serine/threonine protein kinase protein modifying enzyme
          Process(es)
          B cell homeostasis B cell apoptotic process regulation of germinal center formation regulation of immunoglobulin production renal system process chromatin organization chromatin remodeling regulation of transcription from RNA polymerase II promoter protein phosphorylation hyperosmotic response signal transduction epithelial cell migration negative regulation of B cell proliferation intracellular signal transduction post-translational protein modification positive regulation of transcription, DNA-templated spleen development regulation of androgen receptor signaling pathway regulation of cell motility apoptotic process cell adhesion cell projection organization positive regulation of cytokinesis apical junction assembly positive regulation of viral genome replication positive regulation of mitotic cell cycle cell division vascular endothelial cell response to laminar fluid shear stress negative regulation of protein phosphorylation transcription, DNA-templated negative regulation of protein kinase activity histone H3-T11 phosphorylation regulation of transcription, DNA-templated phosphorylation chromatin modification cell cycle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-hn5gs:80/100.66.79.229:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline