Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TRAP220 Antibodies

          Invitrogen

          TRAP220 Polyclonal Antibody

          View all (24) TRAP220 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TRAP220 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          TRAP220 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TRAP220 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRAP220 Antibody (PA5-106503) in WB

          Western blot analysis of TRAP220 in rat muscle lysate. Samples were incubated with TRAP220 polyclonal antibody (Product # PA5-106503). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRAP220 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TRAP220 Polyclonal Antibody

          Product Details

          PA5-106503

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human MED1(Accession Q15648), corresponding to amino acid residues Y1431-S1481.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2854172

          Product Specific Information

          Antibody detects endogenous levels of total TRAP220/MED1.

          Target Information

          Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: activator-recruited cofactor 205 kDa component; ARC205; nuclear receptor interacting protein; p53 regulatory protein RB18A; PBP165; peroxisome proliferator-activated receptor-binding protein; Ppar binding protein; PPAR-binding protein; PPARG binding protein; PPRB; thyroid hormone receptor-associated protein complex 220 kDa component; thyroid hormone receptor-associated protein complex component TRAP220; thyroid receptor interacting protein 2; thyroid receptor-interacting protein 2; TR-interacting protein 2; TRIP-2; unnamed protein product; vitamin D receptor-interacting protein 230 kD; vitamin D receptor-interacting protein complex component DRIP205

          View more View less

          Gene Aliases: CRSP1; CRSP200; CRSP210; DRIP205; DRIP230; l11Jus15; PBP; PPARBP; PPARGBP; RB18A; RGD1559552; TRAP220; TRIP-2; TRIP2

          View more View less

          UniProt ID: (Human) Q15648, (Mouse) Q925J9

          View more View less

          Entrez Gene ID: (Human) 5469, (Rat) 497991, (Mouse) 19014

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding transcription coactivator binding DNA binding chromatin binding transcription cofactor activity transcription coactivator activity transcription corepressor activity protein binding ligand-dependent nuclear receptor binding estrogen receptor binding chromatin DNA binding histone acetyltransferase binding vitamin D receptor binding retinoic acid receptor binding peroxisome proliferator activated receptor binding macromolecular complex binding thyroid hormone receptor binding LBD domain binding RNA polymerase II sequence-specific DNA binding transcription factor binding ubiquitin protein ligase activity general transcription initiation factor binding DNA-binding transcription factor binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter cell morphogenesis angiogenesis in utero embryonic development liver development embryonic placenta development lens development in camera-type eye thyroid hormone mediated signaling pathway ventricular trabecula myocardium morphogenesis retinal pigment epithelium development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter thyroid hormone generation protein import into nucleus androgen biosynthetic process brain development heart development lactation positive regulation of cell proliferation organ morphogenesis positive regulation of gene expression negative regulation of keratinocyte proliferation protein ubiquitination keratinocyte differentiation monocyte differentiation intracellular steroid hormone receptor signaling pathway organ regeneration positive regulation of intracellular estrogen receptor signaling pathway mammary gland epithelial cell proliferation embryonic heart tube development embryonic hindlimb morphogenesis embryonic hemopoiesis peroxisome proliferator activated receptor signaling pathway cellular response to hepatocyte growth factor stimulus megakaryocyte development mRNA transcription from RNA polymerase II promoter camera-type eye development negative regulation of apoptotic process fat cell differentiation positive regulation of keratinocyte differentiation positive regulation of erythrocyte differentiation negative regulation of neuron differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter erythrocyte development enucleate erythrocyte development positive regulation of transcription initiation from RNA polymerase II promoter positive regulation of interferon-gamma-mediated signaling pathway mammary gland branching involved in thelarche mammary gland branching involved in pregnancy epithelial cell proliferation involved in mammary gland duct elongation positive regulation of G0 to G1 transition regulation of vitamin D receptor signaling pathway cellular response to epidermal growth factor stimulus cellular response to steroid hormone stimulus cellular response to thyroid hormone stimulus positive regulation of hepatocyte proliferation regulation of RNA biosynthetic process positive regulation of transcription elongation from RNA polymerase II promoter RNA polymerase II transcriptional preinitiation complex assembly transcription from RNA polymerase II promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0