Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • hnRNP U Antibodies

          Invitrogen

          HNRNPU Polyclonal Antibody

          View all (18) hnRNP U antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HNRNPU Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          HNRNPU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HNRNPU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HNRNPU Antibody (PA5-109827) in ICC/IF

          Immunofluorescence analysis of HNRNPU in C6 cells. Samples were incubated with HNRNPU Polyclonal antibody (Product # PA5-109827) using a dilution of 1:100. Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HNRNPU Antibody in Immunocytochemistry (ICC/IF)
          HNRNPU Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPU Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPU Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HNRNPU Antibody in Western Blot (WB)
          HNRNPU Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          HNRNPU Polyclonal Antibody

          Product Details

          PA5-109827

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -

          Immunoprecipitation (IP)

          0.5 µg-4 µg antibody for 200 µg-400 µg extracts of
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 608-825 of human hnRNP U (NP_1140322)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.60 mg/mL

          Amount

          160 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2855238

          Product Specific Information

          Immunogen sequence: PKDEDYKQRT QKKAEVEGKD LPEHAVLKMK GNFTLPEVAE CFDEITYVEL QKEEAQKLLE QYKEESKKAL PPEKKQNTGS KKSNKNKSGK NQFNRGGGHR GRGGFNMRGG NFRGGAPGNR GGYNRRGNMP QRGGGGGGSG GIGYPYPRAP VFPGRGSYSN RGNYNRGGMP NRGNYNQNFR GRGNNRGYKN QSQGYNQWQQ GQFWGQKPWS QHYHQGYY

          Target Information

          The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: contains signal peptide domain; FLJ30202; FLJ37978; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogenous nuclear ribonucleoprotein U; HNRNPU antisense RNA 1; NCRNA00201; nuclear matrix protein sp120; nuclear p120 ribonucleoprotein; p120 nuclear protein; SAR-DNA binding protein of the nuclear scaffold; scaffold attachment factor A; scaffold-attachment factor A; system N1 Na+ and H+-coupled glutamine transporter; transporter protein; transporter protein; system N1 Na+ and H+-coupled glutamine transporter; U211; unnamed protein product

          View more View less

          Gene Aliases: DEE54; EIEE54; GRIP120; hnRNP U; HNRNPU-AS1; HNRPU; pp120; SAF-A; SAFA; SN1; SP120; U21.1

          View more View less

          UniProt ID: (Human) Q00839, (Rat) Q6IMY8, (Mouse) Q8VEK3

          View more View less

          Entrez Gene ID: (Human) 3192, (Rat) 117280, (Mouse) 51810

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core binding TFIIH-class transcription factor binding DNA binding chromatin binding double-stranded DNA binding single-stranded DNA binding transcription corepressor activity RNA binding double-stranded RNA binding single-stranded RNA binding mRNA 3'-UTR binding actin binding protein binding ATP binding poly(A) binding snRNA binding poly(C) RNA binding chromatin DNA binding poly(G) binding piRNA binding pre-mRNA binding identical protein binding ribonucleoprotein complex binding sequence-specific DNA binding macromolecular complex binding telomerase RNA binding RNA polymerase II C-terminal domain binding lncRNA binding sequence-specific double-stranded DNA binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter alternative mRNA splicing, via spliceosome regulation of alternative mRNA splicing, via spliceosome regulation of mitotic cell cycle dosage compensation by inactivation of X chromosome regulation of gene expression positive regulation of gene expression mRNA metabolic process erythrocyte differentiation chromatin silencing by small RNA negative regulation of telomere maintenance via telomerase circadian regulation of gene expression negative regulation of kinase activity negative regulation of transcription elongation from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter mRNA stabilization maintenance of protein location in nucleus cardiac muscle cell development random inactivation of X chromosome CRD-mediated mRNA stabilization cellular response to glucocorticoid stimulus cellular response to dexamethasone stimulus positive regulation of brown fat cell differentiation dendritic transport of messenger ribonucleoprotein complex negative regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay regulation of mitotic spindle assembly regulation of chromatin organization positive regulation of attachment of mitotic spindle microtubules to kinetochore protein localization to spindle microtubule RNA localization to chromatin cellular response to leukemia inhibitory factor adaptive thermogenesis positive regulation of DNA topoisomerase (ATP-hydrolyzing) activity positive regulation of stem cell proliferation negative regulation of stem cell differentiation positive regulation of cytoplasmic translation mRNA splicing, via spliceosome osteoblast differentiation chromatin organization RNA processing
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0