Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Midkine Antibodies

          Invitrogen

          MDK Polyclonal Antibody

          View all (25) Midkine antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MDK Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          MDK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MDK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MDK Antibody (PA5-109951) in ICC/IF

          Immunofluorescence analysis of MDK in NIH/3T3 cells. Samples were incubated with MDK Polyclonal antibody (Product # PA5-109951) using a dilution of 1:20 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MDK Antibody in Immunocytochemistry (ICC/IF)
          MDK Antibody in Immunocytochemistry (ICC/IF)
          MDK Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MDK Polyclonal Antibody

          Product Details

          PA5-109951

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein (or fragment)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2855362

          Product Specific Information

          Immunogen sequence: KKKDKVKKGG PGSECAEWAW GPCTPSSKDC GVGFREGTCG AQTQRIRCRV PCNWKKEFGA DCKYKFENWG ACDGGTGTKV RQGTLKKARY NAQCQETIRV TKPCTPKTKA KAKAKKGKGK D

          Target Information

          Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Amphiregulin-associated protein; ARAP; FLJ27379; Midgestation and kidney protein; Midkine; MK; neurite growth-promoting factor 2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; Retinoic acid-induced differentiation factor; retinoic acid-responsive protein

          View more View less

          Gene Aliases: ARAP; MDK; Mek; MK; MK1; NEGF2

          View more View less

          UniProt ID: (Human) P21741, (Mouse) P12025

          View more View less

          Entrez Gene ID: (Human) 4192, (Mouse) 17242

          View more View less

          Function(s)
          protein binding growth factor activity heparin binding chondroitin sulfate binding heparan sulfate binding growth factor
          Process(es)
          behavioral fear response leukocyte chemotaxis involved in inflammatory response T cell activation involved in immune response positive regulation of leukocyte chemotaxis cytoskeleton organization negative regulation of cell adhesion signal transduction nervous system development short-term memory response to xenobiotic stimulus response to wounding response to hormone negative regulation of cardiac muscle cell apoptotic process positive regulation of epithelial to mesenchymal transition positive regulation of macrophage chemotaxis positive regulation of keratinocyte proliferation positive regulation of neuron projection development response to auditory stimulus cell migration dentate gyrus development cerebellar granular layer development hippocampus development cerebral cortex development cell differentiation negative regulation of ossification adrenal gland development positive regulation of cell migration defecation regulation of chondrocyte differentiation positive regulation of interleukin-12 production intracellular signal transduction tissue regeneration negative regulation of neuron apoptotic process estrous cycle positive regulation of T cell differentiation negative regulation of regulatory T cell differentiation positive regulation of cell adhesion positive regulation of transcription, DNA-templated regulation of bone remodeling oogenesis positive regulation of oligodendrocyte differentiation positive regulation of inflammatory response regulation of behavior response to glucocorticoid positive regulation of cell division positive regulation of cartilage development positive regulation of smooth muscle cell chemotaxis positive regulation of neutrophil chemotaxis negative regulation of canonical Wnt signaling pathway negative regulation of inflammatory response to wounding positive regulation of inflammatory response to wounding glial cell projection elongation positive regulation of substrate adhesion-dependent cell spreading positive regulation of leukocyte cell-cell adhesion negative regulation of epithelial cell apoptotic process positive regulation of leukocyte adhesion to vascular endothelial cell positive regulation of blood vessel branching positive regulation of vascular endothelial cell proliferation positive regulation of artery morphogenesis positive regulation of neural precursor cell proliferation positive regulation of hepatocyte proliferation positive regulation of neutrophil extravasation positive regulation of neuron migration multicellular organism development negative regulation of neuron death
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-dd86746ff-lmgjf:80/100.66.77.199:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline