Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LGVSFLVLPK YERIMQKYNL LPEKSMYDLV HGSSLWMLCT DVALEFPRPT LPNVVYVGGI LTKPASPLPE DLQRWVNGAN EHGFVLVSFG AGVKYLSEDI ANKLAGA
Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 96%.
UGT8 (UDP-galactose-ceramide galactosyltransferase, 2-hydroxyacylsphingosine 1-beta-galactosyltransferase) is a 541 amino acid, single pass membrane protein of the UDP-glycosyltransferase family. UGT8 is believed to be primarily involved with the metabolism of sphingolipids and galactosylceramide biosynthesis. UGT8 is one of six genes whose elevated expression has been correlated with a significantly increased the risk of lung metastases in breast cancer patients. As such, UGT8 may be a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. UGT8 is ubiquitously expressed with highest levels found in central and peripheral nervous systems and is up-regulated in breast cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ceramide UDP-galactosyltransferase; cerebroside synthase; UDP-galactose-ceramide galactosyltransferase; unnamed protein product; uridine diphosphate glycosyltransferase 8
Gene Aliases: CGT; UGT4
UniProt ID: (Human) Q16880
Entrez Gene ID: (Human) 7368
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support