Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • AMPK alpha-1,2 Antibodies

          Invitrogen

          Phospho-AMPK alpha-1,2 (Thr172) Polyclonal Antibody

          1 Published Figure
          2 References
          View all (20) AMPK alpha-1,2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-AMPK alpha-1,2 (Thr172) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (13)
          • Published Figures (1)
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-AMPK alpha-1,2 (Thr172) Antibody (PA5-114550) in ICC/IF

          Immunocytochemistry analysis of Phospho-AMPK alpha-1,2 (Thr172) in C2C12 cells. Samples were treated with PFA, permeabilized in 0.1% saponin prior to blocking in 10% serum (45 min at 37°C), and incubated with polyclonal antibody (Product # PA5-114550) at a dilution of 1:400 (1 hr at 37°C). Secondary staining was applied with Alexa Fluor 594 conjugated goat anti-rabbit IgG using a dilution of 1:600. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Western Blot (WB)
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Western Blot (WB)
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Western Blot (WB)
          Phospho-AMPK alpha-1,2 (Thr172) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-AMPK alpha-1,2 (Thr172) Polyclonal Antibody

          Product Details

          PA5-114550

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human PRKAA1(Accession Q13131), corresponding to amino acid residues around phosphorylated Thr172.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2899186

          Product Specific Information

          Antibody detects endogenous levels of AMPK alpha 1 when phosphorylated at Thr183, or AMPK alpha 2 when phosphorylated at Thr172.

          Target Information

          The protein encoded by this gene is a catalytic subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. Studies of the mouse counterpart suggest that this catalytic subunit may control whole-body insulin sensitivity and is necessary for maintaining myocardial energy homeostasis during ischemia.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 5'-AMP-activated protein kinase, catalytic alpha-1 chain; 5'-AMP-activated protein kinase, catalytic alpha-2 chain; AAPK2; ACACA kinase; acetyl-CoA carboxylase kinase; AMP -activate kinase alpha 1 subunit; AMP-activated protein kinase; AMP-activated protein kinase, alpha 1 catalytic subunit; AMP-activated protein kinase, catalytic, alpha-1; AMPK alpha 2; AMPK alpha-1 chain; AMPK alpha-2 chain; AMPK alpha-2 variant 2; AMPK alpha-2 variant 3; AMPK subunit alpha-1; AMPK subunit alpha-2; AMPK, alpha, 1; AMPK-alpha-1 chain; AMPK-alpha-2 chain; HMGCR kinase; hydroxymethylglutaryl-CoA reductase kinase; pAMPK; protein kinase; protein kinase, AMP-activated, alpha 1 catalytic subunit; protein kinase, AMP-activated, alpha 2 catalytic subunit; tau-protein kinase PRKAA1; unnamed protein product

          View more View less

          Gene Aliases: 2310008I11Rik; A830082D05; AMPK; AMPK alpha 1; AMPK2; AMPKa1; AMPKa2; AMPKalpha1; AMPKalpha2; C130083N04Rik; PRKAA

          View more View less

          UniProt ID: (Human) Q13131, (Human) P54646, (Rat) P54645, (Mouse) Q5EG47, (Rat) Q09137, (Mouse) Q8BRK8

          View more View less

          Entrez Gene ID: (Human) 5562, (Human) 5563, (Rat) 65248, (Mouse) 105787, (Rat) 78975, (Mouse) 108079

          View more View less

          Function(s)
          chromatin binding protein kinase activity protein serine/threonine kinase activity AMP-activated protein kinase activity cAMP-dependent protein kinase activity protein binding ATP binding macromolecular complex binding [hydroxymethylglutaryl-CoA reductase (NADPH)] kinase activity tau protein binding tau-protein kinase activity protein serine kinase activity histone H2BS36 kinase activity protein serine/threonine/tyrosine kinase activity eukaryotic elongation factor-2 kinase activator activity kinase binding protein binding, bridging
          Process(es)
          chromatin remodeling protein phosphorylation lipid biosynthetic process regulation of gene expression positive regulation of autophagy negative regulation of gene expression response to activity response to muscle activity regulation of macroautophagy peptidyl-serine phosphorylation regulation of lipid metabolic process response to caffeine cellular response to nutrient levels negative regulation of TOR signaling adiponectin-activated signaling pathway cellular response to oxidative stress cellular response to glucose starvation glucose homeostasis regulation of circadian rhythm negative regulation of apoptotic process positive regulation of glycolytic process fatty acid homeostasis regulation of stress granule assembly regulation of microtubule cytoskeleton organization cellular response to calcium ion cellular response to glucose stimulus cellular response to prostaglandin E stimulus cellular response to xenobiotic stimulus energy homeostasis positive regulation of cellular protein localization negative regulation of hepatocyte apoptotic process negative regulation of TORC1 signaling lipid droplet disassembly protein localization to lipid particle cytoplasmic translation phosphatidylethanolamine biosynthetic process phosphatidylcholine biosynthetic process cellular response to starvation positive regulation of catabolic process cellular response to amino acid starvation TORC1 signaling negative regulation of translational initiation positive regulation of translational initiation protein localization to lysosome protein K6-linked ubiquitination positive regulation of release of cytochrome c from mitochondria hepatocyte apoptotic process positive regulation of TORC1 signaling signal transduction positive regulation of macroautophagy positive regulation of T cell mediated immune response to tumor cell glucose metabolic process positive regulation of cell proliferation response to xenobiotic stimulus response to UV cold acclimation response to gamma radiation positive regulation of gene expression negative regulation of translation fatty acid oxidation regulation of peptidyl-serine phosphorylation cellular response to stress response to hydrogen peroxide regulation of vascular permeability response to estrogen positive regulation of gluconeogenesis positive regulation of transcription, DNA-templated positive regulation of fatty acid oxidation positive regulation of glucose import negative regulation of insulin receptor signaling pathway positive regulation of skeletal muscle tissue development positive regulation of T cell activation negative regulation of lipid catabolic process regulation of vesicle-mediated transport motor behavior CAMKK-AMPK signaling cascade neuron cellular homeostasis cellular response to hydrogen peroxide cellular response to ethanol cellular response to hypoxia protein localization to membrane protein localization to plasma membrane negative regulation of ferroptosis positive regulation of transcription from mitochondrial promoter positive regulation of establishment of protein localization to mitochondrion positive regulation of adipose tissue development autophagosome assembly negative regulation of T cell mediated immune response to tumor cell autophagy lysosome organization SCF-dependent proteasomal ubiquitin-dependent protein catabolic process protein destabilization proteasome-mediated ubiquitin-dependent protein catabolic process protein stabilization negative regulation of T cell activation protein-containing complex assembly negative regulation of protein localization to nucleus response to hypoxia positive regulation of cholesterol biosynthetic process negative regulation of glucosylceramide biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline