Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RSK Pan Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for is temporarily unavailable. Please contact Technical Support for suitable replacement options.
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          The product (cat # {{productObj.productId}}) that you're looking for will be discontinued once stock is depleted. Please check out our other {{productObj.targetName}} {{productObj.pendingDiscontinuationTargetLabel}} Please check out our other {{productObj.pendingDiscontinuationTargetLabel}} or contact Technical Support with any questions.

          Invitrogen

          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Polyclonal Antibody

          View all (2) RSK Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (24)
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody (PA5-114637) in IHC (P)

          Immunohistochemistry analysis of Phospho-RSK1/RSK2/RSK3/RSK4 (Ser221, Ser227, Ser218, Ser232) in human ovarian cancer and adjacent normal tissues. Samples were treated with formaldehyde and treated with citrate buffer for antigen retrieval, blocked, and incubated (1.5 hours at 22°C) with polyclonal antibody (Product # PA5-114637) at a dilution of 1:100. Secondary staining was applied with HRP conjugated ant... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-RSK Pan (Ser221, Ser227, Ser218, Ser232) Polyclonal Antibody

          Product Details

          PA5-114637

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human RSK1/2/3/4 around the phosphorylation site of Ser221/227/s218/232.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2899273

          Product Specific Information

          Antibody detects endogenous levels of RSK1/2/3/4 only when phosphorylated at Ser221/227/s218/232.

          Target Information

          The p90 ribosomal S6 kinases (RSKs) comprise a family of serine/threonine kinases that lie at the terminus of the ERK pathway. In humans, the RSK family consists of four isoforms (RSK1 to-4). RSK family members are unusual among serine/threonine kinases in that they contain two distinct kinase domains, both of which are catalytically functional. Theses kinase domains are activated in a sequential manner by a series of phosphorylations. RSK regulates gene expression via association and phosphorylation of transcriptional regulators including c-Fos, estrogen receptor, NFkappaB/IkappaB alpha, cAMP-response element-binding protein (CREB). ERK activates the C-terminal kinase of RSK, leading to activation of the N-terminal kinase. Members of the RSK family are present in the cytoplasm as well as the nucleus. Addition of growth factor to the cells results in the activation of both cytosolic and nuclear RSK and the translocation of the cytosolic RSK into the nucleus upon activation. The activation and nuclear translocation of RSK result in phosphorylation and activation of transcription factors.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 90 kDa ribosomal protein S6 kinase 1; 90 kDa ribosomal protein S6 kinase 2; 90 kDa ribosomal protein S6 kinase 3; 90 kDa ribosomal protein S6 kinase 4; 90 kDa ribosomal protein S6 kinase 6; coiled-coil domain containing 88B; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); epididymis secretory sperm binding protein; insulin-stimulated protein kinase 1; MAP kinase-activated protein kinase 1a; MAP kinase-activated protein kinase 1b; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1a; MAPK-activated protein kinase 1b; MAPK-activated protein kinase 1c; MAPKAP kinase 1a; MAPKAP kinase 1b; MAPKAP kinase 1c; MAPKAPK-1a; MAPKAPK-1b; MAPKAPK-1c; MAPKAPK1A; MAPKAPK1D; mitogen- and stress-activated protein kinase 2; mitogen- and stress-activated protein kinase-2; mitogen-activated protein kinase-activated protein kinase 1C; nuclear mitogen- and stress-activated protein kinase 2; p90 rsk; p90-RSK 1; p90-RSK 2; p90-RSK 3; p90-RSK 6; p90rsk; p90RSK1; p90RSK2; p90S6K; pp90RSK1; pp90RSK3; protein-tyrosine kinase Mpk-9; ribosomal protein kinase B; ribosomal protein S6 kinase 2; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase polypeptide 2; ribosomal protein S6 kinase polypeptide 3; ribosomal protein S6 kinase polypeptide 6; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kD, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 3; ribosomal protein S6 kinase, 90kDa, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 6; ribosomal protein S6 kinase, polypeptide 2; ribosomal protein S6 kinase, polypeptide 4; ribosomal protein S6 kinase, polypeptide A1; ribosomal S6 kinase 1; ribosomal S6 kinase 2; ribosomal S6 kinase 3; ribosomal S6 kinase 4; RLSK; RP11-393H10.3; RPS6KA3; RPS6KA6; RSK-1; RSK-2; RSK-3; RSK-like protein kinase; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha 2; S6K-alpha 6; S6K-alpha-1; S6K-alpha-3; S6K-alpha-4; serine/threonine protein kinase RSK2; ribosomal protein S6 alpha 3; unnamed protein product

          View more View less

          Gene Aliases: 1110069D02Rik; 2610524K04Rik; 90kDa; Ccdc88b; CLS; D17Wsu134e; HU-1; HU-2; HU-3; ISPK-1; Mapkapk-1a; MAPKAPK-1b; MAPKAPK1; MAPKAPK1A; MAPKAPK1B; MAPKAPK1C; mMSK2; MPK-9; MRX19; MSK2; p90-Rsk1; p90-RSK2; p90-RSK3; p90Rsk; p90Rsk1; p90RSK2; p90RSK3; p90RSK6; p90S6K; pp90rsk; pp90RSK2; pp90RSK3; PP90RSK4; RGD1560817; RGD1563860; Rps6ka-rs1; RSK; Rsk-1; RSK-4; RSK-B; RSK1; RSK2; RSK3; RSK4; S6K-alpha; S6K-alpha 1; S6K-alpha-1; S6K-alpha-4; S6K-alpha-6; S6K-alpha2; S6K-alpha3; XLID19

          View more View less

          UniProt ID: (Human) Q15418, (Human) Q15349, (Human) P51812, (Human) O75676, (Human) Q9UK32, (Mouse) P18653, (Rat) Q63531, (Mouse) Q9WUT3, (Mouse) P18654, (Mouse) Q9Z2B9, (Mouse) Q7TPS0

          View more View less

          Entrez Gene ID: (Human) 6195, (Human) 6196, (Human) 6197, (Human) 8986, (Human) 27330, (Mouse) 20111, (Rat) 81771, (Rat) 117269, (Mouse) 20112, (Rat) 501560, (Mouse) 110651, (Rat) 361715, (Mouse) 56613, (Rat) 317203, (Mouse) 67071

          View more View less

          Function(s)
          magnesium ion binding protein kinase activity protein serine/threonine kinase activity ribosomal protein S6 kinase activity protein serine/threonine/tyrosine kinase activity protein binding ATP binding PDZ domain binding mitogen-activated protein kinase binding protein serine kinase activity protein kinase binding histone kinase activity (H3-S10 specific) histone kinase activity (H3-S28 specific) kinase activity DNA-binding transcription factor binding nucleotide binding
          Process(es)
          DNA damage response, signal transduction by p53 class mediator intracellular signal transduction TORC1 signaling negative regulation of embryonic development negative regulation of ERK1 and ERK2 cascade negative regulation of mesoderm development toll-like receptor signaling pathway biological_process response to lipopolysaccharide negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter protein phosphorylation negative regulation of TOR signaling positive regulation of transcription, DNA-templated hepatocyte proliferation positive regulation of hepatic stellate cell activation negative regulation of cytokine production chromatin remodeling regulation of transcription, DNA-templated post-translational protein modification interleukin-1-mediated signaling pathway oocyte maturation brain renin-angiotensin system heart development negative regulation of cell proliferation positive regulation of gene expression cardiac muscle cell apoptotic process peptidyl-serine phosphorylation positive regulation of apoptotic process negative regulation of cell cycle negative regulation of meiotic nuclear division heart contraction regulation of protein processing cellular response to carbohydrate stimulus skeletal system development signal transduction chemical synaptic transmission central nervous system development positive regulation of cell growth regulation of translation in response to stress positive regulation of cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0