Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Dicer Antibodies

          Invitrogen

          Dicer Polyclonal Antibody

          1 Published Figure
          1 Reference
          View all (44) Dicer antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Dicer Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (16)
          • Published Figures (1)
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Dicer Antibody (PA5-115124) in IHC (P)

          Immunohistochemistry analysis of Dicer in human normal tissues adjacent to mammary cancer. Samples were treated with formaldehyde and treated with citrate buffer for antigen retrieval, blocked, and incubated (1.5 hours at 22°C) with polyclonal antibody (Product # PA5-115124) at a dilution of 1:100. Secondary staining was applied with HRP conjugated anti-Rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Dicer Antibody in Western Blot (WB)
          Dicer Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Dicer Polyclonal Antibody

          Product Details

          PA5-115124

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human DICER1(Accession Q9UPY3), corresponding to amino acid residues K1516-L1566.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2899760

          Product Specific Information

          Antibody detects endogenous levels of total Dicer.

          Target Information

          Dicer is a double-stranded RNA (dsRNA) endoribonuclease playing a central role in short dsRNA-mediated post-transcriptional gene silencing. It cleaves naturally occurring long dsRNAs and short hairpin pre-microRNAs (miRNA) into fragments of twenty-one to twenty-three nucleotides with 3' overhang of two nucleotides, producing respectively short interfering RNAs (siRNA) and mature microRNAs. SiRNAs and miRNAs serve as guide to direct the RNA-induced silencing complex (RISC) to complementary RNAs to degrade them or prevent their translation. Gene silencing mediated by siRNAs, also called RNA interference, controls the elimination of transcripts from mobile and repetitive DNA elements of the genome but also the degradation of exogenous RNA of viral origin for instance. The miRNA pathway on the other side is a means to specifically regulate the expression of target genes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: antiviral Dicer; antiviral isoform of Dicer; DCR; DCR-1; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease type III; Dicer1, Dcr-1 homolog; double-strand-specific ribonuclease mDCR-1; helicase MOI; helicase with RNAse motif; helicase-moi; mDCR-1; unnamed protein product; variant of Dicer1

          View more View less

          Gene Aliases: 1110006F08Rik; aviD; D12Ertd7e; DCR1; Dicer; Dicer1e; GLOW; HERNA; K12H4.8-LIKE; mKIAA0928; MNG1; RMSE2

          View more View less

          UniProt ID: (Human) Q9UPY3

          View more View less

          Entrez Gene ID: (Human) 23405, (Rat) 299284, (Mouse) 192119

          View more View less

          Function(s)
          nucleic acid binding RNA binding double-stranded RNA binding endoribonuclease activity ribonuclease III activity deoxyribonuclease I activity protein binding ATP binding endoribonuclease activity, producing 5'-phosphomonoesters protein domain specific binding siRNA binding regulatory RNA binding pre-miRNA binding DNA binding hydrolase activity miRNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter meiotic spindle organization angiogenesis trophectodermal cell proliferation hair follicle development apoptotic DNA fragmentation RNA processing spermatogonial cell division cell proliferation post-embryonic development zygote asymmetric cell division regulation of gene expression positive regulation of endothelial cell migration negative regulation of Schwann cell proliferation positive regulation of gene expression negative regulation of gene expression regulation of muscle cell apoptotic process negative regulation of tumor necrosis factor-mediated signaling pathway positive regulation of Schwann cell differentiation myoblast differentiation involved in skeletal muscle regeneration posttranscriptional gene silencing stem cell population maintenance spinal cord motor neuron differentiation nerve development olfactory bulb interneuron differentiation cerebral cortex development lung development embryonic limb morphogenesis production of siRNA involved in RNA interference pre-miRNA processing hair follicle morphogenesis pericentric heterochromatin assembly regulation of myelination positive regulation of myelination peripheral nervous system myelin formation negative regulation of tumor necrosis factor production positive regulation of collagen biosynthetic process embryonic hindlimb morphogenesis posttranscriptional gene silencing by RNA production of miRNAs involved in gene silencing by miRNA multicellular organism growth regulation of odontogenesis of dentin-containing tooth regulation of viral genome replication regulation of regulatory T cell differentiation regulation of cell differentiation regulation of neuron differentiation positive regulation of transcription from RNA polymerase II promoter mRNA stabilization spleen development digestive tract development reproductive structure development regulation of oligodendrocyte differentiation epidermis morphogenesis branching morphogenesis of an epithelial tube neuron projection morphogenesis regulation of inflammatory response regulation of neurogenesis cartilage development spindle assembly regulation of multicellular organismal process regulation of RNA metabolic process defense response to virus regulation of cell cycle cardiac muscle cell development inner ear receptor cell development negative regulation of glial cell proliferation intestinal epithelial cell development cardiac neural crest cell development involved in outflow tract morphogenesis ganglion development regulation of enamel mineralization cellular response to hydrogen peroxide small RNA loading onto RISC cellular response to glucose stimulus hair follicle cell proliferation cellular response to tumor necrosis factor cellular response to hypoxia positive regulation of establishment of endothelial barrier positive regulation of hepatic stellate cell proliferation positive regulation of endothelial cell-matrix adhesion via fibronectin positive regulation of vascular endothelial cell proliferation regulation of miRNA metabolic process positive regulation of miRNA metabolic process regulation of stem cell differentiation rRNA processing regulation of Notch signaling pathway global gene silencing by mRNA cleavage miRNA metabolic process tRNA catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-q2vkv:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline