Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Lass2 Antibodies

          Invitrogen

          Lass2 Polyclonal Antibody

          View all (18) Lass2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Lass2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (32)
          Lass2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Lass2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 32

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Lass2 Antibody (PA5-115496) in ICC/IF

          Immunocytochemistry analysis of Lass2 in HepG2 cells. Samples were treated with PFA, permeabilized in 0.1% Triton X-100, blocked in 10% serum (45 min at 25°C), and incubated with polyclonal antibody (Product # PA5-115496). Secondary staining was applied with mouse anti-beta tubulin (1 hr at 37°, AlexaFluor 594 conjugated goat anti-rabbit IgG (Red), AlexaFluor 488 conjugated goat anti-mouse IgG (Green) and DAPI. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Lass2 Antibody in Immunocytochemistry (ICC/IF)
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lass2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Lass2 Polyclonal Antibody

          Product Details

          PA5-115496

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CERS2(Accession Q96G23), corresponding to amino acid residues T346-D380.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2900132

          Product Specific Information

          Antibody detects endogenous levels of total LASS2.

          Target Information

          LASS2 is a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. This gene encodes a protein that has sequence similarity to yeast longevity assurance gene 1. Mutation or overexpression of the related gene in yeast has been shown to alter yeast lifespan. The human protein may play a role in the regulation of cell growth. Alternatively spliced transcript variants encoding the same protein have been described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ10243; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; longevity assurance (LAG1, S. cerevisiae) homolog 2; longevity assurance homolog 2; member of the TRAM superfamily; MGC987; sphingosine N-acyltransferase CERS2; TRAM homolog 3; translocating chain-associating membrane protein homolog 3; tumor metastasis-suppressor gene 1 protein; unnamed protein product; very-long-chain ceramide synthase CERS2

          View more View less

          Gene Aliases: 0610013I17Rik; L3; LASS2; SP260; TMSG1; TRH3

          View more View less

          UniProt ID: (Human) Q96G23, (Mouse) Q924Z4

          View more View less

          Entrez Gene ID: (Human) 29956, (Rat) 310667, (Mouse) 76893

          View more View less

          Function(s)
          DNA binding protein binding transferase activity, transferring acyl groups other than amino-acyl groups sphingosine N-acyltransferase activity
          Process(es)
          sphingolipid metabolic process glycosphingolipid biosynthetic process membrane budding response to wounding gene expression negative regulation of Schwann cell proliferation regulation of lipid metabolic process sphingolipid biosynthetic process ceramide biosynthetic process negative regulation of axon regeneration negative regulation of Schwann cell migration response to immobilization stress
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0