Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Positive Samples: HT-1080, SKOV3, A-549, 22Rv1, Mouse brain, Mouse skeletal muscle, Rat skeletal muscle
Immunogen sequence: MERAGPSFGQ QRQQQQPQQQ KQQQRDQDSV EAWLDDHWDF TFSYFVRKAT REMVNAWFAE RVHTIPVCKE GIRGHTESCS CPLQQSPRAD NSAPGTPTRK ISASEFDRPL RPIVVKDSEG TVSFLSDSEK KEQMPLTPPR FDHDEGDQCS
The cyclic monophosphate nucleotides (cyclic adenosine monophosphate [cAMP] and cyclic guanosine monophosphate [cGMP] are found ubiquitously in mammalian cells and act as second messenger transducers to effect the intracellular actions of a variety of G protein coupled receptors (GPCRs) for hormones, cytokines, and neurotransmitters. Cyclic nucleotides are important intracellular second messenger which play important role in a variety of signal transduction process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CGB-PDE; cGMP-binding cGMP specific phosphodiesterase 5A2; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-binding; cGMP-specific phosphodiesterase; cyclic nucleotide phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; PDE5A1; phosphodiesterase-5A1; PDE5A2; phosphodiesterase-5A2; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5; phosphodiesterase type 5; unnamed protein product
Gene Aliases: CGB-PDE; Cgbpde; CN5A; Cn5n; PDE5; Pde5a1; PDE5A2
UniProt ID: (Human) O76074, (Rat) O54735, (Mouse) Q8CG03
Entrez Gene ID: (Human) 8654, (Rat) 171115, (Mouse) 242202
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support