Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FGFR1/FGFR2 Antibodies

          Invitrogen

          Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Polyclonal Antibody

          View all (7) FGFR1/FGFR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Antibody (PA5-118586) in ICC/IF

          Immunocytochemical analysis of Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) in A549 cells(4h of LPS treatment) cells using a Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) polyclonal antibody (Product # PA5-118586). The samples were fixed with PFA and permeabilized in 0.1% Triton X-100,then blocked in 10% serum for 45 minutes at 25°C. Samples were then incubated with the primary antibody and mouse anti-beta tubulin Ab for 1 ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Antibody in Immunocytochemistry (ICC/IF)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-FGFR1/FGFR2 (Tyr463, Tyr466) Polyclonal Antibody

          Product Details

          PA5-118586

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human FGFR1/2 around the phosphorylation site of Tyr463/466.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2903086

          Product Specific Information

          Antibody detects endogenous levels of FGFR1/2 only when phosphorylated at Tyr463/466.

          Target Information

          FGFR1 is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein consists of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds both acidic and basic fibroblast growth factors and is involved in limb induction.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alternatively spliced; bacteria-expressed kinase; basic fibroblast growth factor receptor 1; BEK fibroblast growth factor receptor; FGF Receptor 2; FGF receptor precursor; FGF receptor-1; FGF receptor-1 precursor; fgfr 1; FGFR1-IIIb; receptor tyrosine kinase; FGFR1/PLAG1 fusion; FGFR2; fibroblast growth factor receptor (FGFr) transmembrane form; fibroblast growth factor receptor FGFR-1 secreted form; fibroblast growth factor-receptor precursor; FMS-like tyrosine kinase 2; fms-related tyrosine kinase 2; heparin-binding growth factor receptor; hydroxyaryl-protein kinase; keratinocyte growth factor receptor; orf 5' of fibroblast growth factor receptor 1 FGFR-1; protein tyrosine kinase, receptor like 14; proto-oncogene c-Fgr; soluble form; FGF-R1; Soluble KGFR; transmembrane protein tyrosine kinase; unnamed protein product

          View more View less

          Gene Aliases: BBDS; BEK; bFGF-R; bFGF-R-1; BFGFR; BFR-1; c-fgr; CD331; CD332; CEK; CEK3; CFD1; Eask; ECCL; ECT1; FGFBR; FGFR-1; Fgfr-2; Fgfr-7; FGFR-I; Fgfr2b; Fgfr7; FLG; FLT-2; FLT2; Fr1; HBGFR; HH2; HRTFDS; Hspy; JWS; K-SAM; KAL2; KGFR; KGFRTr; MFR; N-SAM; OGD; svs; TK14; TK25

          View more View less

          UniProt ID: (Human) P11362, (Human) P21802, (Mouse) P16092, (Rat) Q04589, (Mouse) P21803

          View more View less

          Entrez Gene ID: (Human) 2260, (Human) 2263, (Mouse) 14182, (Rat) 79114, (Mouse) 14183, (Rat) 25022

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity fibroblast growth factor-activated receptor activity protein binding ATP binding heparin binding fibroblast growth factor binding identical protein binding protein homodimerization activity receptor-receptor interaction receptor binding SH2 domain binding macromolecular complex binding cell adhesion molecule binding nucleotide binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter angiogenesis ureteric bud development in utero embryonic development organ induction epithelial to mesenchymal transition positive regulation of mesenchymal cell proliferation chondrocyte differentiation regulation of mitotic cell cycle brain development salivary gland morphogenesis sensory perception of sound positive regulation of cell proliferation fibroblast growth factor receptor signaling pathway gene expression regulation of gene expression regulation of cell cycle process negative regulation of gene expression positive regulation of cell communication positive regulation of fibroblast migration regulation of phosphate transport positive regulation of neuron projection development peptidyl-tyrosine phosphorylation stem cell population maintenance orbitofrontal cortex development postnatal olfactory bulb interneuron migration ventricular zone neuroblast division forebrain generation of neurons central nervous system neuron development tangential migration from the subventricular zone to the olfactory bulb positive regulation of signaling lung development embryonic limb morphogenesis epithelial cell differentiation midbrain development neuron projection development biomineral tissue development fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development regulation of cell proliferation inner ear morphogenesis outer ear morphogenesis middle ear morphogenesis regulation of apoptotic process positive regulation of MAP kinase activity positive regulation of MAPK cascade positive regulation of blood vessel endothelial cell migration ear development cellular response to fibroblast growth factor stimulus positive regulation of cell differentiation positive regulation of neuron differentiation negative regulation of osteoblast differentiation positive regulation of cell cycle positive regulation of transcription from RNA polymerase II promoter protein autophosphorylation paraxial mesoderm development regulation of lateral mesodermal cell fate specification cell maturation blood vessel morphogenesis generation of neurons mesenchymal cell differentiation regulation of phosphorus metabolic process positive regulation of protein kinase B signaling calcium ion homeostasis positive regulation of cardiac muscle cell proliferation auditory receptor cell development branching involved in salivary gland morphogenesis lung-associated mesenchyme development regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling vasculogenesis involved in coronary vascular morphogenesis vitamin D3 metabolic process diphosphate metabolic process cementum mineralization regulation of stem cell proliferation positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway negative regulation of fibroblast growth factor production regulation of postsynaptic density assembly positive regulation of mitotic cell cycle DNA replication response to sodium phosphate positive regulation of vascular endothelial cell proliferation positive regulation of hepatic stellate cell activation positive regulation of parathyroid hormone secretion positive regulation of endothelial cell chemotaxis regulation of extrinsic apoptotic signaling pathway in absence of ligand response to ischemia female pregnancy response to nutrient levels cellular response to lipopolysaccharide cellular response to growth factor stimulus cellular response to histamine cellular response to nerve growth factor stimulus MAPK cascade skeletal system development neuron migration protein phosphorylation positive regulation of phospholipase activity cell migration chordate embryonic development regulation of cell differentiation phosphatidylinositol-mediated signaling skeletal system morphogenesis outflow tract septum morphogenesis membranous septum morphogenesis endochondral bone growth cell-cell signaling axonogenesis negative regulation of cell proliferation regulation of smoothened signaling pathway post-embryonic development embryonic pattern specification organ morphogenesis negative regulation of keratinocyte proliferation morphogenesis of embryonic epithelium pyramidal neuron development gland morphogenesis positive regulation of Wnt signaling pathway bone mineralization otic vesicle formation hair follicle morphogenesis response to lipopolysaccharide lacrimal gland development regulation of osteoblast proliferation organ growth fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow fibroblast growth factor receptor signaling pathway involved in hemopoiesis fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow odontogenesis cell fate commitment response to ethanol regulation of osteoblast differentiation lung alveolus development mesodermal cell differentiation embryonic digestive tract morphogenesis embryonic organ morphogenesis digestive tract development embryonic organ development reproductive structure development embryonic cranial skeleton morphogenesis epidermis morphogenesis branching morphogenesis of a nerve regulation of epithelial cell proliferation positive regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation regulation of smooth muscle cell differentiation positive regulation of cell division ventricular cardiac muscle tissue morphogenesis limb bud formation bone development bone morphogenesis branching involved in prostate gland morphogenesis bud elongation involved in lung branching lung lobe morphogenesis positive regulation of epithelial cell proliferation involved in lung morphogenesis prostate gland morphogenesis prostate epithelial cord elongation prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis squamous basal epithelial stem cell differentiation involved in prostate gland acinus development lateral sprouting from an epithelium mammary gland bud formation epithelial cell proliferation involved in salivary gland morphogenesis branch elongation involved in salivary gland morphogenesis branching involved in labyrinthine layer morphogenesis regulation of morphogenesis of a branching structure mesenchymal cell differentiation involved in lung development mesenchymal cell proliferation involved in lung development regulation of ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade cellular response to retinoic acid cellular response to hypoxia cellular response to transforming growth factor beta stimulus positive regulation of canonical Wnt signaling pathway positive regulation of vascular smooth muscle cell proliferation cranial skeletal system development neuromuscular junction development positive regulation of gene expression forebrain development negative regulation of mitotic nuclear division synaptic vesicle transport regulation of developmental growth positive regulation of developmental process positive regulation of multicellular organismal process coronal suture morphogenesis epithelial tube branching involved in lung morphogenesis endodermal digestive tract morphogenesis lens fiber cell development fibroblast growth factor receptor apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-qfp4r:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0