Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD148 Antibodies

          Invitrogen

          CD148 Polyclonal Antibody

          View all (15) CD148 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD148 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (9)
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD148 Antibody (PA5-119082) in IHC (P)

          Immunohistochemistry (Paraffin) analysis of CD148 in human tonsil tissue using CD148 Polyclonal Antibody (Product # PA5-119082) (10 µg/mL). The sample was then stained with HRP-Goat anti-Rabbit IgG polyclonal secondary antibody (2 µg/mL). Vector Red staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD148 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD148 Antibody in Western Blot (WB)
          CD148 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CD148 Polyclonal Antibody

          Product Details

          PA5-119082

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.01-3 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          5-20 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein Density Enhanced Phosphatase 1. The antigen corresponds to amino acid range 567-842 of the target protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography, Protein A

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2903582

          Target Information

          CD148 is a highly glycosylated transmembrane protein of up to 250 kDa, containing eight fibronectin type III domains in the extracellular region and one tyrosine phosphatase domain in the cytoplasmic region. Expression is found broadly on fibroblasts, epithelial cells, lymphocytes (B, T and NK cells), and myeloid cells (monocytes and granulocytes). CD148 can dephosphorylate proteins such as LAT and PLC-gamma, and is involved in the downregulation of T cell activation. Conversely, CD148 expression can be upregulated following T cell activation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD148 antigen; colon tumor susceptibility 1; density-enhanced phosphatase 1; HPTP beta-like tyrosine phosphatase; human density enhanced phosphatase-1; protein tyrosine phosphatase, receptor type, J polypeptide; protein-tyrosine phosphatase eta; receptor tyrosine phosphatase; susceptibility to colon cancer 1; susceptibility to colon cancer 1, mouse, homolog of; unnamed protein product

          View more View less

          Gene Aliases: BET; Byp; CD148; DEP-1; DEP1; HPTP eta; HPTPeta; Ptpb2; PTPbeta2; R-PTP-ETA; R-PTP-J; RPTPJ; Scc-1; SCC1; THC10

          View more View less

          UniProt ID: (Human) Q12913, (Mouse) Q64455

          View more View less

          Entrez Gene ID: (Human) 5795, (Mouse) 19271

          View more View less

          Function(s)
          protein tyrosine phosphatase activity platelet-derived growth factor receptor binding protein binding beta-catenin binding phosphatase activity protein kinase binding gamma-catenin binding cadherin binding mitogen-activated protein kinase binding delta-catenin binding identical protein binding
          Process(es)
          vasculogenesis positive regulation of cell-matrix adhesion immune system process signal transduction axon guidance heart development blood coagulation negative regulation of cell proliferation anatomical structure morphogenesis positive regulation of platelet activation negative regulation of platelet-derived growth factor receptor signaling pathway positive regulation of macrophage chemotaxis hemopoiesis regulation of cell adhesion B cell differentiation platelet formation negative regulation of cell growth negative regulation of cell migration positive regulation of tumor necrosis factor production peptidyl-tyrosine dephosphorylation negative regulation of epidermal growth factor receptor signaling pathway glucose homeostasis negative regulation of vascular permeability negative regulation of MAP kinase activity positive regulation of MAPK cascade positive regulation of cell adhesion negative regulation of insulin receptor signaling pathway platelet-derived growth factor receptor signaling pathway oligodendrocyte differentiation positive regulation of phagocytosis positive regulation of immune response positive regulation of calcium-mediated signaling negative regulation of T cell receptor signaling pathway positive chemotaxis positive regulation of cellular component organization positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling negative regulation of protein kinase B signaling positive regulation of Fc receptor mediated stimulatory signaling pathway positive regulation of intracellular signal transduction peptidyl-tyrosine dephosphorylation involved in inactivation of protein kinase activity negative regulation of receptor activity cytokine-mediated signaling pathway T cell receptor signaling pathway contact inhibition
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0