Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TREM2 Antibodies

          Invitrogen

          TREM2 Polyclonal Antibody

          1 Published Figure
          2 References
          View all (41) TREM2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TREM2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Published Figures (1)
          TREM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          TREM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TREM2 Antibody (PA5-119690) in IHC (P)

          Immunohistochemistry (Paraffin) analysis of paraffin-embedded human colon carcinoma tissue using TREM2 Polyclonal Antibody (Product # PA5-119690). The section was pre-treated using heat mediated antigen retrieval with Tris-EDTA buffer (pH 8.0-8.4) for 20 minutes. The tissues were blocked in 5% BSA for 30 minutes at room temperature, washed with ddH2O and PBS, and then probed with the TREM2 antibody at a dil... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TREM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TREM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TREM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TREM2 Antibody in Western Blot (WB)
          TREM2 Antibody in Flow Cytometry (Flow)
          TREM2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TREM2 Polyclonal Antibody

          Product Details

          PA5-119690

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          1:50-1:100
          -
          Product Specifications

          Species Reactivity

          Human

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide within human TREM2 aa 30-60.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 0.2% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2913263

          Product Specific Information

          Positive Control: Mouse brain tissue lysates, human liver tissue, human liver carcinoma tissue, human colon carcinoma tissue, THP-1.

          Subcellular Location: Cell membrane, Secreted.

          Target Information

          TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; TREM-2; triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells 2a; unnamed protein product

          View more View less

          Gene Aliases: AD17; PLOSL2; TREM-2; Trem2a; Trem2b; Trem2c

          View more View less

          UniProt ID: (Human) Q9NZC2

          View more View less

          Entrez Gene ID: (Human) 54209

          View more View less

          Function(s)
          lipopolysaccharide binding beta-amyloid binding phosphatidylserine binding transmembrane signaling receptor activity protein binding phospholipid binding beta-catenin binding high-density lipoprotein particle binding lipid binding phosphatidylethanolamine binding semaphorin receptor activity kinase activator activity low-density lipoprotein particle binding semaphorin receptor binding apolipoprotein binding apolipoprotein A-I binding very-low-density lipoprotein particle binding signaling receptor activity peptidoglycan binding macromolecular complex binding lipoteichoic acid binding lipoprotein particle binding scaffold protein binding carbohydrate derivative binding sulfatide binding protein tyrosine kinase binding
          Process(es)
          microglial cell activation microglial cell activation involved in immune response T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell positive regulation of antigen processing and presentation of peptide antigen via MHC class II negative regulation of inflammatory response to antigenic stimulus response to ischemia phagocytosis phagocytosis, recognition immune response humoral immune response signal transduction cell surface receptor signaling pathway memory regulation of gene expression negative regulation of autophagy positive regulation of gene expression positive regulation of cholesterol efflux negative regulation of cholesterol storage negative regulation of sequestering of triglyceride positive regulation of high-density lipoprotein particle clearance regulation of lipid metabolic process osteoclast differentiation regulation of TOR signaling positive regulation of TOR signaling detection of lipopolysaccharide detection of peptidoglycan regulation of interleukin-6 production negative regulation of interleukin-1 beta production negative regulation of tumor necrosis factor production positive regulation of interleukin-10 production negative regulation of toll-like receptor 2 signaling pathway negative regulation of toll-like receptor 4 signaling pathway regulation of toll-like receptor 6 signaling pathway positive regulation of macrophage fusion negative regulation of glial cell apoptotic process social behavior CXCL12-activated CXCR4 signaling pathway defense response to bacterium negative regulation of apoptotic process negative regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of potassium ion transport apoptotic cell clearance regulation of innate immune response positive regulation of osteoclast differentiation respiratory burst after phagocytosis positive regulation of complement activation, classical pathway astrocyte activation cell development response to axon injury positive regulation of protein secretion positive regulation of phagocytosis defense response to Gram-negative bacterium positive regulation of calcium-mediated signaling negative regulation of cell activation positive regulation of chemotaxis positive regulation of protein kinase B signaling negative regulation of protein kinase B signaling lipid homeostasis regulation of resting membrane potential positive regulation of phagocytosis, engulfment microglial cell proliferation negative regulation of astrocyte activation pyroptosis negative regulation of fat cell proliferation positive regulation of ERK1 and ERK2 cascade detection of lipoteichoic acid cellular response to lipoteichoic acid cellular response to peptidoglycan cellular response to glucose stimulus cellular response to lipid cellular response to lipoprotein particle stimulus cellular response to hypoxia semaphorin-plexin signaling pathway regulation of macrophage inflammatory protein 1 alpha production dendritic cell differentiation dendritic spine maintenance beta-amyloid clearance import into cell regulation of hippocampal neuron apoptotic process regulation of plasma membrane bounded cell projection organization cellular response to oxidised low-density lipoprotein particle stimulus complement-mediated synapse pruning neuroinflammatory response positive regulation of neuroinflammatory response negative regulation of neuroinflammatory response amyloid-beta clearance by cellular catabolic process regulation of cytokine production involved in inflammatory response negative regulation of cytokine production involved in inflammatory response positive regulation of beta-amyloid clearance negative regulation of NLRP3 inflammasome complex assembly positive regulation of engulfment of apoptotic cell positive regulation of NIK/NF-kappaB signaling positive regulation of proteasomal protein catabolic process negative regulation of macrophage colony-stimulating factor signaling pathway positive regulation of intracellular signal transduction positive regulation of protein localization to plasma membrane positive regulation of C-C chemokine receptor CCR7 signaling pathway regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway negative regulation of p38MAPK cascade positive regulation of microglial cell activation negative regulation of autophagic cell death positive regulation of microglial cell migration cellular response to beta-amyloid positive regulation of establishment of protein localization positive regulation of CAMKK-AMPK signaling cascade positive regulation of low-density lipoprotein particle clearance excitatory synapse pruning positive regulation of synapse pruning negative regulation of amyloid fibril formation positive regulation of CD40 signaling pathway positive regulation of ATP biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline