Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Filamin A Antibodies

          Invitrogen

          Filamin A Polyclonal Antibody

          View all (30) Filamin A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Filamin A Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          Filamin A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Filamin A Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Filamin A Antibody (PA5-120445) in ICC/IF

          Immunofluorescence analysis of Filamin A in NIH/3T3 cells. Samples were incubated with Filamin A Polyclonal antibody (Product # PA5-120445) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Filamin A Antibody in Immunocytochemistry (ICC/IF)
          Filamin A Antibody in Immunocytochemistry (ICC/IF)
          Filamin A Antibody in Immunocytochemistry (ICC/IF)
          Filamin A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Filamin A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Filamin A Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Filamin A Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Filamin A Polyclonal Antibody

          Product Details

          PA5-120445

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 2348-2647 of human Filamin A (NP_001104026.1).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.51 mg/mL

          Amount

          51 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2914017

          Product Specific Information

          Positive test controls include: Jurkat, HeLa, NIH/3T3, 293T, U-251MG, Mouse lung. The target is usually found in the following locations: Cytoplasm, cell cortex, cytoskeleton.

          Immunogen sequence: NQPASFAVSL NGAKGAIDAK VHSPSGALEE CYVTEIDQDK YAVRFIPREN GVYLIDVKFN GTHIPGSPFK IRVGEPGHGG DPGLVSAYGA GLEGGVTGNP AEFVVNTSNA GAGALSVTID GPSKVKMDCQ ECPEGYRVTY TPMAPGSYLI SIKYGGPYHI GGSPFKAKVT GPRLVSNHSL HETSSVFVDS LTKATCAPQH GAPGPGPADA SKVVAKGLGL SKAYVGQKSS FTVDCSKAGN NMLLVGVHGP RTPCEEILVK HVGSRLYSVS YLLKDKGEYT LVVKWGDEHI PGSPYRVVVP

          Target Information

          Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: actin binding protein 280; actin binding protein 280; involved in actin cytoskeleton organization; actin-binding protein 280; alpha-filamin; dilated pupils 2; endothelial actin-binding protein; Endothelial actin-binding protein 280); epididymis secretory sperm binding protein; filamin 1; filamin A, alpha; filamin, alpha; filamin-1; Filamin1; FilaminA; non-muscle filamin; unnamed protein product

          View more View less

          Gene Aliases: ABP-280; ABPX; CSBS; CVD1; Dilp2; F730004A14Rik; FGS2; filamin-1; FLN; FLN-A; FLN1; FMD; GENA 379; MNS; NHBP; OPD; OPD1; OPD2; RGD1560614; XLVD; XMVD

          View more View less

          UniProt ID: (Human) P21333, (Mouse) Q8BTM8

          View more View less

          Entrez Gene ID: (Human) 2316, (Rat) 293860, (Mouse) 192176

          View more View less

          Function(s)
          G-protein coupled receptor binding RNA binding protein binding potassium channel regulator activity kinase binding small GTPase binding Fc-gamma receptor I complex binding protein homodimerization activity ion channel binding macromolecular complex binding cadherin binding SMAD binding actin filament binding GTPase binding DNA-binding transcription factor binding protein sequestering activity protein kinase C binding actin binding
          Process(es)
          angiogenesis epithelial to mesenchymal transition blood vessel remodeling heart morphogenesis adenylate cyclase-inhibiting dopamine receptor signaling pathway negative regulation of neuron projection development negative regulation of transcription from RNA polymerase I promoter formation of radial glial scaffolds cerebral cortex development actin cytoskeleton organization regulation of cell migration regulation of actin filament bundle assembly positive regulation of actin filament bundle assembly protein localization to cell surface negative regulation of protein catabolic process positive regulation of protein import into nucleus mRNA transcription from RNA polymerase II promoter negative regulation of apoptotic process receptor clustering wound healing, spreading of cells early endosome to late endosome transport establishment of protein localization cell-cell junction organization positive regulation of axon regeneration synapse organization protein stabilization actin crosslink formation cilium morphogenesis semaphorin-plexin signaling pathway protein localization to plasma membrane tubulin deacetylation mitotic spindle assembly establishment of Sertoli cell barrier positive regulation of substrate adhesion-dependent cell spreading positive regulation of potassium ion transmembrane transport protein localization to bicellular tight junction regulation of membrane repolarization during cardiac muscle cell action potential positive regulation of neural precursor cell proliferation positive regulation of integrin-mediated signaling pathway positive regulation of neuron migration blood coagulation, intrinsic pathway positive regulation of platelet activation megakaryocyte development positive regulation of I-kappaB kinase/NF-kappaB signaling release of sequestered calcium ion into cytosol regulation of membrane repolarization during atrial cardiac muscle cell action potential platelet aggregation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline