Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HIPK2 Antibodies

          Invitrogen

          Phospho-HIPK2 (Tyr361) Polyclonal Antibody

          Advanced Verification
          7 Published Figures
          7 References
          View all (21) HIPK2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-HIPK2 (Tyr361) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (7)
          • Advanced Verification (1)
          Phospho-HIPK2 (Tyr361) Antibody in Dot Blot (DB)
          Group 53 Created with Sketch.
          Phospho-HIPK2 (Tyr361) Antibody in Dot Blot (DB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-HIPK2 (Tyr361) Antibody (PA5-13045) in DB

          Dot blot analysis of Phospho-HIPK2 (Tyr361). Samples were incubated with Phospho-HIPK2 (Tyr361) polyclonal antibody (Product # PA5-13045) using a dilution of 0.5 µg/mL. 50 ng of Phospho-peptide or Non Phospho-peptide per dot were adsorbed on nitrocellulose membrane. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-HIPK2 (Tyr361) Antibody in Dot Blot (DB)
          Phospho-HIPK2 (Tyr361) Antibody in Western Blot (WB)
          Phospho-HIPK2 (Tyr361) Antibody in Western Blot (WB)
          Phospho-HIPK2 (Tyr361) Antibody in Western Blot (WB)
          Phospho-HIPK2 (Tyr361) Antibody in Western Blot (WB)
          Phospho-HIPK2 (Tyr361) Antibody in Western Blot (WB)
          Phospho-HIPK2 (Tyr361) Antibody in Western Blot (WB)
          Phospho-HIPK2 (Tyr361) Antibody in Immunoprecipitation (IP)
          Phospho-HIPK2 (Tyr361) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-HIPK2 (Tyr361) Polyclonal Antibody

          Product Details

          PA5-13045

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Dot blot (DB)

          1:500
          -
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y361 of human HIPK2
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          200 µg

          Purification

          Protein A, Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10987115

          Product Specific Information

          This antibody is predicted to react with mouse and rat based on sequence homology.

          Target Information

          The HIPK2 gene encodes a member of the homeodomain-interacting protein kinase family, specifically a serine/threonine nuclear protein kinase. HIPK2 plays versatile roles in cellular processes, including transcription regulation, cell proliferation, differentiation, and apoptosis. It interacts with various transcription factors and proteins, contributing significantly to the modulation of gene expression. Notably, HIPK2 is involved in the activation of the tumor suppressor protein p53, with p53-dependent pro-apoptotic activity, positioning HIPK2 as a potential tumor suppressor gene. Recent studies highlight its downregulation in multiple tumor types and its emerging role in cancer biology and therapy. Its expression is subject to quality control mechanisms, including autophosphorylation at serine and threonine sites. Structurally, the crystal structure of HIPK2 reveals unique conformational attributes that contribute to its functionality in different signaling pathways. HIPK2's influence extends to physiological processes, such as fibrosis and central nervous system regulation, indicating its broad biological significance and potential therapeutic applications.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: hHIPk2; HIPK2; predicted protein of HQ0593

          View more View less

          Gene Aliases: PRO0593

          View more View less

          UniProt ID: (Human) Q9H2X6

          View more View less

          Entrez Gene ID: (Human) 28996

          View more View less

          Function(s)
          transcription coactivator activity transcription corepressor activity protein kinase activity protein serine/threonine kinase activity protein tyrosine kinase activity protein binding ATP binding SMAD binding virion binding RNA polymerase II sequence-specific DNA binding transcription factor binding protein serine kinase activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter eye development protein phosphorylation smoothened signaling pathway retina layer formation peptidyl-serine phosphorylation neuron differentiation erythrocyte differentiation DNA damage response, signal transduction by p53 class mediator positive regulation of transforming growth factor beta receptor signaling pathway negative regulation of BMP signaling pathway PML body organization regulation of gene expression, epigenetic intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator positive regulation of angiogenesis positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade embryonic camera-type eye morphogenesis regulation of cell cycle embryonic retina morphogenesis in camera-type eye lens induction in camera-type eye SMAD protein signal transduction iris morphogenesis cellular response to hypoxia intrinsic apoptotic signaling pathway regulation of signal transduction by p53 class mediator negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0