Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • DJ-1 Antibodies

          Invitrogen

          DJ-1 Polyclonal Antibody

          View all (33) DJ-1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DJ-1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          DJ-1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DJ-1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DJ-1 Antibody (PA5-143504) in ICC/IF

          Immunofluorescent analysis of DJ-1 in SH-SY5Y cells. Samples were stained with DJ-1 polyclonal antibody (Product # PA5-143602) using a dilution of 1:1,000 (red). Costained with mouse mAb to actin (Product # MA5-47454) dilution 1:200 in green. The blue is Hoechst staining of nuclear DNA. The DJ-1 antibody reveals strong cytoplasmic staining for DJ1 protein, while the actin antibody labels the submembranous actin-rich cytoskeleton, stress fibers and bundles of actin associated with cell adhesion sites. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DJ-1 Antibody in Immunocytochemistry (ICC/IF)
          DJ-1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          DJ-1 Polyclonal Antibody

          Product Details

          PA5-143504

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          -

          Immunohistochemistry (IHC)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Full length recombinant human DJ1 expressed in and purified from E. coli.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          5mM sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942733

          Target Information

          The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CAP1; contraception-associated protein 1; CTA-215D11.1; early onset 7; epididymis secretory sperm binding protein Li 67p; fertility protein SP22; FLJ27376; FLJ34360; FLJ92274; maillard deglycase; oncogene DJ1; oncogene Dj1; Parkinson disease protein 7; OTTHUMP00000001350; OTTHUMP00000001351; PARK7 GENE; PARK7/DJ1; Parkinson disease (autosomal recessive, early onset) 7; parkinson disease protein 7 homolog; parkinson protein 7; parkinsonism-associated deglycase; protein deglycase DJ-1; protein DJ-1; protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit; unnamed protein product

          View more View less

          Gene Aliases: CAP1; DJ-1; DJ1; GATD2; HEL-S-67p; SP22

          View more View less

          UniProt ID: (Human) Q99497, (Mouse) Q99LX0, (Rat) O88767

          View more View less

          Entrez Gene ID: (Human) 11315, (Mouse) 57320, (Rat) 117287

          View more View less

          Function(s)
          core promoter sequence-specific DNA binding double-stranded DNA binding single-stranded DNA binding transcription coactivator activity mRNA binding receptor binding copper ion binding protein binding enzyme activator activity peptidase activity superoxide dismutase copper chaperone activity oxidoreductase activity, acting on peroxide as acceptor oxygen sensor activity enzyme binding kinase binding cytokine binding peptidase inhibitor activity receptor activator activity tyrosine 3-monooxygenase activator activity L-dopa decarboxylase activator activity protein deglycase activity identical protein binding protein homodimerization activity small protein activating enzyme binding ubiquitin-like protein conjugating enzyme binding cadherin binding mercury ion binding androgen receptor binding peroxiredoxin activity ubiquitin-protein transferase inhibitor activity scaffold protein binding DNA-binding transcription factor binding cupric ion binding cuprous ion binding ubiquitin-specific protease binding glyoxalase (glycolic acid-forming) activity RNA binding
          Process(es)
          synaptic transmission, dopaminergic positive regulation of acute inflammatory response to antigenic stimulus DNA repair response to oxidative stress mitochondrion organization adult locomotory behavior methylglyoxal metabolic process detoxification of copper ion positive regulation of gene expression negative regulation of gene expression negative regulation of smooth muscle cell migration lactate biosynthetic process removal of superoxide radicals insulin secretion protein repair androgen receptor signaling pathway positive regulation of protein complex assembly negative regulation of protein ubiquitination negative regulation of proteasomal ubiquitin-dependent protein catabolic process positive regulation of interleukin-8 production negative regulation of protein sumoylation cellular response to stress positive regulation of NAD(P)H oxidase activity cellular response to oxidative stress cellular response to reactive oxygen species cellular response to glyoxal negative regulation of protein catabolic process response to hydrogen peroxide glucose homeostasis hydrogen peroxide metabolic process regulation of neuron apoptotic process negative regulation of neuron apoptotic process positive regulation of transcription from RNA polymerase II promoter glycolate biosynthetic process negative regulation of protein export from nucleus regulation of inflammatory response detoxification of mercury ion protein stabilization dopamine uptake involved in synaptic transmission regulation of mitochondrial membrane potential positive regulation of protein kinase B signaling membrane depolarization membrane hyperpolarization regulation of androgen receptor signaling pathway detoxification of hydrogen peroxide cellular response to hydrogen peroxide detection of oxidative stress guanine deglycation guanine deglycation, glyoxal removal cellular detoxification of aldehyde cellular detoxification of methylglyoxal positive regulation of protein localization to nucleus regulation of synaptic vesicle endocytosis cellular response to oxygen-containing compound negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of mitochondrial electron transport, NADH to ubiquinone negative regulation of death-inducing signaling complex assembly negative regulation of protein K48-linked deubiquitination negative regulation of TRAIL-activated apoptotic signaling pathway positive regulation of dopamine biosynthetic process glyoxal metabolic process glyoxal catabolic process regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway negative regulation of hydrogen peroxide-induced neuron intrinsic apoptotic signaling pathway negative regulation of reactive oxygen species biosynthetic process positive regulation of reactive oxygen species biosynthetic process negative regulation of intrinsic apoptotic signaling pathway in response to hydrogen peroxide negative regulation of vascular smooth muscle cell proliferation negative regulation of NMDA glutamate receptor activity negative regulation of nitrosative stress-induced intrinsic apoptotic signaling pathway positive regulation of fertilization positive regulation of oxidative phosphorylation uncoupler activity positive regulation of reactive oxygen species metabolic process negative regulation of extrinsic apoptotic signaling pathway spermatogenesis response to xenobiotic stimulus fertilization regulation of TRAIL production maternal process involved in female pregnancy cellular response to lipopolysaccharide cellular response to hypoxia Ras protein signal transduction response to testosterone regulation of fibril organization positive regulation of mitophagy
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0