Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CYP11B1/CYP11B2 Antibodies

          Invitrogen

          CYP11B1/CYP11B2 Polyclonal Antibody

          View all (3) CYP11B1/CYP11B2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CYP11B1/CYP11B2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CYP11B1/CYP11B2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CYP11B1/CYP11B2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CYP11B1/CYP11B2 Antibody (PA5-143908) in WB

          Western blot analysis of CYP11B1/C11B2/CYP11B2 using CYP11B1/C11B2/CYP11B2 antibody (Product # PA5-143908). Electrophoresis was performed on a 5-20% SDS-PAGE gel at 70V (Stacking gel) / 90V (Resolving gel) for 2-3 hours. The sample well of each lane was loaded with 50 µg of sample under reducing conditions. Lane 1: rat kidney tissue lysates; Lane 2: mouse kidney tissue lysates. After Electrophoresis, proteins were transferred to a Nitrocellulose membrane at 150mA for 50-90 minutes. Blocked the membrane with 5% Non-fat Milk/ TBS for 1.5 hou... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CYP11B1/CYP11B2 Antibody in Western Blot (WB)
          CYP11B1/CYP11B2 Antibody in Flow Cytometry (Flow)
          CYP11B1/CYP11B2 Polyclonal Antibody

          Product Details

          PA5-143908

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.25-0.5 µg/mL
          -

          Flow Cytometry (Flow)

          1-3 µg/10x^6 cells
          -

          ELISA (ELISA)

          0.1-0.5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived human CYP11B1/C11B2/CYP11B2 recombinant protein (Position: M1-P491).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 4mg trehalose

          Contains

          0.01mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3075122

          Product Specific Information

          Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.

          Positive Control - WB: rat kidney tissue, mouse kidney tissue. Flow: 293T cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

          Target Information

          The steroid 11beta-hydroxylase gene, also designated Cyp11b-1, is a marker for the functional differentiation of cells in the zonae fasciculata reticularis. The deduced protein CYP11B1 consists of 466 amino acids containing a secretory signal, epidermal growth factor-like repeats, and a proteolytically inactive cathepsin B-related sequence. A related protein, human aldosterone synthase (CYP11B2), is involved in substrate recognition and conversion, with a functionally significant residue 112 in the N-terminal region of human CYP11B2. The inherited disorder glucocorticoid-remediable aldosteronism is caused by a chimeric gene duplication between the CYP11B1 and CYP11B2 genes. This disorder is characterized by hyperaldosteronism and high levels of 18-hydroxycortisol and 18-oxocortisol, which are under ACTH control.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALDOS; Aldosterone synthase; Aldosterone-synthesizing enzyme; Corticosterone 18-monooxygenase, CYP11B2; CYP11B1; CYP11B1-V316M; CYPXIB1; CYPXIB2; CYPXIB3; Cytochrome P-450Aldo; Cytochrome P-450c11; Cytochrome P-450C18; Cytochrome P450 11B1, mitochondrial; Cytochrome P450 11B2, mitochondrial; cytochrome P450 11B3, mitochondrial; Cytochrome P450 subfamily XIB polypeptide 2 (aldosterone synthase); cytochrome p450 XIB1; cytochrome P450(11 beta)-DS; cytochrome P450, family 11, subfamily B, polypeptide 1; cytochrome P450, family 11, subfamily B, polypeptide 2; cytochrome P450, subfamily 11B, polypeptide 1; cytochrome P450, subfamily 11B, polypeptide 2; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2; Cytochrome P450, subfamily XIB, polypeptide 1 (steroid 11-beta-hydroxylase); Cytochrome P450, subfamily XIB, polypeptide 2 (aldosterone synthase); cytochrome P450-Aldo-1; cytochrome P450-Aldo-2; Cytochrome P450C11; mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2; P450(11 beta)-DS; P450-Aldo-1; P450-Aldo-2; P450C11; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B1; Steroid 11-beta-hydroxylase, CYP11B2; steroid 11-beta-monooxygenase; steroid 11-beta/18-hydroxylase; Steroid 18-hydroxylase; steroid 18-hydroxylase, aldosterone synthase, P450C18, P450aldo; steroid-11-beta-hydroxylase; unnamed protein product

          View more View less

          Gene Aliases: AA389061; ALDOS; Cp11ba; Cp45as; CPN1; CPN2; CYP11B; Cyp11b-1; Cyp11b-2; CYP11B1; CYP11B2; Cyp11b3; CYP11BL; CYPXIB2; FHI; P-450C18; P450aldo; P450C11; P450C18; RATCP11BA; RNCP45AS; S11BH

          View more View less

          UniProt ID: (Human) P15538, (Human) P19099, (Mouse) P15539

          View more View less

          Entrez Gene ID: (Human) 1584, (Human) 1585, (Mouse) 110115, (Rat) 500892, (Mouse) 13072, (Rat) 24294

          View more View less

          Function(s)
          monooxygenase activity steroid 11-beta-monooxygenase activity iron ion binding oxidoreductase activity oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen heme binding metal ion binding corticosterone 18-monooxygenase activity steroid hydroxylase activity steroid binding
          Process(es)
          regulation of blood volume by renal aldosterone C21-steroid hormone biosynthetic process glucocorticoid biosynthetic process aldosterone biosynthetic process potassium ion homeostasis sodium ion homeostasis oxidation-reduction process regulation of systemic arterial blood pressure by circulatory renin-angiotensin steroid biosynthetic process regulation of blood pressure cortisol biosynthetic process drinking behavior positive regulation of blood pressure ion homeostasis response to nutrient response to salt stress cellular response to nutrient response to insulin cellular response to hormone stimulus cellular response to potassium ion response to drug response to peptide hormone cellular response to potassium ion starvation cellular response to mechanical stimulus cellular response to peptide hormone stimulus alcohol metabolic process lipid metabolic process immune response cholesterol metabolic process sterol metabolic process cortisol metabolic process glucose homeostasis renal water homeostasis mineralocorticoid biosynthetic process steroid metabolic process glucocorticoid metabolic process dibenzo-p-dioxin metabolic process adenosine metabolic process cellular response to interferon-gamma response to dexamethasone
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-hn5gs:80/100.66.79.229:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline