Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EHD3 Antibodies

          Invitrogen

          EHD3 Polyclonal Antibody

          View all (13) EHD3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite EHD3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (13)
          EHD3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          EHD3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          EHD3 Antibody (PA5-143924) in ICC/IF

          Immunocytochemistry analysis of EHD3 in MCF-7 cells. Enzyme antigen retrieval was performed using IHC enzyme antigen retrieval reagent for 15 mins. The cells were blocked with 10% goat serum. Samples were then incubated in EHD3 Polyclonal antibody (Product # PA5-143924) using a dilution of 5 μg/mL. DyLight®488 Conjugated Goat Anti-Rabbit IgG was used as secondary antibody at 1:100 dilution and incubated for 30 minutes at 37°C. The section was counterstained with DAPI. Visualize using a fluorescence microscope and filter sets appropriate fo... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          EHD3 Antibody in Immunocytochemistry (ICC/IF)
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          EHD3 Antibody in Western Blot (WB)
          EHD3 Antibody in Western Blot (WB)
          EHD3 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          EHD3 Polyclonal Antibody

          Product Details

          PA5-143924

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.25 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          2-5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          -

          Flow Cytometry (Flow)

          1-3 µg/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to a sequence of human EHD3 (NELPAHLLPPSKRKVAE).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 4mg trehalose

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3075138

          Product Specific Information

          Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.

          Positive Control - WB: human placenta tissue, human A549 whole cell, human Hela whole cell, human CCRF-CEM whole cell, human U-87MG whole cell, human SY-SY5Y whole cell, monkey heart tissue, rat heart tissue, rat brain tissue, rat kidney tissue, rat liver tissue, mouse heart tissue, mouse brain tissue, mouse kidney tissue, mouse liver tissue. IHC: mouse brain tissue, human ovarian serous adenocarcinoma tissue, human ovarian serous adenocarcinoma tissue, human renal clear cell carcinoma tissue, human breast cancer tissue, human lung cancer tissue, human rectal cancer tissue, human gastric adenocarcinoma tissue, rat brain tissue. ICC/IF: MCF-7 cell. Flow: MCF-7 cell.|Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

          Target Information

          The EHD3 gene encodes the EH-domain containing protein 3, part of the EHD protein family involved in the regulation of endocytic recycling and membrane trafficking. EHD3 plays a crucial role in the endosomal transport system, impacting how receptors and other membrane proteins are recycled back to the cell surface after internalization. This protein aids in maintaining cellular homeostasis by ensuring the efficient sorting and transport of cargo through the endosomal system. EHD3 has been implicated in various cellular functions, including insulin signaling, receptor recycling, and neuronal development, and significantly influences synaptic transmission and plasticity. Alterations in EHD3 expression or function can disrupt endosomal trafficking processes and are linked to conditions such as neurodegenerative diseases and cancer. Research on EHD3 is focused on uncovering its role in membrane dynamics and identifying potential therapeutic strategies for disorders involving dysfunctional endocytic pathways.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EH-domain containing protein 2; EHD2; PAST homolog 3; similar to EHD1; unnamed protein product

          View more View less

          Gene Aliases: Ehd2; PAST3

          View more View less

          UniProt ID: (Human) Q9NZN3, (Rat) Q8R491, (Mouse) Q9QXY6

          View more View less

          Entrez Gene ID: (Human) 30845, (Rat) 192249, (Mouse) 57440

          View more View less

          Function(s)
          nucleic acid binding calcium ion binding protein binding GTP binding protein binding, bridging
          Process(es)
          receptor recycling endocytosis endocytic recycling early endosome to Golgi transport protein homooligomerization regulation of cardiac muscle contraction cilium morphogenesis protein localization to plasma membrane regulation of cardiac muscle cell membrane potential Golgi to lysosome transport positive regulation of voltage-gated calcium channel activity regulation of Golgi organization regulation of cardiac conduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline