Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Reconstitute at 0.2 mg/mL in sterile PBS.
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne's syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic transcription factor 2 89 kDa subunit; DNA 3'-5' helicase/translocase XPB; DNA excision repair protein ERCC-3; DNA repair gene; DNA repair helicase; DNA repair protein complementing XP-B cells; excision repair cross-complementation group 3; excision repair cross-complementing rodent repair deficiency, complementation group 3; general transcription and DNA repair factor IIH helicase subunit XPB; TFIIH 89 kDa subunit; TFIIH basal transcription factor complex 89 kDa subunit; TFIIH basal transcription factor complex helicase XPB subunit; TFIIH subunit XPB; unnamed protein product; xeroderma pigmentosum group B-complementing protein; xeroderma pigmentosum, complementation group B
Gene Aliases: BTF2; BTF2 p89; Ercc-3; GTF2H; RAD25; Ssl2; TFIIH; TTD2; XPB
UniProt ID: (Human) P19447, (Mouse) P49135, (Rat) Q4G005
Entrez Gene ID: (Human) 2071, (Mouse) 13872, (Rat) 291703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support