Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • RACK1 Antibodies

          Invitrogen

          RACK1 Polyclonal Antibody

          Advanced Verification
          2 Published Figures
          3 References
          View all (20) RACK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite RACK1 Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (2)
          • Advanced Verification (1)
          RACK1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RACK1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          RACK1 Antibody (PA5-17484) in ICC/IF

          Immunofluorescence analysis of RACK1 was performed using 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with RACK1 Polyclonal Antibody (Product # PA5-17484) at 1:200 dilution in 0.1% BSA, incubated at 4 degree Celsius overnight and then labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product # A2703... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          RACK1 Antibody in Immunocytochemistry (ICC/IF)
          RACK1 Antibody in Western Blot (WB)
          RACK1 Antibody in Western Blot (WB)
          RACK1 Antibody in Western Blot (WB)
          RACK1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          RACK1 Antibody in Western Blot (WB)
          RACK1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-17484

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:200
          -
          Product Specifications

          Species Reactivity

          Fruit fly, Human, Mouse, Rat, Zebrafish

          Published species

          Yeast

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to human RACK1

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          29 µg/mL

          Amount

          2.9 µg

          Purification

          Antigen affinity chromatography, Protein A

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10980517

          Product Specific Information

          It is not recommended to aliquot this antibody.

          Target Information

          RACK1 (receptor for activated C kinase 1) was identified through its binding to various PKC isoforms. Its main function is to recruit PKC and various other proteins to specific locations to form multiprotein complexes, mediating various signal pathways. RACK1 is required for VANGL2 membrane localization and for PTK2/FAK1 phosphorylation and dephosphorylation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 12-3; activated protein kinase C receptor; cell proliferation-inducing gene 21 protein; CG7111-PA; CG7111-PB; CG7111-PC; CG7111-PD; GNB2L1; guanine nucleotide binding protein (G protein) beta polypeptide 2-like 1; guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1 sequence 1; guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1; guanine nucleotide binding protein beta polypeptide 2-like 1; guanine nucleotide binding protein related; guanine nucleotide binding protein, beta 2, related sequence 1; guanine nucleotide binding protein, beta polypeptide 2-like 1; guanine nucleotide-binding protein beta-subunit like protein; guanine nucleotide-binding protein subunit beta-2-like 1; guanine nucleotide-binding protein subunit beta-like protein 12.3; human lung cancer oncogene 7 protein; lung cancer oncogene 7; OTTHUMP00000223870; OTTHUMP00000223891; OTTHUMP00000223893; OTTHUMP00000223900; OTTHUMP00000223901; OTTHUMP00000223902; OTTHUMP00000223930; OTTHUMP00000223931; PIG21; proliferation-inducing gene 21; protein homologous to chicken B complex protein, guanine nucleotide binding; protein kinase C receptor; RACK; rack-1; Rack1-PA; Rack1-PB; Rack1-PC; Rack1-PD; RE74715p; receptor for activated C kinase 1; receptor for activated protein kinase C; receptor for activated protein kinase C1; receptor of activated PKC; receptor of activated protein C kinase 1; receptor of activated protein kinase C; receptor of activated protein kinase C 1; similar to mammalian RACK1; unnamed protein product

          View more View less

          Gene Aliases: CG7111; Dmel\CG7111; Dmel_CG7111; dRack1; GB-like; Gnb2-rs1; GNB2L1; H12.3; HLC-7; i173; p205; PIG21; RACK1; wu:fb80d08; wu:fk65d12

          View more View less

          UniProt ID: (Human) P63244, (Mouse) P68040, (Fruit fly) O18640, (Rat) P63245, (Zebrafish) O42248

          View more View less

          Entrez Gene ID: (Human) 10399, (Mouse) 14694, (Fruit fly) 34070, (Rat) 83427, (Zebrafish) 30722

          View more View less

          Function(s)
          RNA binding protein kinase C binding receptor binding protein binding enzyme activator activity ion channel inhibitor activity enzyme binding protein phosphatase binding protein serine/threonine kinase inhibitor activity protein tyrosine kinase inhibitor activity cyclin binding receptor tyrosine kinase binding signaling adaptor activity SH2 domain binding identical protein binding protein homodimerization activity ribosome binding translation regulator activity cadherin binding BH3 domain binding binding, bridging cysteine-type endopeptidase activator activity involved in apoptotic process signaling receptor activity
          Process(es)
          positive regulation of protein phosphorylation translation apoptotic process protein localization negative regulation of gene expression protein ubiquitination negative regulation of translation negative regulation of Wnt signaling pathway negative regulation of cell growth positive regulation of cell migration positive regulation of protein complex assembly positive regulation of proteasomal ubiquitin-dependent protein catabolic process regulation of protein localization tumor necrosis factor-mediated signaling pathway intracellular signal transduction positive regulation of Golgi to plasma membrane protein transport positive regulation of apoptotic process pigmentation positive regulation of GTPase activity negative regulation of smoothened signaling pathway negative regulation of phagocytosis regulation of cell division regulation of cell cycle negative regulation of protein kinase B signaling positive regulation of mitochondrial depolarization cellular response to glucose stimulus cellular response to growth factor stimulus rescue of stalled ribosome regulation of adenylate cyclase-activating G protein-coupled receptor signaling pathway regulation of protein localization to plasma membrane negative regulation of intrinsic apoptotic signaling pathway in response to hydrogen peroxide regulation of establishment of cell polarity positive regulation of gastrulation negative regulation of translational frameshifting positive regulation of intrinsic apoptotic signaling pathway mRNA splicing, via spliceosome negative regulation of protein phosphorylation locomotory behavior cellular response to starvation regulation of autophagosome size oviposition positive regulation of actin filament polymerization wing disc development cuticle development positive regulation by host of viral genome replication positive regulation of glycogen biosynthetic process muscle cell cellular homeostasis oogenesis IRES-dependent viral translational initiation angiogenesis convergent extension involved in gastrulation osteoblast differentiation phospholipase C-activating G-protein coupled receptor signaling pathway cytoplasmic translation negative regulation of protein binding negative regulation of endoplasmic reticulum unfolded protein response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-x4rxd:80/100.66.76.16:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0