Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PKC alpha/beta-2 Antibodies

          Invitrogen

          Phospho-PKC alpha/beta-2 (Thr638, Thr641) Polyclonal Antibody

          View all (4) PKC alpha/beta-2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-PKC alpha/beta-2 (Thr638, Thr641) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}

          Product Details

          PA5-17904

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunoprecipitation (IP)

          1:100
          -
          Product Specifications

          Species Reactivity

          Hamster, Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phosphopeptide corresponding to residues surrounding pThr638 of human PKC alpha

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10989347

          Product Specific Information

          It is not recommended to aliquot this antibody.

          Target Information

          Protein Kinase c alpha (PKC alpha) is an 77 kDa member of the conventional group (cPKCs: sensitive to calcium, diacylglycerol, phosphatidylserine and phorbol esters) of the PKC family of serine/ threonine kinases that are involved in a wide range of physiological processes including mitogenesis, cell survival and transcriptional regulation. PKC alpha is an ubiquitously expressed PKC isozyme that has been implicated in the regulation of a broad range of cellular functions including proliferation, differentiation, development, migration, cell cell adhesion, cell extracellular matrix adhesion, and solute transport. The activation loop threonine (threonine 497 in PKC alpha) of conventional PKCs is phosphorylated by phosphoinositide dependent kinase 1 (PDK1). This phosphorylation is necessary for the autophosphorylation of threonine 638 in the carboxy terminus of PKC alpha, a step that is critical for regulating the rate of PKC alpha dephosphorylation and inactivation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AAG6; aging-associated gene 6; PKC-A; PKC-alpha; PKC-B; PKC-beta; PRKC Pan; protein kinase; protein kinase C alpha-polypeptide; protein kinase C beta I; protein kinase C beta II; protein kinase C beta-1; protein kinase C beta-II; protein kinase C type III; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide; protein kinase c-alpha; unnamed protein product

          View more View less

          Gene Aliases: AAG6; PKC-alpha; PKC-B; PKC-Beta; PKCA; PKCalpha; PKCB; PKCbeta; PKCI(2); PKCI+/-; PRKCB1; PRKCB2

          View more View less

          UniProt ID: (Human) P17252, (Human) P05771, (Mouse) P20444, (Rat) P05696, (Rat) P68403, (Mouse) P68404

          View more View less

          Entrez Gene ID: (Human) 5578, (Human) 5579, (Mouse) 18750, (Rat) 24680, (Rat) 25023, (Mouse) 18751

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity protein kinase C activity calcium-dependent protein kinase C activity receptor binding integrin binding protein binding ATP binding zinc ion binding enzyme binding diacylglycerol binding histone kinase activity (H3-T6 specific) protein serine kinase activity chromatin binding transcription coactivator activity protein kinase C binding calcium channel regulator activity histone binding androgen receptor binding lipid binding
          Process(es)
          response to reactive oxygen species negative regulation of protein phosphorylation positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation regulation of the force of heart contraction chondrocyte differentiation desmosome assembly chromatin remodeling protein phosphorylation negative regulation of protein kinase activity cellular calcium ion homeostasis regulation of muscle contraction learning or memory negative regulation of cell proliferation positive regulation of signal transduction positive regulation of endothelial cell migration positive regulation of cardiac muscle hypertrophy negative regulation of glucose transport negative regulation of translation central nervous system neuron axonogenesis signal transduction involved in regulation of gene expression positive regulation of cell migration neutrophil chemotaxis positive regulation of lipopolysaccharide-mediated signaling pathway response to estradiol negative regulation of glial cell apoptotic process intracellular signal transduction peptidyl-serine autophosphorylation proteasome-mediated ubiquitin-dependent protein catabolic process negative regulation of MAPK cascade protein kinase B signaling positive regulation of blood vessel endothelial cell migration positive regulation of cellular component biogenesis response to ethanol positive regulation of macrophage differentiation positive regulation of angiogenesis positive regulation of bone resorption positive regulation of cell adhesion positive regulation of exocytosis positive regulation of mitotic cell cycle negative regulation of glucose import negative regulation of insulin receptor signaling pathway protein autophosphorylation positive regulation of smooth muscle cell proliferation positive regulation of inflammatory response regulation of peptidyl-tyrosine phosphorylation induction of positive chemotaxis positive regulation of developmental process positive regulation of multicellular organismal process positive regulation of protein kinase B signaling positive regulation of synapse assembly renal sodium ion absorption positive regulation of ERK1 and ERK2 cascade protein kinase C signaling response to interleukin-1 cellular response to carbohydrate stimulus regulation of platelet aggregation intrinsic apoptotic signaling pathway positive regulation of angiotensin-activated signaling pathway negative regulation of cytokine production involved in inflammatory response response to phorbol 13-acetate 12-myristate regulation of synaptic vesicle exocytosis positive regulation of dense core granule biogenesis regulation of transcription from RNA polymerase II promoter mitotic nuclear envelope disassembly signal transduction phospholipase C-activating G-protein coupled acetylcholine receptor signaling pathway regulation of glucose transport positive regulation of vascular endothelial growth factor receptor signaling pathway B cell activation lipoprotein transport positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of transcription, DNA-templated B cell receptor signaling pathway regulation of transport negative regulation of dopamine receptor signaling pathway positive regulation of ferroptosis response to mechanical stimulus response to toxic substance positive regulation of insulin secretion response to peptide hormone regulation of mRNA stability response to corticosterone apoptotic signaling pathway positive regulation of adenylate cyclase-activating G protein-coupled receptor signaling pathway response to hypoxia calcium ion transport response to xenobiotic stimulus response to glucose regulation of signal transduction regulation of dopamine secretion dibenzo-p-dioxin metabolic process response to vitamin D regulation of growth positive regulation of odontogenesis of dentin-containing tooth regulation of secretion by cell response to angiotensin regulation of exocytosis establishment of protein localization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-gglmw:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0