Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ARC Antibodies

          Invitrogen

          ARC Polyclonal Antibody

          View all (31) ARC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ARC Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          ARC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ARC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ARC Antibody (PA5-19934) in WB

          Western Blot analysis of ARC in HeLa whole cell lysates with ARC Polyclonal Antibody (Product # PA5-19934) at 1:500 dilution. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ARC Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          ARC Polyclonal Antibody

          Product Details

          PA5-19934

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A peptide corresponding to amino acids 2 to 18 of human origin
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11154320

          Product Specific Information

          In Western blot applications, this antibody detects a band at ~25kDa. A suggested positive control is Hela cell lysate.

          PA5-19934 can be used with blocking peptide PEP-0059.

          Target Information

          ARC (also known as NOL3 - Nucleolar protein 3) has two isoforms. Isoform 1 is involved in RNA splicing. Isoform 2 functions as an apoptosis repressor that blocks multiple modes of cell death. ARC inhibits extrinsic apoptotic pathways two different ways. The first is by interacting with FAS and FADD upon FAS activation blocking death-inducing signaling complex (DISC) assembly. The second way is by interacting with CASP8 in a mitochondria localization-and phosphorylation-dependent manner, which in turn limits the amount of soluble CASP8 avaliable for DISC-mediated activation. ARC also inhibits intrinsic apoptotic pathways in response to a wide range of stresses through interactions with BAX. This results in BAX inactivation and prevents mitochondrial dysfunction and release of pro-apoptotic factors. Mutations affecting the gene can results in myoclonus, familiam cortical.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: apoptosis repressor with CARD; ARC; muscle specific protein; muscle-enriched cytoplasmic protein; Nop 30; nucleolar protein 3 (apoptosis repressor with CARD domain); nucleolar protein of 30 kDa; unnamed protein product

          View more View less

          Gene Aliases: ARC; B430311C09Rik; FCM; MYC; MYOCL1; MYP; NOP; NOP30

          View more View less

          UniProt ID: (Human) O60936, (Mouse) Q9D1X0, (Rat) Q62881

          View more View less

          Entrez Gene ID: (Human) 8996, (Mouse) 78688, (Rat) 85383

          View more View less

          Function(s)
          RNA binding receptor binding death receptor binding calcium ion binding protein binding death effector domain binding identical protein binding cysteine-type endopeptidase inhibitor activity involved in apoptotic process caspase binding cysteine-type endopeptidase inhibitor activity kinase binding phosphatase binding
          Process(es)
          response to hypoxia blood vessel remodeling response to ischemia mRNA splice site selection negative regulation of signal transduction regulation of gene expression cardiac muscle cell apoptotic process negative regulation of striated muscle cell apoptotic process negative regulation of cardiac muscle cell apoptotic process negative regulation of tumor necrosis factor-mediated signaling pathway regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum negative regulation of muscle atrophy release of sequestered calcium ion into cytosol by sarcoplasmic reticulum response to injury involved in regulation of muscle adaptation regulation of apoptotic process negative regulation of apoptotic process myoblast differentiation smooth muscle cell proliferation protein oligomerization negative regulation of cytosolic calcium ion concentration negative regulation of mitochondrial calcium ion concentration negative regulation of programmed necrotic cell death cellular response to hypoxia negative regulation of release of cytochrome c from mitochondria intrinsic apoptotic signaling pathway regulation of NIK/NF-kappaB signaling negative regulation of extrinsic apoptotic signaling pathway via death domain receptors negative regulation of mitochondrial membrane permeability involved in apoptotic process negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway regulation of intracellular signal transduction negative regulation of death-inducing signaling complex assembly negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway negative regulation of establishment of protein localization to mitochondrion negative regulation of extrinsic apoptotic signaling pathway negative regulation of intrinsic apoptotic signaling pathway cellular response to hydrogen peroxide RNA splicing
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-8lbpj:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0