Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cripto Antibodies

          Invitrogen

          Cripto Polyclonal Antibody, Biotin

          View all (20) Cripto antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Cripto Polyclonal Antibody, Biotin

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Cripto Polyclonal Antibody, Biotin

          Product Details

          PA5-22698

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1:1,000
          -

          Flow Cytometry (Flow)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide made to an N-terminal portion of mouse Cripto1 (between residues 1-50).
          3D Epitope / Immunogen

          Conjugate

          Biotin Biotin Biotin

          Form

          Liquid

          Concentration

          0.85 mg/mL

          Amount

          85 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11151971

          Product Specific Information

          In Western blot a band is seen at ~18 kDa representing the core form of Cripto1, and at ~36 kDa reporesenting the glycosylated/ post-translationally modified form.

          Suggested positive control: MDA-MB231 (human breast cancer) lysates.

          Target Information

          Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cripto; cripto growth factor; Cripto-1; cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; epidermal growth factor-like cripto protein CR1; TDGF2; teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR); unnamed protein product

          View more View less

          Gene Aliases: CR; CR-1; CR1; CRGF; TDGF1

          View more View less

          UniProt ID: (Human) P13385, (Mouse) P51865

          View more View less

          Entrez Gene ID: (Human) 6997, (Mouse) 21667

          View more View less

          Function(s)
          receptor binding protein binding growth factor activity nodal binding activin receptor binding coreceptor activity Wnt-protein binding type I activin receptor binding
          Process(es)
          blood vessel development morphogenesis of a branching structure cell migration involved in sprouting angiogenesis signal transduction epidermal growth factor receptor signaling pathway determination of left/right symmetry heart development positive regulation of cell proliferation anterior/posterior axis specification, embryo embryo development ending in birth or egg hatching anterior/posterior pattern specification regulation of signal transduction positive regulation of endothelial cell migration cell differentiation positive regulation of cell migration mammary gland development cellular response to hepatocyte growth factor stimulus nodal signaling pathway negative regulation of apoptotic process positive regulation of MAP kinase activity positive regulation of MAPK cascade cellular response to fibroblast growth factor stimulus cellular response to interferon-gamma cellular response to interleukin-6 cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus vasculogenesis in utero embryonic development positive regulation of cell-matrix adhesion gastrulation anterior/posterior axis specification regulation of cell migration negative regulation of transforming growth factor beta receptor signaling pathway positive regulation of transcription from RNA polymerase II promoter positive regulation of fibroblast proliferation cardiac muscle cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline