Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DISC1 Antibodies

          Invitrogen

          DISC1 Polyclonal Antibody, DyLight™ 550

          View all (34) DISC1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite DISC1 Polyclonal Antibody, DyLight™ 550

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          DISC1 Polyclonal Antibody, DyLight™ 550

          Product Details

          PA5-22840

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          2.5-5 µg/mL
          -

          Immunohistochemistry (Frozen) (IHC (F))

          2.5-5.0 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide made to an internal region (within residues 400-500) of the rat DISC1 protein.
          3D Epitope / Immunogen

          Conjugate

          DyLight™ 550 DyLight™ 550 DyLight™ 550

          Excitation/Emission Max

          556/570 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.85 mg/mL

          Amount

          85 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          50mM sodium borate

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11153752

          Product Specific Information

          The theoretical molecular weight of DISC1 is ~90 kDa.

          Due to alternative splicing, DISC1 may also run at ~75 and 100 kDa.

          Suggested positive control: human brain sections (neurons and glia).

          Target Information

          DISC1 (Disrupted in Schizophrenia 1) is a multifunctional protein that interacts with multiple proteins of the centrosome and cytoskeletal system, and is involved in neuronal functions that are dependent upon proper cytoskeletal regulation, including neuronal migration, neurite architecture and intracellular transport. Regions of the brain which express DISC1 include the hippocampus, lateral septum, amygdala, cerebral cortex, cerebellum and paraventricular hypothalamus, and loss of DISC1 function results in differences in structural organization.The DISC1 gene locus is associated with patients afflicted with schizophrenia as a result of chromosomal translocations. DISC-1 encodes a large protein predicted to contain a globular N-terminal domain and a helical C-terminal domain, both of which have the potential to form interactions with other proteins. DISC-1 interacts with proteins involved in the centrosome and cytoskeletal system, including MIPT3, MAP1A and NUDEL; proteins which localize receptors to membranes, including alpha-actinin 2, beta 4-spectrin; and proteins which transduce signals from membrane receptors, including ATF-4 and ATF-5. Therefore, DISC-1 is thought to be involved in intracellular transport, neurite architecture and/or neuronal migration, all of which are thought to be pathogenic in the schizophrenic brain. Decreased expression of DISC1 in neurons caused an accelerated rate of neuronal integration, resulting in aberrant morphological development, suggesting that DISC1 plays a role in dendritic development and synapse formation. DISC-1 is thought to be involved in intracellular transport, neurite architecture and/or neuronal migration, all of which are thought to be pathogenic in the schizophrenic brain.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Disrupted in schizophrenia 1 homolog; Disrupted in schizophrenia 1 protein; RP4-730B13.1

          View more View less

          Gene Aliases: C1orf136; DISC1; KIAA0457; SCZD9

          View more View less

          UniProt ID: (Human) Q9NRI5, (Mouse) Q811T9, (Rat) Q810H6

          View more View less

          Entrez Gene ID: (Human) 27185, (Mouse) 244667, (Rat) 307940

          View more View less

          Function(s)
          protein binding kinesin binding identical protein binding macromolecular complex binding binding, bridging protein complex binding cytoskeletal protein binding
          Process(es)
          microtubule cytoskeleton organization neuron migration positive regulation of cell-matrix adhesion positive regulation of neuroblast proliferation multicellular organism development nervous system development protein localization regulation of neuron projection development positive regulation of neuron projection development Wnt signaling pathway cerebral cortex radially oriented cell migration cell proliferation in forebrain pyramidal neuron migration positive regulation of Wnt signaling pathway TOR signaling cellular protein localization nonmotile primary cilium assembly cilium assembly positive regulation of axon extension mitochondrial calcium ion homeostasis canonical Wnt signaling pathway regulation of dendritic spine development regulation of synapse maturation positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process response to electrical stimulus ubiquitin-dependent protein catabolic process detection of temperature stimulus involved in sensory perception of pain regulation of synaptic transmission, glutamatergic cilium morphogenesis neuron cellular homeostasis protein localization to centrosome regulation of postsynapse organization non-motile cilium assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline