Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ABCA1 Antibodies

          Invitrogen

          ABCA1 Polyclonal Antibody, HRP

          View all (20) ABCA1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite ABCA1 Polyclonal Antibody, HRP

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          ABCA1 Polyclonal Antibody, HRP

          Product Details

          PA5-22909

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-Dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          ChIP assay (ChIP)

          Assay-dependent
          -

          Neutralization (Neu)

          Assay-dependent
          -

          Gel Shift (EMSA)

          Assay-dependent
          -

          In Situ Hybridization (ISH)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Dog, Chicken, Horse, Hamster, Human, Mustelid, Mouse, Non-human primate, Pig, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Partial peptide sequence (within residues 1100-1300) of human ABCA1.
          3D Epitope / Immunogen

          Conjugate

          HRP HRP HRP

          Form

          Liquid

          Concentration

          0.66 mg/mL

          Amount

          66 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11151837

          Target Information

          Mutations in the ABCA1 gene (ATP-binding cassette transporter 1) have been reported in Tangier disease (TD). TD is an autosomal recessive disorder characterized by an absence of plasma HDL, cholesterol ester depositing in the reticulo-endothelial system and deviations in cellular lipid trafficking. ABCA1 mediates the apo-A1 associated export of cholesterol and phospholipids from the cell. It is expressed on the plasma membrane and the Golgi complex, and is regulated by cholesterol flux. Regulation of the cholesterol flux between HDL and macrophages is competitive between ABCA1 and SR-BI.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ABC 1; ABC 1 antibody; ABC transporter; ABC Transporter 1; ABC-1; ABC1 antibody; ATP-binding cassette 1; ATP-binding cassette sub-family A member 1; ATP-binding cassette transporter 1; ATP-binding cassette transporter A1; ATP-binding cassette, sub-family A (ABC1), member 1; CERP antibody; Cholesterol efflux regulatory protein; membrane-bound; Phospholipid-transporting ATPase ABCA1

          View more View less

          Gene Aliases: ABC-1; ABC1; ABCA1; CERP; HDLCQTL13; HDLDT1; HPALP1; TGD

          View more View less

          UniProt ID: (Human) O95477, (Mouse) P41233

          View more View less

          Entrez Gene ID: (Chicken) 373945, (Horse) 100054241, (Dog) 481651, (Rabbit) 100356765, (Human) 19, (Pig) 100152112, (Mouse) 11303, (Rat) 313210, (Chinese hamster) 100761226

          View more View less

          Function(s)
          nucleotide binding receptor binding protein binding ATP binding phospholipid transporter activity high-density lipoprotein particle binding protein transmembrane transporter activity cholesterol binding ATPase activity syntaxin binding phosphatidylcholine binding small GTPase binding apolipoprotein binding apolipoprotein A-I binding apolipoprotein A-I receptor activity ATPase activity, coupled to transmembrane movement of substances sphingolipid floppase activity ATPase binding phosphatidylcholine-translocating ATPase activity phosphatidylserine-translocating ATPase activity intermembrane cholesterol transfer activity ATPase-coupled intramembrane lipid transporter activity floppase activity ABC-type transporter activity transporter activity anion transmembrane transporter activity cholesterol transporter activity ATP-binding cassette (ABC) transporter primary active transporter transporter
          Process(es)
          peptide secretion lipid metabolic process phagocytosis, engulfment lysosome organization G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway response to nutrient steroid metabolic process cholesterol metabolic process protein secretion response to xenobiotic stimulus negative regulation of macrophage derived foam cell differentiation positive regulation of cholesterol efflux negative regulation of cholesterol storage organic hydroxy compound transport phospholipid transport endosomal transport signal release cholesterol transport response to nutrient levels intracellular cholesterol transport regulation of Cdc42 protein signal transduction cholesterol efflux response to vitamin B3 phospholipid efflux high-density lipoprotein particle assembly response to laminar fluid shear stress apolipoprotein A-I-mediated signaling pathway lipoprotein metabolic process lipoprotein biosynthetic process cholesterol homeostasis reverse cholesterol transport phospholipid translocation establishment of localization in cell transmembrane transport phospholipid homeostasis platelet dense granule organization cellular response to lipopolysaccharide cellular response to retinoic acid cellular response to cytokine stimulus cellular response to cholesterol cellular response to low-density lipoprotein particle stimulus cellular response to xenobiotic stimulus protein transmembrane transport regulation of high-density lipoprotein particle assembly positive regulation of high-density lipoprotein particle assembly sphingolipid translocation export across plasma membrane protein lipidation transport positive regulation of cAMP biosynthetic process interleukin-1 beta secretion response to high density lipoprotein particle anion transmembrane transport ion transmembrane transport response to drug response to low-density lipoprotein particle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-2x6nv:80/100.66.78.26:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline