Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CCR2 (CD192) Antibodies

          Invitrogen

          CCR2 Polyclonal Antibody, HRP

          View all (34) CCR2 (CD192) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CCR2 Polyclonal Antibody, HRP

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          CCR2 Polyclonal Antibody, HRP

          Product Details

          PA5-23041

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-Dependent
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Cat, Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide made to an N-terminal portion of the human CCR2 protein (within residues 20-100).
          3D Epitope / Immunogen

          Conjugate

          HRP HRP HRP

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11153550

          Product Specific Information

          The immunogen sequence similarity with other species: Rhesus monkey and several other primate species (100%), Mouse (69%).

          Target Information

          CCR2, a chemokine receptor, is a regulator of monocyte/macrophage traffickings and is upregulated in response to inflammation. Resistance to HIV-1 infection or delayed progression to AIDS may be linked to CCR2 polymorphisms. The ligand for CCR2 is monocyte chemoattractant protein (MCP-1), a chemokine produced by endothelial cells, smooth muscle cells, and macrophages. Activation of monocytes by MCP-1 may mediate the monocytic infiltration of tissues in atherosclerosis and other inflammatory disorders including rheumatoid arthritis, alveolitis, and tumor infiltration. Two isoforms are produced by alternative splicing. CCR2 has been reported to be expressed in a wide variety of tissues, including blood, brain, heart, kidney, liver, lung, ovary, pancreas, spinal cord, spleen, thymus, and vessel. Increased expression of CCR2 has been documented in CD3+ lymphocytes in rheumatoid arthritis. ESTs have been isolated from liver/spleen and prostate libraries.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: C-C chemokine receptor type 2; C-C CKR-2; CC-CKR-2; Cc-ckr2; CCR-2; CD192 antigen; chemokine (C-C motif) receptor 2; chemokine receptor CCR2; hCG_14621; MCP-1 receptor; Mcp1 receptor; Mcp1r; monocyte chemoattractant protein 1 receptor; monocyte chemotactic protein 1 receptor; unnamed protein product

          View more View less

          Gene Aliases: CC-CKR-2; CCR-2; CCR2A; CCR2B; CD192; CKR2; CKR2A; CKR2B; CMKBR2; MCP-1-R; PCLUD

          View more View less

          UniProt ID: (Human) P41597, (Rat) O55193

          View more View less

          Entrez Gene ID: (Bovine) 539002, (Human) 729230, (Cat) 100049057, (Rat) 60463

          View more View less

          Function(s)
          G-protein coupled receptor activity chemokine receptor activity protein binding C-C chemokine receptor activity cytokine binding C-C chemokine binding CCR2 chemokine receptor binding chemokine (C-C motif) ligand 2 binding chemokine (C-C motif) ligand 12 binding chemokine (C-C motif) ligand 7 binding identical protein binding chemokine (C-C motif) ligand 5 binding
          Process(es)
          blood vessel remodeling dendritic cell chemotaxis regulation of T cell cytokine production positive regulation of T-helper 1 type immune response negative regulation of type 2 immune response cellular calcium ion homeostasis chemotaxis inflammatory response immune response cellular defense response G-protein coupled receptor signaling pathway negative regulation of adenylate cyclase activity positive regulation of cytosolic calcium ion concentration JAK-STAT cascade response to wounding regulation of vascular endothelial growth factor production positive regulation of T cell chemotaxis negative regulation of angiogenesis cytokine-mediated signaling pathway sensory perception of pain calcium-mediated signaling cellular homeostasis positive regulation of interferon-gamma production positive regulation of interleukin-2 production positive regulation of tumor necrosis factor production monocyte extravasation T-helper 17 cell chemotaxis negative regulation of eosinophil degranulation regulation of T cell differentiation positive regulation of alpha-beta T cell proliferation regulation of inflammatory response positive regulation of inflammatory response positive regulation of T cell activation positive regulation of synaptic transmission, glutamatergic cell chemotaxis chemokine-mediated signaling pathway cellular response to cytokine stimulus positive regulation of monocyte chemotaxis positive regulation of immune complex clearance by monocytes and macrophages inflammatory response to wounding positive regulation of cold-induced thermogenesis positive regulation of glutamate receptor signaling pathway macrophage migration positive regulation of thymocyte migration positive regulation of monocyte extravasation positive regulation of CD8-positive, alpha-beta T cell extravasation positive regulation of astrocyte chemotaxis positive regulation of hematopoietic stem cell migration response to hypoxia inflammatory response to antigenic stimulus monocyte chemotaxis humoral immune response hemopoiesis regulation of cell migration homeostasis of number of cells within a tissue response to hyperoxia leukocyte adhesion to vascular endothelial cell neutrophil clearance positive regulation of p38MAPK cascade positive regulation of leukocyte tethering or rolling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-6ndwg:80/100.66.72.244:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0