Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CRHR2 Antibodies

          Invitrogen

          CRHR2 Polyclonal Antibody

          2 References
          View all (15) CRHR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CRHR2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CRHR2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CRHR2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CRHR2 Antibody (PA5-23129) in WB

          Western blot analysis of CRHR2 in 0.5 mg/mL Human Brain lysate. Samples were incubated in CRHR2 polyclonal antibody (Product # PA5-23129). This experiment was performed under reducing conditions using the 12-230 kDa separation system. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CRHR2 Antibody in Western Blot (WB)
          CRHR2 Antibody in Western Blot (WB)
          CRHR2 Polyclonal Antibody

          Product Details

          PA5-23129

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.5-2 µg/mL
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Bovine, Dog, Chicken, Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A portion of amino acids 75-125 of human CRHR2.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.0 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2540656

          Product Specific Information

          Suggested positive control: human brain lysate, human brain protein.

          94% sequence identity with Rhesus monkey.

          Target Information

          Corticotropin-releasing hormone (CRH; 122560) is a 41-amino acid peptide synthesized in the hypothalamus, and play a role in the neuroregulation, responses to stress and immune challenge. Corticotropin Releasing Factor Receptor 2 (CRHR2) was the receptor of CRH. The sepression of CRHR2 has been reported in various regions of the brain as well as in placenta, umbilical vein, heart, epididymis, gastrointestinal tract, adrenal and skeletal muscle.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: corticotrophin releasing hormone receptor 2; corticotropin releasing factor receptor type 2 beta; corticotropin-releasing factor 2; Corticotropin-releasing factor receptor 2; corticotropin-releasing factor receptor subtype 2; Corticotropin-releasing hormone receptor 2; corticotropin-releasing hormone receptor type 2 variant 2; CRF 2 receptor; CRF-R 2; CRF-R-2; CRF-R2; CRF-RB; CRF2; CRF2a-desQ; CRF2d; CRF2e; CRF2f; CRF2g; CRFR-2; CRH-R 2; CRH-R-2; CRH-R2; Crh2 receptor; G protein-coupled receptor; G-protein linked receptor; Heart and muscle corticotropin-releasing factor receptor; HM-CRF; putative corticotropin-releasing hormone receptor type 2; unnamed protein product

          View more View less

          Gene Aliases: CRF-R2; CRF-RB; CRF2; CRF2R; CRFR2; CRFR2alpha; CRFR2beta; CRH-R2; CRH2R; CRHR2; HM-CRF

          View more View less

          UniProt ID: (Human) Q13324, (Rat) P47866, (Mouse) Q60748

          View more View less

          Entrez Gene ID: (Human) 1395, (Bovine) 521831, (Chicken) 395105, (Dog) 482384, (Rat) 64680, (Mouse) 12922

          View more View less

          Function(s)
          transmembrane signaling receptor activity G-protein coupled receptor activity protein binding G-protein coupled peptide receptor activity corticotrophin-releasing factor receptor activity peptide hormone binding corticotropin-releasing hormone receptor activity hormone activity signal transducer activity transmembrane signal receptor
          Process(es)
          signal transduction cell surface receptor signaling pathway G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway hormone-mediated signaling pathway long-term synaptic potentiation cellular response to corticotropin-releasing hormone stimulus actin filament organization protein kinase C-activating G-protein coupled receptor signaling pathway feeding behavior positive regulation of heart rate positive regulation of gene expression negative regulation of gene expression negative regulation of norepinephrine secretion positive regulation of serotonin secretion negative regulation of angiogenesis sensory perception of pain cerebellum development hypothalamus development positive regulation of cAMP metabolic process negative regulation of cAMP biosynthetic process positive regulation of cAMP biosynthetic process epithelial cell differentiation positive regulation of interleukin-6 production negative regulation of epinephrine secretion positive regulation of stress-activated MAPK cascade negative regulation of luteinizing hormone secretion gastric motility catecholamine biosynthetic process positive regulation of cAMP-mediated signaling negative regulation of cAMP-mediated signaling positive regulation of blood pressure negative regulation of follicle-stimulating hormone secretion skeletal muscle tissue growth positive regulation of NF-kappaB transcription factor activity negative regulation of insulin secretion involved in cellular response to glucose stimulus regulation of ERK1 and ERK2 cascade cellular response to glucocorticoid stimulus negative regulation of calcium ion import negative regulation of feeding behavior negative regulation of defecation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline