Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 33
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, PC-12, A549, SiHa.
Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (92%), Xenopus laevis (89%), Pig (100%), Chicken (96%), Rhesus Monkey (100%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PCNA (polymerase delta auxiliary protein) is essential for DNA replication and is involved in DNA excision and mismatch repair pathways. PCNA binds to the CDK inhibitor p21, the structure-specific endonucleases Fen1 and XPG, and DNA cytosine 5-methyltransferase (MCMT). PCNA is a potentially useful marker of cells with proliferative potential and for identifying the proliferation status of tumor tissue (i.e. relevant to prognosis). PCNA is a marker for cells in early G1 phase and S phase of the cell cycle. PCNA is found in the nucleus and is a cofactor of DNA polymerase delta, and acts as a homotrimer and helps increase the processing of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Two transcript variants encoding the same protein have been found for PCNA. Pseudogenes of PCNA have been described on chromosome 4 and on the X chromosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CG9193-PA; CG9193-PB; cyclin; DNA polymerase delta auxiliary protein; HGCN8729; LD45889p; MGC8367; mutagen-sensitive 209; PCNA; PCNA-PA; PCNA-PB; proliferating-cell nuclear antigen; Proliferating-cell-nuclear-antigen; unnamed protein product
Gene Aliases: 53/13; ATLD2; CG9193; Dmel\CG9193; Dmel_CG9193; DmPCNA; DmPCNA1; dPCNA; l(2)02448; l(2)56F; l(2)56Fa; l(2)s1534; mus 209; MUS209; Pcna; Pcna/cyclin; PCNA/mus209; PCNA1; PCNAR
UniProt ID: (Fruit fly) P17917, (Human) P12004, (Chinese hamster) P57761, (Mouse) P17918, (Rat) P04961
Entrez Gene ID: (Fruit fly) 37290, (Human) 5111, (Chinese hamster) 100689335, (Mouse) 18538, (Rat) 25737
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support