Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ataxin 3 Antibodies

          Invitrogen

          Ataxin 3 Polyclonal Antibody

          Advanced Verification
          View all (19) Ataxin 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Ataxin 3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          • Advanced Verification (2)
          Ataxin 3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Ataxin 3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ataxin 3 Antibody (PA5-27391) in ICC/IF

          Immunocytochemistry-Immunofluorescence analysis of Ataxin 3 was performed in SK N SH cells fixed in 4% paraformaldehyde at RT for 15 min. Green: Ataxin 3 Polyclonal Antibody (Product # PA5 27391) diluted at 1:250. Blue: Hoechst 33342 staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ataxin 3 Antibody in Immunocytochemistry (ICC/IF)
          Ataxin 3 Antibody in Immunocytochemistry (ICC/IF)
          Ataxin 3 Antibody in Western Blot (WB)
          Ataxin 3 Antibody in Western Blot (WB)
          Ataxin 3 Antibody in Western Blot (WB)
          Ataxin 3 Antibody in Western Blot (WB)
          Ataxin 3 Antibody in Western Blot (WB)
          Ataxin 3 Antibody
          Ataxin 3 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-27391

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:20,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human Ataxin 3. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.55 mg/mL

          Amount

          55 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 10% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2544867

          Product Specific Information

          Recommended positive controls: 293T, A431, HeLa, HepG2, SK-N-SH, SK-N-AS, human Ataxin3-transfected 293T.

          Predicted reactivity: Mouse (88%), Rat (82%), Pig (89%), Sheep (92%), Bovine (93%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Machado-Joseph disease is an autosomal dominant neurologic disorder, and is now known to be the same as previously described spinocerebellar ataxia-3. MJD protein (Ataxin-3) contains (CAG)n repeats in the coding region, and the expansion of these repeats from the normal 13-36 to 68-79 is the cause of Machado-Joseph disease. There is a negative correlation between the age of onset and CAG repeat numbers. This protein interacts with key regulators (CBP, p300 and PCAF) of transcription and represses transcription, and also acts as a histone-binding protein that regulates transcription. MJD is a deubiquitinating enzyme.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ataxin3; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1; machado-Joseph disease protein 1 homolog; olivopontocerebellar ataxia 3; spin; spinocerebellar ataxia type 3 protein; unnamed protein product

          View more View less

          Gene Aliases: 2210008M02Rik; AT3; ataxin-3; ATX3; JOS; MJD; MJD1; SCA3

          View more View less

          UniProt ID: (Human) P54252, (Mouse) Q9CVD2

          View more View less

          Entrez Gene ID: (Human) 4287, (Mouse) 110616

          View more View less

          Function(s)
          thiol-dependent ubiquitin-specific protease activity protein binding ubiquitin protein ligase binding identical protein binding ATPase binding Lys63-specific deubiquitinase activity Lys48-specific deubiquitinase activity RNA polymerase II regulatory region sequence-specific DNA binding transcription corepressor binding histone deacetylase activity histone deacetylase binding
          Process(es)
          microtubule cytoskeleton organization nucleotide-excision repair ubiquitin-dependent protein catabolic process misfolded or incompletely synthesized protein catabolic process chemical synaptic transmission nervous system development regulation of cell-substrate adhesion protein deubiquitination actin cytoskeleton organization cellular response to amino acid starvation cellular response to heat monoubiquitinated protein deubiquitination proteasome-mediated ubiquitin-dependent protein catabolic process intermediate filament cytoskeleton organization protein K63-linked deubiquitination protein K48-linked deubiquitination cellular response to misfolded protein negative regulation of TORC1 signaling positive regulation of ERAD pathway protein localization to cytosolic proteasome complex positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process cytoplasmic translation chromatin remodeling cellular response to starvation cellular response to nutrient levels exploration behavior TORC1 signaling negative regulation of translational initiation positive regulation of translational initiation positive regulation of TORC1 signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline