Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Lyn Antibodies

          Invitrogen

          Lyn Polyclonal Antibody

          View all (42) Lyn antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Lyn Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          Lyn Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Lyn Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Lyn Antibody (PA5-29329) in ICC/IF

          Immunocytochemistry analysis of Lyn in methanol-fixed A431. Samples were incubated with Lyn polyclonal antibody (Product # PA5-29329) using a dilution of 1:500. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Lyn Antibody in Immunocytochemistry (ICC/IF)
          Lyn Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Lyn Antibody in Western Blot (WB)
          Lyn Antibody in Western Blot (WB)
          Lyn Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-29329

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human LYN. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.86 mg/mL

          Amount

          86 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.1M tris glycine, pH 7, with 10% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2546805

          Product Specific Information

          Recommended positive controls: A431, mouse brain, K562.

          Predicted reactivity: Mouse (98%), Rat (97%), Zebrafish (85%), Xenopus laevis (90%), Chicken (92%), Sheep (99%), Bovine (99%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          Lyn proto oncogene (Lcl/Yes related novel protein tyrosine kinase) is encoded by the Lyn gene located on chromosome 8 in humans and belongs to the Src family kinase. It is a non-receptor tyrosine kinase that acts as a mediator in transducing cellular signaling. The most well studied expression of Lyn is in hematopoietic cell types, both lymphoid and myeloid cells with exclusion in T lymphocytes. It has also been observed that Lyn plays diverse roles such as in regulation of B cell receptor signaling, mast cell signaling and Epithelial - Mesenchymal Transition (EMT).

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ26625; lck/Yes-related novel protein tyrosine kinase; lyn; lyn tyrosine kinase; protein-tyrosine kinase (EC 2.7.1.112); unnamed protein product; V-yes-1 Yamaguchi sarcoma viral related oncogene homolog; Yamaguchi sarcoma viral (v-yes-1) oncogene homolog

          View more View less

          Gene Aliases: Hck-2; JTK8; p53Lyn; p56Lyn; SAIDV

          View more View less

          UniProt ID: (Human) P07948, (Mouse) P25911

          View more View less

          Entrez Gene ID: (Human) 4067, (Mouse) 17096

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding platelet-derived growth factor receptor binding integrin binding protein binding ATP binding kinase activity enzyme binding receptor activator activity ubiquitin protein ligase binding immunoglobulin receptor binding gamma-tubulin binding glycosphingolipid binding ion channel binding macromolecular complex binding ephrin receptor binding phosphoprotein binding scaffold protein binding phosphorylation-dependent protein binding erythropoietin receptor binding SH3 domain binding phosphatidylinositol 3-kinase activator activity
          Process(es)
          DNA damage checkpoint cell morphogenesis B cell homeostasis regulation of cytokine production negative regulation of protein phosphorylation hematopoietic progenitor cell differentiation Fc receptor mediated stimulatory signaling pathway tolerance induction to self antigen histamine secretion by mast cell platelet degranulation negative regulation of myeloid leukocyte differentiation immune response-regulating cell surface receptor signaling pathway Fc receptor mediated inhibitory signaling pathway regulation of B cell apoptotic process protein phosphorylation autophagy defense response inflammatory response cellular response to DNA damage stimulus transmembrane receptor protein tyrosine kinase signaling pathway positive regulation of cell proliferation negative regulation of cell proliferation response to hormone positive regulation of neuron projection development fatty acid transport vesicle-mediated transport peptidyl-tyrosine phosphorylation hemopoiesis erythrocyte differentiation eosinophil differentiation positive regulation of cell migration negative regulation of B cell proliferation neuron projection development lipopolysaccharide-mediated signaling pathway regulation of mast cell activation regulation of cell adhesion mediated by integrin negative regulation of toll-like receptor 2 signaling pathway toll-like receptor 4 signaling pathway negative regulation of toll-like receptor 4 signaling pathway positive regulation of toll-like receptor 4 signaling pathway positive regulation of toll-like receptor 9 signaling pathway intracellular signal transduction interleukin-5-mediated signaling pathway C-X-C chemokine receptor CXCR4 signaling pathway B cell proliferation positive regulation of phosphorylation hemoglobin biosynthetic process regulation of mast cell degranulation negative regulation of MAP kinase activity positive regulation of MAPK cascade regulation of erythrocyte differentiation protein autophosphorylation ephrin receptor signaling pathway regulation of inflammatory response B cell receptor signaling pathway regulation of B cell receptor signaling pathway negative regulation of B cell receptor signaling pathway regulation of release of sequestered calcium ion into cytosol positive regulation of glial cell proliferation positive regulation of Fc receptor mediated stimulatory signaling pathway regulation of ERK1 and ERK2 cascade negative regulation of ERK1 and ERK2 cascade positive regulation of oligodendrocyte progenitor proliferation negative regulation of mast cell proliferation positive regulation of mast cell proliferation cellular response to retinoic acid regulation of monocyte chemotaxis regulation of platelet aggregation dendritic cell differentiation neuroinflammatory response negative regulation of intracellular signal transduction positive regulation of amyloid precursor protein catabolic process positive regulation of protein localization to plasma membrane positive regulation of dendritic cell apoptotic process regulation of protein phosphorylation stimulatory C-type lectin receptor signaling pathway innate immune response-activating signal transduction negative regulation of inflammatory response to antigenic stimulus response to sterol depletion signal transduction response to xenobiotic stimulus response to toxic substance response to carbohydrate oligodendrocyte development T cell costimulation response to insulin cellular response to heat Fc-epsilon receptor signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis response to amino acid response to peptide hormone response to axon injury negative regulation of immune response leukocyte migration JAK-STAT cascade involved in growth hormone signaling pathway cellular response to lipid
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0