Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Atpalpha Antibodies

          Invitrogen

          Atpalpha Polyclonal Antibody

          View all (2) Atpalpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Atpalpha Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Atpalpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Atpalpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Atpalpha Antibody (PA5-32093) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human colon carcinoma, using Atpalpha (Product # PA5-32093) antibody at 1:250 dilution. Antigen Retrieval: EDTA based buffer, pH 8.0, 15 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Atpalpha Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Atpalpha Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-32093

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:20,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Fruit fly, Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of Drosophila Atpalpha. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 1% BSA, 20% glycerol

          Contains

          0.01% thimerosal

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2549566

          Product Specific Information

          Recommended positive controls: Drosophila lysate.

          Predicted reactivity: Drosophila (100%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          The ATPa, or ATPalpha gene, encodes a subunit of the Na+/K+ ATPase enzyme, which is crucial for maintaining cellular ion gradients by actively transporting Na+ out of cells and K+ into cells. This gene is essential in various organisms, including Drosophila melanogaster (fruit fly). Mutations in the ATPalpha gene can lead to significant developmental and physiological abnormalities, such as neurodegeneration, altered longevity, and myodegeneration. One of its critical roles is in neuromuscular functioning, where it affects locomotor activity, with mutations causing conditional bang-sensitive or temperature-sensitive paralysis. Additionally, ATPalpha mutations disrupt the formation and function of septate junctions in Drosophila, structures analogous to vertebrate tight junctions, further underscoring its role in maintaining cellular integrity and signal transduction.

          ⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: (Na[+] + K[+])-ATPase alpha-subunit; (Na[+],K[+])ATPase alpha subunit; alpha-subunit of ATPase; ATPalpha; Atpalpha-PA; Atpalpha-PB; Atpalpha-PC; Atpalpha-PD; Atpalpha-PE; Atpalpha-PF; Atpalpha-PG; Atpalpha-PH; Atpalpha-PI; Atpalpha-PJ; Atpalpha-PK; ATPase alpha; ATPase alpha subunit; CG5670-PA; CG5670-PB; CG5670-PC; CG5670-PD; CG5670-PE; CG5670-PF; CG5670-PG; CG5670-PH; CG5670-PI; CG5670-PJ; CG5670-PK; Na+,K+ ATPase alpha-subunit; Na, K ATPase a subunit; Na,K ATPase; Na,K ATPase alpha subunit; Na,K-ATPase; Na,K-ATPase alpha; Na,K-ATPase alpha subunit; Na,K-ATPase alpha-subunit; Na[ +], K[ +] -ATPase; Na[+] pump alpha subunit; Na[+], K[+] ATPase alpha; Na[+],K[+] ATPase alpha subunit; Na[+],K[+]-ATPase alpha subunit; Na[+]-K[+] ATPase; Na[+]/K[+] ATPase (alpha subunit); Na[+]/K[+] ATPase alpha subunit; Na[+]K[+] ATPase alpha subunit; Na[+]K[+]ATPase; NaK-ATPase; sodium pump alpha subunit; sodium pump alpha-subunit

          View more View less

          Gene Aliases: (Na[+]/k[+])-ATPase; alpha-ATPase; Atp; Atp a; ATP-alpha; ATPa; ATPalpha; ATPBagr; CG5670; Da-47; Dmel\CG5670; Dmel_CG5670; dnATPase; JY; K ATPase; K-ATPase; K[+] -ATPase; K[+]-ATPase; K[+]-ATPase alpha subunit; K[+]-ATPase alpha-subunit; l(3)01164; l(3)01453; l(3)04492; l(3)j3B2; l(3)j3B7; l(3)j6A4; l(3)rM454; l(3)ry136; Na; Na+-K+-ATPase; Na+/K+ ATPase alpha subunit; Na+/K+-ATPase; Na-p; Na/K ATPase; Na/K-ATPase; Na/K-ATPase alpha-subunit; Na[+]; Na[+]-K[+]-ATPase alpha[[5]]-subunit; Na[+]/K[+] ATPase; Na[+]/K[+]-ATPase; Na[+]/K[+]ATPase; Na[+]K[+] ATPase; Na[+]K[+]-ATPase; Na[+]K[+]ATPase; NaK; NaK ATPase; NaK-ATPase; NKA; sodium/potassium transporting ATPase alpha

          View more View less

          UniProt ID: (Fruit fly) P13607

          View more View less

          Entrez Gene ID: (Fruit fly) 48971

          View more View less

          Function(s)
          sodium:potassium-exchanging ATPase activity ATP binding cation transmembrane transporter activity primary active transporter
          Process(es)
          tissue homeostasis cation transport cellular sodium ion homeostasis chemical synaptic transmission sensory perception of sound locomotory behavior jump response determination of adult lifespan adult locomotory behavior response to temperature stimulus response to mechanical stimulus septate junction assembly cellular potassium ion homeostasis regulation of tube diameter, open tracheal system regulation of tube length, open tracheal system sodium ion export from cell neuromuscular process synaptic growth at neuromuscular junction trachea morphogenesis hydrogen ion transmembrane transport potassium ion import across plasma membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0