Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PP2A alpha Antibodies

          Invitrogen

          Phospho-PP2A alpha (Tyr307) Polyclonal Antibody

          Advanced Verification
          5 Published Figures
          10 References
          View all (34) PP2A alpha antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-PP2A alpha (Tyr307) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (5)
          • Advanced Verification (1)
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PP2A alpha (Tyr307) Antibody (PA5-36874) in WB

          Western blot analysis of Phospho-PP2A alpha (Tyr307) in Lane 1: HEK293T cell treated with EGF (0.1 ng/mL, 30 min), Lane 2: mouse brain tissue lysate, Lane 3: H9C2 cell treated with EGF (0.1 ng/mL, 30 min). Samples were incubated with Phospho-PP2A alpha (Tyr307) polyclonal antibody (Product # PA5-36874) at a dilution of 1:500. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Phospho-PP2A alpha (Tyr307) Antibody in Western Blot (WB)
          Phospho-PP2A alpha (Tyr307) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-PP2A alpha (Tyr307) Polyclonal Antibody

          Product Details

          PA5-36874

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phosphopeptide derived from human PP2A-alpha around the phosphorylation site of Tyrosine 307
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2553794

          Product Specific Information

          This antibody detects endogenous protein at a molecular weight of 38 kDa.

          Purity is >95% by SDS-PAGE.

          Target Information

          In eukaryotes, the phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions, including division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the protein phosphatases. In general, the protein phosphatase (PP) holoenzyme is a trimeric complex composed of a regulatory subunit, a variable subunit and a catalytic subunit. Four major families of protein phosphatase catalytic subunits have been identified, designated PP1, PP2A, PP2B (calcineurin) and PP2C. The PP2A family comprises subfamily members PP2A alpha and PP2A beta. An additional protein phosphatase catalytic subunit, PPX (also known as PP4) is a putative member of a novel PP family.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: B55-GAMMA; IMYPNO; IMYPNO1; Methyl-PPP2CA; MGC33570; phosphatase 2A; pp2A; PP2A alpha; PP2A C terminal; PP2A L9; PP2A Leucine 9; PP2A-alpha; PPP2CA; PR52; PR55G; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2A catalytic alpha subunit; protein phosphatase-2A-alpha; replication protein C; type-2A protein phosphatase catalytic subunit; unnamed protein product

          View more View less

          Gene Aliases: HJS3; NEDLBA; PP2A; Pp2a1; PP2Ac; PP2CA; PP2Calpha; RP-C

          View more View less

          UniProt ID: (Human) P67775, (Mouse) P63330, (Rat) P63331

          View more View less

          Entrez Gene ID: (Human) 5515, (Mouse) 19052, (Rat) 24672

          View more View less

          Function(s)
          phosphoprotein phosphatase activity protein serine/threonine phosphatase activity protein tyrosine phosphatase activity protein binding hydrolase activity protein heterodimerization activity tau protein binding GABA receptor binding RNA polymerase II CTD heptapeptide repeat S2 phosphatase activity RNA polymerase II CTD heptapeptide repeat S5 phosphatase activity RNA polymerase II CTD heptapeptide repeat S7 phosphatase activity enzyme binding protein kinase binding protein phosphatase binding protein domain specific binding beta-2 adrenergic receptor binding identical protein binding protein kinase B binding ion channel binding macromolecular complex binding protein phosphatase 2A binding protein antigen binding
          Process(es)
          mitotic cell cycle regulation of transcription from RNA polymerase II promoter protein dephosphorylation response to lead ion negative regulation of epithelial to mesenchymal transition snRNA processing regulation of microtubule polymerization regulation of transcription elongation from RNA polymerase II promoter regulation of hippo signaling negative regulation of hippo signaling intracellular signal transduction regulation of growth T cell homeostasis regulation of cell differentiation negative regulation of protein kinase B signaling negative regulation of canonical Wnt signaling pathway vascular endothelial cell response to oscillatory fluid shear stress RNA polymerase II transcription initiation surveillance positive regulation of NLRP3 inflammasome complex assembly negative regulation of glycolytic process through fructose-6-phosphate regulation of G1/S transition of mitotic cell cycle regulation of protein phosphorylation transcription from RNA polymerase II promoter transcription elongation from RNA polymerase II promoter mesoderm development regulation of receptor activity regulation of protein autophosphorylation peptidyl-threonine dephosphorylation regulation of protein catabolic process positive regulation of glycolytic process regulation of cell cycle negative regulation of protein phosphorylation cardiac ventricle development heart development cellular response to insulin stimulus positive regulation of dephosphorylation negative regulation of protein import into nucleus cellular response to calcium ion cellular response to glucose stimulus cellular response to cytokine stimulus cellular response to ethanol cellular response to follicle-stimulating hormone stimulus cellular response to steroid hormone stimulus negative regulation of calcium ion transmembrane transporter activity positive regulation of extrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-nct7h:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0