Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • WNT3A Antibodies

          Invitrogen

          WNT3A Polyclonal Antibody

          View all (19) WNT3A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite WNT3A Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          WNT3A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          WNT3A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          WNT3A Antibody (PA5-37320) in WB

          Western blot analysis of WNT3A in NCI-H292 cells. Samples were incubated with WNT3A polyclonal antibody (Product # PA5-37320). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          WNT3A Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          WNT3A Polyclonal Antibody

          Product Details

          PA5-37320

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant full length human Wnt3a
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2554007

          Product Specific Information

          This antibody detects endogenous protein at a molecular weight of 39 and 68 kDa.

          Purity is >95% by SDS-PAGE.

          Target Information

          Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: MGC119418; MGC119419; MGC119420; unnamed protein product; vestigial tail; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family, member 3A; Wnt 3a; WNT3A protein

          View more View less

          Gene Aliases: vt; Wnt-3a

          View more View less

          UniProt ID: (Human) P56704, (Mouse) P27467

          View more View less

          Entrez Gene ID: (Human) 89780, (Mouse) 22416, (Rat) 303181

          View more View less

          Function(s)
          transcription coactivator activity receptor binding frizzled binding cytokine activity protein binding protein domain specific binding co-receptor binding identical protein binding receptor agonist activity
          Process(es)
          positive regulation of receptor internalization heart development COP9 signalosome assembly gene expression regulation of gene expression positive regulation of gene expression negative regulation of neuron projection development Wnt signaling pathway cell proliferation in forebrain neurogenesis neuron differentiation midbrain development cell proliferation in midbrain non-canonical Wnt signaling pathway synaptic vesicle recycling cell fate commitment positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of mesodermal cell fate specification animal organ development positive regulation of skeletal muscle tissue development negative regulation of neurogenesis modulation of synaptic transmission regulation of synapse organization canonical Wnt signaling pathway cardiac muscle cell fate commitment positive regulation of dermatome development secondary palate development cellular response to retinoic acid positive regulation of canonical Wnt signaling pathway calcium ion transmembrane transport via low voltage-gated calcium channel presynapse assembly positive regulation of protein localization to plasma membrane negative regulation of dopaminergic neuron differentiation midbrain dopaminergic neuron differentiation regulation of postsynapse to nucleus signaling pathway regulation of presynapse assembly positive regulation of cardiac muscle cell differentiation osteoblast differentiation in utero embryonic development somitogenesis positive regulation of cytokine production heart looping signal transduction cell-cell signaling determination of left/right symmetry axonogenesis axon guidance mesoderm development protein localization positive regulation of cell proliferation organ morphogenesis anterior/posterior pattern specification spinal cord association neuron differentiation hippocampus development dorsal/ventral neural tube patterning hemopoiesis platelet activation extracellular matrix organization BMP signaling pathway mammary gland development positive regulation of B cell proliferation positive regulation of telomere maintenance via telomerase skeletal muscle cell differentiation post-anal tail morphogenesis inner ear morphogenesis regulation of cell differentiation negative regulation of fat cell differentiation somatic stem cell division paraxial mesodermal cell fate commitment positive regulation of collateral sprouting in absence of injury negative regulation of axon extension involved in axon guidance regulation of axonogenesis positive regulation of protein tyrosine kinase activity regulation of microtubule cytoskeleton organization platelet aggregation dopaminergic neuron differentiation axis elongation involved in somitogenesis positive regulation of cell-cell adhesion mediated by cadherin positive regulation of neural precursor cell proliferation positive regulation of hepatocyte proliferation regulation of RNA biosynthetic process biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0