Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Caveolin 1 Antibodies

          Invitrogen

          Phospho-Caveolin 1 (Tyr14) Polyclonal Antibody

          View all (56) Caveolin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-Caveolin 1 (Tyr14) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Phospho-Caveolin 1 (Tyr14) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-Caveolin 1 (Tyr14) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Caveolin 1 (Tyr14) Antibody (PA5-37506) in ICC/IF

          Immunocytochemical analysis of Phospho-Caveolin 1 (Tyr14) in methanol-fixed Hela cells, using Phospho-Caveolin 1 (Tyr14) Polyclonal Antibody (Product # PA5-37506). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Caveolin 1 (Tyr14) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Caveolin 1 (Tyr14) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-Caveolin 1 (Tyr14) Polyclonal Antibody

          Product Details

          PA5-37506

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide sequence around phosphorylation site of tyrosine 14 (H-L-Y(p)-T-V) derived from Human CAVEOLIN-1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2554115

          Product Specific Information

          A suggested positive control for Western blot is NIH-3T3 cells; suggested positive control for ICC/IF is Hela cells.

          Target Information

          Caveolin-1 or CAV1 is a scaffolding proteins within caveolar membranes. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-1 is also involved in the co-stimulatory signal essential for T-cell receptor (TCR) mediated T-cell activation. It can bind with DPP4 and induces T-cell proliferation and NF-kappa-B activation in a T-cell receptor/CD3-dependent manner. Mutations affecting the CAV1 gene can result in congenital generalized lipodystrophy 3, pulmonary hypertension primary 3, or partial lipodystrophy/congenital cataracts, and neurodegeneration syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: caveolae; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-like; cell growth-inhibiting protein 32; GIG32; HGNC:1527; OTTHUMP00000196475; unnamed protein product

          View more View less

          Gene Aliases: BSCL3; Cav; Cav-1; CGL3; LCCNS; MSTP085; PPH3; VIP21

          View more View less

          UniProt ID: (Human) Q03135, (Mouse) P49817

          View more View less

          Entrez Gene ID: (Human) 857, (Rat) 25404, (Mouse) 12389

          View more View less

          Function(s)
          receptor binding patched binding protein binding oxysterol binding cholesterol binding peptidase activator activity enzyme binding protein kinase binding protein tyrosine kinase inhibitor activity protein binding, bridging small GTPase binding nitric-oxide synthase inhibitor activity identical protein binding ion channel binding macromolecular complex binding protein heterodimerization activity nitric-oxide synthase binding ATPase binding binding, bridging inward rectifier potassium channel inhibitor activity protein sequestering activity SNARE binding kinase binding receptor serine/threonine kinase binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter angiogenesis vasculogenesis response to hypoxia negative regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation negative regulation of cytokine-mediated signaling pathway regulation of the force of heart contraction response to ischemia regulation of the force of heart contraction by chemical signal triglyceride metabolic process calcium ion transport cellular calcium ion homeostasis regulation of smooth muscle contraction skeletal muscle tissue development response to nutrient lactation protein localization negative regulation of cell proliferation cellular response to starvation response to xenobiotic stimulus response to mechanical stimulus response to bacterium positive regulation of signal transduction negative regulation of signal transduction response to gamma radiation positive regulation of calcium ion transport into cytosol posttranscriptional regulation of gene expression positive regulation of gene expression negative regulation of muscle cell apoptotic process positive regulation of cholesterol efflux negative regulation of smooth muscle cell migration protein transport vesicle organization receptor-mediated endocytosis of virus by host cell regulation of fatty acid metabolic process lipid storage cell differentiation regulation of blood coagulation positive regulation of cell migration negative regulation of BMP signaling pathway negative regulation of epithelial cell differentiation mammary gland development organ regeneration positive regulation of microtubule polymerization T cell costimulation negative regulation of protein ubiquitination positive regulation of protein ubiquitination receptor internalization maintenance of protein location in cell response to progesterone cholesterol efflux nitric oxide homeostasis insulin receptor internalization angiotensin-activated signaling pathway vasoconstriction negative regulation of tyrosine phosphorylation of STAT protein cholesterol homeostasis positive regulation of catalytic activity positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of MAP kinase activity negative regulation of MAPK cascade response to estrogen protein localization to plasma membrane raft negative regulation of nitric oxide biosynthetic process negative regulation of neuron differentiation positive regulation of endocytosis positive regulation of vasoconstriction negative regulation of JAK-STAT cascade microtubule polymerization negative regulation of pinocytosis negative regulation of smooth muscle cell proliferation negative regulation of nitric-oxide synthase activity response to glucocorticoid regulation of cytosolic calcium ion concentration response to calcium ion membrane depolarization regulation of peptidase activity calcium ion homeostasis mammary gland involution canonical Wnt signaling pathway negative regulation of necroptotic process negative regulation of ERK1 and ERK2 cascade caveola assembly cellular response to misfolded protein cellular response to mechanical stimulus cellular response to peptide hormone stimulus cellular response to hyperoxia cellular response to hypoxia cellular response to transforming growth factor beta stimulus basement membrane organization caveolin-mediated endocytosis regulation of heart rate by cardiac conduction negative regulation of canonical Wnt signaling pathway cellular senescence apoptotic signaling pathway regulation of membrane repolarization during action potential regulation of cardiac muscle cell action potential involved in regulation of contraction regulation of ventricular cardiac muscle cell action potential positive regulation of cold-induced thermogenesis regulation of ruffle assembly negative regulation of potassium ion transmembrane transport regulation of cell communication by electrical coupling involved in cardiac conduction protein localization to basolateral plasma membrane positive regulation of gap junction assembly regulation of potassium ion export across plasma membrane positive regulation of ERAD pathway negative regulation of vascular smooth muscle cell proliferation negative regulation of calcium ion import across plasma membrane regulation of entry of bacterium into host cell negative regulation of anoikis positive regulation of extrinsic apoptotic signaling pathway positive regulation of intrinsic apoptotic signaling pathway cholesterol transport nitric oxide metabolic process endocytosis toll-like receptor 4 signaling pathway regulation of toll-like receptor 4 signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nv6df:80/100.66.78.131:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline