Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • IGF1R beta Antibodies

          Invitrogen

          Phospho-IGF1R beta (Tyr1165, Tyr1166) Polyclonal Antibody

          4 References
          View all (22) IGF1R beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-IGF1R beta (Tyr1165, Tyr1166) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Phospho-IGF1R beta (Tyr1165, Tyr1166) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-IGF1R beta (Tyr1165, Tyr1166) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-IGF1R beta (Tyr1165, Tyr1166) Antibody (PA5-37602) in IHC (P)

          Immunohistochemistry analysis of Phospho-IGF1R beta (Tyr1165, Tyr1166) in human colorectal cancer formalin fixed paraffin embedded tissue section. Samples were incubated with Phospho-IGF1R beta (Tyr1165, Tyr1166) polyclonal antibody (Product #PA5-37602). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-IGF1R beta (Tyr1165, Tyr1166) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-IGF1R beta (Tyr1165, Tyr1166) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-IGF1R beta (Tyr1165, Tyr1166) Polyclonal Antibody

          Product Details

          PA5-37602

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:100
          -

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide sequence around phosphorylation site of tyrosine 1165/tyrosine 1166 (T-D-Y(p)-Y(p)-R-K) derived from Human IGF-1R.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2554210

          Product Specific Information

          A suggested positive control for Western blot is 293 cells; suggested positive control for ICC/IF is MCF-7 cells.

          Target Information

          IGF-1R (Insulin-like Growth Factor-1 Receptor) consists of alpha- and beta-subunits, which are disulfide-linked in a beta-alpha-alpha-beta configuration in the mature receptor. The alpha-subunit is completely extracellular, while the beta-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: IGF-I receptor; insulin-like growth factor I receptor; insulin-like growth factor-I receptor precursor; line 186; protein-tyrosine kinase (EC 2.7.1.112); type-I IGF receptor; unnamed protein product

          View more View less

          Gene Aliases: A330103N21Rik; CD221; D930020L01; hyft; IGF-1 receptor; IGF-1R; IGFIR; IGFIRC; IGFR; Igfr1; JTK13

          View more View less

          UniProt ID: (Human) P08069, (Mouse) Q60751, (Rat) P24062

          View more View less

          Entrez Gene ID: (Human) 3480, (Mouse) 16001, (Rat) 25718

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity insulin-activated receptor activity insulin-like growth factor-activated receptor activity insulin receptor binding structural molecule activity protein binding insulin-like growth factor binding ATP binding insulin-like growth factor I binding identical protein binding phosphatidylinositol 3-kinase binding insulin binding insulin receptor substrate binding G-protein alpha-subunit binding macromolecular complex binding protein transporter activity
          Process(es)
          protein phosphorylation immune response transmembrane receptor protein tyrosine kinase signaling pathway axonogenesis brain development positive regulation of cell proliferation insulin receptor signaling pathway epidermis development organ morphogenesis negative regulation of muscle cell apoptotic process cerebellum development establishment of cell polarity male sex determination adrenal gland development positive regulation of cell migration mammary gland development exocrine pancreas development neuron projection development positive regulation of cytokinesis cellular response to insulin stimulus positive regulation of osteoblast proliferation peptidyl-tyrosine autophosphorylation negative regulation of apoptotic process positive regulation of protein complex disassembly negative regulation of MAPK cascade positive regulation of MAPK cascade estrous cycle transcytosis response to ethanol positive regulation of mitotic nuclear division negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated regulation of JNK cascade protein autophosphorylation insulin-like growth factor receptor signaling pathway positive regulation of developmental growth positive regulation of smooth muscle cell proliferation positive regulation of axon regeneration positive regulation of DNA metabolic process positive regulation of meiotic cell cycle positive regulation of protein kinase B signaling negative regulation of protein kinase B signaling prostate gland epithelium morphogenesis cellular response to glucose stimulus cellular response to transforming growth factor beta stimulus positive regulation of steroid hormone biosynthetic process positive regulation of cell cycle process dendritic spine maintenance beta-amyloid clearance postsynaptic modulation of chemical synaptic transmission positive regulation of cold-induced thermogenesis response to L-glutamate negative regulation of hepatocyte apoptotic process negative regulation of cholangiocyte apoptotic process cellular response to beta-amyloid cellular response to insulin-like growth factor stimulus signal transduction protein kinase B signaling cellular response to oxygen-containing compound cardiac atrium development response to hormone hippocampus development response to nutrient levels response to insulin response to vitamin E cellular response to zinc ion starvation response to nicotine response to alkaloid cellular response to mechanical stimulus cellular response to estradiol stimulus cellular response to progesterone stimulus cellular response to testosterone stimulus cellular response to dexamethasone stimulus cellular senescence cellular response to aldosterone cellular response to angiotensin
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0