Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PI3K p85/p55 Antibodies

          Invitrogen

          Phospho-PI3K p85/p55 (Tyr467, Tyr199) Polyclonal Antibody

          View all (13) PI3K p85/p55 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-PI3K p85/p55 (Tyr467, Tyr199) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Phospho-PI3K p85/p55 (Tyr467, Tyr199) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PI3K p85/p55 (Tyr467, Tyr199) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PI3K p85/p55 (Tyr467, Tyr199) Antibody (PA5-37820) in WB

          Western Blot analysis of Phospho-PI3K p85/p55 (Tyr467, Tyr199) was performed using extracts from 293 cells. The membrane was probed with Phospho-PI3K p85/p55 (Tyr467, Tyr199) Polyclonal Antibody (Product # PA5-37820). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PI3K p85/p55 (Tyr467, Tyr199) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-PI3K p85/p55 (Tyr467, Tyr199) Polyclonal Antibody

          Product Details

          PA5-37820

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Peptide sequence around phosphorylation site of tyrosine 467 (L-Y(p)-E-E-Y) derived from Human PI3 Kinase p85/p55.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2554428

          Product Specific Information

          A suggested positive control for Western blot is 293 cells.

          PBS does not contain Mg2+ or Ca2+.

          Target Information

          Phosphoinositide 3-kinase (PI3K, phospho PI3K) Binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phospho PI3K is necessary for the insulin-stimulated increase in glucose uptake and glycogen synthesis in insulin-sensitive tissues. During insulin stimulation, it also binds to IRS-1. Growth factors and hormones trigger this phosphorylation event, which in turn coordinates cell growth, cell cycle entry, cell migration, and cell survival.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: growth factor receptor bound 1; OTTHUMP00000009783; OTTHUMP00000009784; p55PIK; p85-alpha; GRB1; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 3 (p55); phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase p55 subunit; phosphatidylinositol 3-kinase p85 alpha; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphatidylinositol 3-kinase-associated p-85 alpha; Phosphoinositide 3-kinase p85 (other splicing variants: p55 and p50); Phosphoinositide 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3 kinase-associated p85; PI3-kinase p85 subunit alpha; PI3-kinase p85 subunit beta; PI3-kinase p85 subunit gamma; PI3-kinase regulatory subunit alpha; PI3-kinase regulatory subunit beta; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; PI3-kinase subunit p85-alpha; PI3-kinase subunit p85-beta; PI3K regulatory subunit alpha; PI3K regulatory subunit beta; PI3K regulatory subunit gamma; PI3K-p46; polypeptide 3; ptdIns-3-kinase p85-alpha; ptdIns-3-kinase p85-beta; ptdIns-3-kinase p85-gamma; ptdIns-3-kinase regulatory subunit alpha; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; PtdIns-3-kinase regulatory subunit p85-al; ptdIns-3-kinase regulatory subunit p85-alpha; ptdIns-3-kinase regulatory subunit p85-beta; RP4-533D7.2; unnamed protein product

          View more View less

          Gene Aliases: AGM7; GRB1; IMD36; MPPH; MPPH1; p50alpha; p55; p55-GAMMA; p55alpha; p55PIK; p85; p85-ALPHA; p85-BETA; p85alpha; P85B; p85beta; PI3K; PI3KA

          View more View less

          UniProt ID: (Human) P27986, (Human) O00459, (Human) Q92569, (Mouse) P26450, (Rat) Q63787, (Mouse) O08908, (Rat) Q63789, (Mouse) Q64143

          View more View less

          Entrez Gene ID: (Human) 5295, (Human) 5296, (Human) 8503, (Mouse) 18708, (Rat) 25513, (Mouse) 18709, (Rat) 29741, (Rat) 60664, (Mouse) 18710

          View more View less

          Function(s)
          phosphotyrosine binding transmembrane receptor protein tyrosine kinase adaptor activity insulin receptor binding insulin-like growth factor receptor binding neurotrophin TRKA receptor binding protein binding kinase regulator activity kinase activator activity protein phosphatase binding phosphatidylinositol 3-kinase regulator activity phosphatidylinositol 3-kinase regulatory subunit binding ErbB-3 class receptor binding phosphatidylinositol 3-kinase binding insulin binding insulin receptor substrate binding 1-phosphatidylinositol-3-kinase regulator activity protein heterodimerization activity phosphatidylinositol kinase activity enzyme-substrate adaptor activity phosphatidylinositol 3-kinase activator activity receptor tyrosine kinase binding 1-phosphatidylinositol-3-kinase activity receptor binding platelet-derived growth factor receptor binding calmodulin binding protein kinase binding protein domain specific binding estrogen receptor binding ubiquitin protein ligase binding ATPase binding phosphoprotein binding
          Process(es)
          cell migration involved in sprouting angiogenesis insulin receptor signaling pathway positive regulation of gene expression protein kinase B signaling cellular glucose homeostasis positive regulation of protein phosphorylation negative regulation of cell-matrix adhesion glucose metabolic process protein phosphorylation protein import into nucleus negative regulation of cell adhesion signal transduction extrinsic apoptotic signaling pathway via death domain receptors intrinsic apoptotic signaling pathway in response to DNA damage negative regulation of heart rate positive regulation of lamellipodium assembly negative regulation of muscle cell apoptotic process cytokine-mediated signaling pathway negative regulation of cell-cell adhesion B cell differentiation positive regulation of cell migration positive regulation of tumor necrosis factor production response to insulin cellular response to insulin stimulus positive regulation of RNA splicing regulation of toll-like receptor 4 signaling pathway cellular response to UV response to endoplasmic reticulum stress interleukin-18-mediated signaling pathway phosphatidylinositol-3-phosphate biosynthetic process natural killer cell mediated cytotoxicity positive regulation of protein import into nucleus negative regulation of apoptotic process positive regulation of cellular component biogenesis positive regulation of myoblast differentiation negative regulation of osteoclast differentiation negative regulation of blood pressure negative regulation of proteolysis positive regulation of transcription from RNA polymerase II promoter phosphatidylinositol metabolic process phosphatidylinositol phosphorylation insulin-like growth factor receptor signaling pathway positive regulation of smooth muscle cell proliferation negative regulation of smooth muscle cell proliferation protein stabilization positive regulation of cellular component organization negative regulation of multicellular organismal process response to glucocorticoid positive regulation of filopodium assembly regulation of stress fiber assembly negative regulation of stress fiber assembly response to cAMP positive regulation of synapse assembly growth hormone receptor signaling pathway T follicular helper cell differentiation positive regulation of focal adhesion disassembly positive regulation of endoplasmic reticulum unfolded protein response regulation of protein localization to plasma membrane protein polyubiquitination immune response T cell differentiation substrate adhesion-dependent cell spreading positive regulation of glucose import positive regulation of protein localization to plasma membrane regulation of autophagy negative regulation of MAPK cascade regulation of actin filament polymerization positive regulation of cell adhesion negative regulation of cell migration involved in sprouting angiogenesis response to yeast response to nutrient response to xenobiotic stimulus response to fructose response to iron(II) ion response to estradiol response to progesterone response to testosterone response to amino acid response to ethanol response to fatty acid response to growth factor cellular response to fatty acid response to dexamethasone regulation of locomotion negative regulation of anoikis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline