Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TSKU Antibodies

          Invitrogen

          TSKU Polyclonal Antibody

          Advanced Verification
          View all (7) TSKU antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite TSKU Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (8)
          • Advanced Verification (1)
          TSKU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TSKU Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TSKU Antibody (PA5-52523) in ICC/IF

          Immunofluorescent staining of TSKU in human cell line A549 using a TSKU Polyclonal Antibody (Product # PA5-52523) shows localization to nucleus. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TSKU Antibody in Immunocytochemistry (ICC/IF)
          TSKU Antibody in Immunohistochemistry (IHC)
          TSKU Antibody in Immunohistochemistry (IHC)
          TSKU Antibody in Immunohistochemistry (IHC)
          TSKU Antibody in Immunohistochemistry (IHC)
          TSKU Antibody in Western Blot (WB)
          TSKU Antibody in Western Blot (WB)
          TSKU Antibody in Western Blot (WB)
          TSKU Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          TSKU Polyclonal Antibody

          Product Details

          PA5-52523

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.04-0.4 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          0.25-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to Human TSKU. Recombinant protein control fragment (Product #RP-89879).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          10 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 40% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2648998

          Product Specific Information

          Immunogen sequence: DTAHLDLSSN RLEMVNESVL AGPGYTTLAG LDLSHNLLTS ISPTAFSRLR YLESLDLSHN GLTALPAESF TSSPLSDVNL SHNQLREVSV SAFTTHSQGR ALHVDLSHNL IHR

          Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

          Target Information

          The TSKU gene encodes Tsukushi, a member of the small leucine-rich proteoglycan (SLRP) family, playing crucial roles in cellular signaling, growth, and development. Tsukushi is characterized by its involvement in various biological processes, including immune regulation, tissue remodeling, and metabolic homeostasis. It is a hepatokine released in response to conditions like nonalcoholic fatty liver disease (NAFLD), influencing metabolic traits and contributing to conditions such as dyslipidemia and cardiovascular diseases. Tsukushi impacts energy expenditure by modulating brown fat thermogenesis, thereby affecting metabolic homeostasis and obesity-related metabolic traits. As an extracellular matrix component, Tsukushi participates in signaling pathways that regulate cell proliferation, migration, and differentiation, impacting organ development and pathophysiological processes. Its role in metabolic and inflammatory diseases presents it as a potential biomarker and therapeutic target in conditions characterized by altered metabolic and inflammatory states. Ongoing research further elucidates Tsukushi's mechanisms and interactions, contributing to our understanding of its multifunctional roles in health and disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E2-induced gene 4 protein; early insulin-induced hepatic gene protein; hepatic protein EIIH; leucine rich repeat containing 54; leucine-rich repeat-containing protein 54; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushin; unnamed protein product

          View more View less

          Gene Aliases: 9530051K01Rik; E2IG4; Eiih; LRRC54; TSK

          View more View less

          UniProt ID: (Human) Q8WUA8, (Mouse) Q8CBR6, (Rat) Q6QMY6

          View more View less

          Entrez Gene ID: (Human) 25987, (Mouse) 244152, (Rat) 308843

          View more View less

          Function(s)
          cytokine binding transforming growth factor beta binding protein binding signaling receptor activity
          Process(es)
          cholesterol metabolic process anatomical structure morphogenesis regulation of gene expression negative regulation of neuron projection development corpus callosum morphogenesis lateral ventricle development hippocampus development anterior commissure morphogenesis negative regulation of Wnt signaling pathway negative regulation of transforming growth factor beta1 production cholesterol efflux wound healing cholesterol homeostasis positive regulation of hair cycle camera-type eye development inner ear receptor stereocilium organization ciliary body morphogenesis energy homeostasis negative regulation of myofibroblast differentiation growth plate cartilage chondrocyte development bone growth
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-5rqsh:80/100.66.74.151:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0