Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EphB Pan Antibodies

          Invitrogen

          Phospho-EphB Pan (Tyr600, Tyr602, Tyr614, Tyr596) Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-EphB Pan (Tyr600, Tyr602, Tyr614, Tyr596) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          Phospho-EphB Pan (Tyr600, Tyr602, Tyr614, Tyr596) Polyclonal Antibody

          Product Details

          PA5-64578

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human EPHB1/2/3/4 (Phospho-Tyr600/602/614/596)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol, 0.5% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2662980

          Product Specific Information

          Phospho-EphB Pan (Tyr600, Tyr602, Tyr614, Tyr596) Polyclonal Antibody detects endogenous levels of EphB Pan only when phosphorylated at Tyr600, Tyr602, Tyr614, Tyr596.

          Target Information

          Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: developmental kinase 2; developmental kinase 5; developmentally-regulated Eph-related tyrosine kinase; EK6; ELK-related protein tyrosine kinase; elk-related tyrosine kinase; embryonic kinase 2; Eph receptor B2 (ELK-related protein tyrosine kinase); eph tyrosine kinase 2; eph tyrosine kinase 3; EPH-like kinase 5; EPH-like kinase 6; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin B4; ephrin receptor EphB4; ephrin type-B receptor 2-like; EphrinB1; EphrinB2; EphrinB3; EphrinB4; HEK-6; hepatoma transmembrane kinase; human embryo kinase 2; mDK-2; membrane protein; MGC87492; neural kinase; neuronally-expressed EPH-related tyrosine kinase; nuk receptor tyrosine kinase; protein-tyrosine kinase HEK5; renal carcinoma antigen NY-REN-47; Sek-3; Sek-4; tyrosine kinase MYK-1; tyrosine-protein kinase receptor EPH-2; tyrosine-protein kinase receptor EPH-3; tyrosine-protein kinase receptor HTK; tyrosine-protein kinase receptor SEK-3; tyrosine-protein kinase receptor SEK-4; tyrosine-protein kinase TYRO11; tyrosine-protein kinase TYRO5; tyrosine-protein kinase TYRO6; unnamed protein product

          View more View less

          Gene Aliases: 9330129L11; b2b2412Clo; BDPLT22; C130099E04Rik; CAPB; Cek10; Cek5; Cek6; CMAVM2; DRT; EK2; EK5; ELK; Elkh; Ephb2; EPHT2; EPHT3; ERK; ETECK; ETK2; HEK2; Hek5; Hek6; HFASD; HTK; LMPHM7; mDK-5; MDK2; MDK5; MYK1; NET; Nuk; PCBC; Prkm5; Qek5; RGD1564232; Sek3; Sek4; TYRO11; Tyro5; TYRO6

          View more View less

          UniProt ID: (Human) P54762, (Human) P29323, (Human) P54753, (Human) P54760, (Mouse) Q8CBF3, (Rat) P09759, (Mouse) P54763, (Mouse) P54754, (Mouse) P54761

          View more View less

          Entrez Gene ID: (Human) 2047, (Human) 2048, (Human) 2049, (Human) 2050, (Mouse) 270190, (Rat) 24338, (Mouse) 13844, (Rat) 313633, (Rat) 287989, (Mouse) 13845, (Mouse) 13846, (Rat) 686310

          View more View less

          Function(s)
          protein kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity ephrin receptor activity transmembrane-ephrin receptor activity protein binding ATP binding axon guidance receptor activity macromolecular complex binding beta-amyloid binding receptor binding signaling receptor activity identical protein binding nucleotide binding
          Process(es)
          angiogenesis urogenital system development positive regulation of immunoglobulin production negative regulation of cell adhesion transmembrane receptor protein tyrosine kinase signaling pathway nervous system development axon guidance axonal fasciculation learning or memory learning positive regulation of gene expression phosphorylation peptidyl-tyrosine phosphorylation hindbrain tangential cell migration corpus callosum development regulation of blood coagulation positive regulation of cell migration positive regulation of B cell proliferation positive regulation of synaptic plasticity positive regulation of tumor necrosis factor production B cell activation inner ear morphogenesis regulation of JAK-STAT cascade negative regulation of Ras protein signal transduction ephrin receptor signaling pathway regulation of neuronal synaptic plasticity regulation of body fluid levels positive regulation of multicellular organismal process regulation of filopodium assembly regulation of synapse assembly positive regulation of synapse assembly palate development dendritic spine development dendritic spine morphogenesis positive regulation of dendritic spine morphogenesis negative regulation of ERK1 and ERK2 cascade cellular response to lipopolysaccharide commissural neuron axon guidance postsynaptic membrane assembly neuron projection retraction vesicle-mediated intercellular transport tight junction assembly negative regulation of cytokine production involved in inflammatory response positive regulation of long-term synaptic potentiation negative regulation of glutamate receptor signaling pathway positive regulation of glutamate receptor signaling pathway positive regulation of protein localization to plasma membrane cellular response to beta-amyloid neuron projection maintenance positive regulation of protein localization to cell surface regulation of T-helper 17 type immune response regulation of behavioral fear response cell migration involved in sprouting angiogenesis heart morphogenesis cell adhesion animal organ development venous blood vessel morphogenesis regulation of developmental process positive regulation of aorta morphogenesis positive regulation of T cell costimulation immunological synapse formation cell activation skeletal muscle satellite cell activation cell migration cranial nerve development optic nerve morphogenesis central nervous system projection neuron axonogenesis neurogenesis establishment of cell polarity retinal ganglion cell axon guidance cell-substrate adhesion regulation of JNK cascade protein autophosphorylation camera-type eye morphogenesis modulation of synaptic transmission detection of temperature stimulus involved in sensory perception of pain cell chemotaxis neural precursor cell proliferation regulation of ERK1 and ERK2 cascade negative regulation of skeletal muscle satellite cell proliferation negative regulation of satellite cell differentiation central nervous system development regulation of cell-cell adhesion neuron projection development substrate adhesion-dependent cell spreading thymus development digestive tract morphogenesis regulation of axonogenesis cell morphogenesis protein phosphorylation organ morphogenesis positive regulation of long-term neuronal synaptic plasticity negative regulation of axonogenesis regulation of autophagosome assembly system development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch