Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GRK1/GRK2 Antibodies

          Invitrogen

          Phospho-GRK1/GRK2 (Tyr13) Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-GRK1/GRK2 (Tyr13) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          Phospho-GRK1/GRK2 (Tyr13) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-GRK1/GRK2 (Tyr13) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-GRK1/GRK2 (Tyr13) Antibody (PA5-64755) in WB

          Western Blot analysis of Phospho-GRK1/GRK2 (Tyr13) was performed using lysates of (1) Jurkat cell and (2) Jurkat cell treated with 100 ng/mL LPS for 30 min. The membrane was probed with Phospho-GRK1/GRK2 (Tyr13) Polyclonal Antibody (Product # PA5-64755) using a dilution of 1:1,000. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-GRK1/GRK2 (Tyr13) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-GRK1/GRK2 (Tyr13) Polyclonal Antibody

          Product Details

          PA5-64755

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human GRK1/2 (Phospho-Tyr13)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2665189

          Product Specific Information

          Phospho-GRK1/GRK2 (Tyr13) Polyclonal Antibody detects endogenous levels of GRK1/GRK2 only when phosphorylated at Tyr13.

          Target Information

          The sensation of sight is the result of a cascade of events starting with the interaction of photoactivated rhodopsin with a protein called transducin. Rhodopsin kinase is a G-protein-coupled Ser/Thr kinase, also known as GRK1, which is a key element in the regulation of this cascade. Following phosphorylation by GRK1, arrestin is recruited to phospho-rhodopsin quenching its phototransductive activity by preventing further interaction with transducin. By breaking the cycle of phototransduction, GRK1 plays an important role in the restoration of the system for subsequent visual events.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: adrenergic beta receptor kinase 1; adrenergic receptor kinase, beta 1; adrenergic, beta, receptor kinase 1; beta-adrenergic receptor kinase 1; beta-adrenergic receptor kinase 1 beta ARK1; beta-AR kinase-1; beta-ARK-1; G protein-coupled receptpr kinase 1; G-protein receptor kinase 1; grk 2; GRK2; RK; unnamed protein product

          View more View less

          Gene Aliases: Adrbk-1; ADRBK1; Bark-1; BARK1; beta ARK; BETA-ARK1; betaARK1; GPRK1; GRK-2; RHOK; RK

          View more View less

          UniProt ID: (Human) Q15835, (Human) P25098, (Rat) Q63651, (Mouse) Q99MK8, (Rat) P26817

          View more View less

          Entrez Gene ID: (Human) 6011, (Human) 156, (Mouse) 24013, (Rat) 81760, (Mouse) 110355, (Rat) 25238

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity G-protein coupled receptor kinase activity ATP binding rhodopsin kinase activity G-protein coupled receptor binding protein binding alpha-2A adrenergic receptor binding Edg-2 lysophosphatidic acid receptor binding beta-adrenergic receptor kinase activity ubiquitin protein ligase binding delta-type opioid receptor binding kappa-type opioid receptor binding scaffold protein binding
          Process(es)
          regulation of the force of heart contraction desensitization of G-protein coupled receptor protein signaling pathway negative regulation of the force of heart contraction by chemical signal protein phosphorylation response to oxidative stress signal transduction G-protein coupled receptor signaling pathway tachykinin receptor signaling pathway heart development positive regulation of muscle cell apoptotic process negative regulation of peptidyl-threonine phosphorylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation viral genome replication receptor internalization positive regulation of interleukin-6 production regulation of inositol trisphosphate biosynthetic process negative regulation of peptidyl-serine phosphorylation positive regulation of catecholamine secretion intraciliary retrograde transport vasodilation follicle-stimulating hormone signaling pathway negative regulation of apoptotic process negative regulation of G-protein coupled receptor protein signaling pathway positive regulation of smoothened signaling pathway positive regulation of vasoconstriction negative regulation of striated muscle contraction negative regulation of glucose import viral entry into host cell positive regulation of fibroblast proliferation oligodendrocyte differentiation maintenance of protein location in nucleus cardiac muscle contraction positive regulation of ERK1 and ERK2 cascade cellular response to glucose stimulus cellular response to epidermal growth factor stimulus negative regulation of adenylate cyclase-activating G protein-coupled receptor signaling pathway negative regulation of cellular response to insulin stimulus negative regulation of relaxation of smooth muscle negative regulation of protein localization to ciliary membrane positive regulation of vascular smooth muscle cell proliferation positive regulation of intraciliary retrograde transport cellular response to chemokine positive regulation of excitatory postsynaptic potential negative regulation of apoptotic signaling pathway visual perception photoreceptor cell morphogenesis response to light stimulus regulation of signal transduction regulation of rhodopsin mediated signaling pathway positive regulation of phosphorylation protein autophosphorylation post-embryonic retina morphogenesis in camera-type eye G-protein coupled acetylcholine receptor signaling pathway regulation of G-protein coupled receptor protein signaling pathway rhodopsin mediated signaling pathway response to xenobiotic stimulus response to muscle activity response to hydrogen peroxide response to leptin response to caloric restriction cellular response to catecholamine stimulus cellular response to rapamycin cellular response to phorbol 13-acetate 12-myristate
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch